Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
7141 - 7170 of 48881 products displayed
  • APC7 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520948
  • Size 100ug
Applications
Immunofluorescence (20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Cyclosome subunit 7; Prediabetic NOD sera-reactive autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520948 349.00
Check your price
  • Apelin Receptor Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521285
  • Size 100ug
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
G-protein coupled receptor HG11; G-protein coupled receptor APJ; Angiotensin receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521285 349.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Marsupial, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523466
  • Size 100ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Presenilin-stabilization factor; Aph-1alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523466 349.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518739
  • Size 100ug
Applications
Western Blot (0.05-0.1 ug/ml)
Alternate names:
Presenilin-stabilization factor; Aph-1alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518739 349.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520318
  • Size 100ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Aph-1alpha; Presenilin-stabilization factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520318 349.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520317
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Aph-1alpha; Presenilin-stabilization factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520317 349.00
Check your price
  • APOA1BP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518975
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
YjeF N-terminal domain-containing protein 1; NAD(P)HX epimerase; Apolipoprotein A-I-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518975 349.00
Check your price
  • APOBEC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519014
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
mRNA(cytosine(6666)) deaminase 1; Apolipoprotein B mRNA-editing enzyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519014 349.00
Check your price
  • APOBEC2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518910
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
mRNA(cytosine(6666)) deaminase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518910 349.00
Check your price
  • APOBEC2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518887
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
mRNA(cytosine(6666)) deaminase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518887 349.00
Check your price
  • APOBEC3C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518873
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Phorbolin I; APOBEC1-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518873 349.00
Check your price
  • APOBEC3G Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520052
  • Size 100ug
Applications
Immunohistochemistry (1 µg/ml), Western Blot (5 ug/ml)
Alternate names:
Deoxycytidine deaminase; CEM-15; APOBEC-related protein 9; APOBEC-related cytidine deaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520052 349.00
Check your price
  • APOE Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518361
  • Size 100ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
APOE; AD2; MGC1571; apoprotein; apolipoprotein E; apolipoprotein E3; Alzheimer disease 2 (APOE*E4-associated, late onset); LPG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518361 349.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514668
  • Size 100ug
Applications
ELISA (1µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514668 349.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514670
  • Size 100ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514670 349.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514732
  • Size 100ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514732 349.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514667
  • Size 100ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514667 349.00
Check your price
  • Apolipoprotein A4 Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518637
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
APOA4; apolipoprotein A-IV ; MGC142154; MGC142156
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518637 349.00
Check your price
  • Apolipoprotein A4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # PA521165
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
apo-AIV; apoA-IV; apolipoprotein A4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521165 349.00
Check your price
  • Apolipoprotein A4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # PA521167
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
apo-AIV; apoA-IV; apolipoprotein A4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521167 349.00
Check your price
  • Apolipoprotein A5 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514759
  • Size 100ug
Applications
Western Blot (1/500-1/2000)
Alternate names:
Regeneration-associated protein 3; Apolipoprotein A5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514759 349.00
Check your price
  • Apolipoprotein A5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518757
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Regeneration-associated protein 3; Apolipoprotein A5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518757 349.00
Check your price
  • Apolipoprotein A5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # PA521168
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Regeneration-associated protein 3; Apolipoprotein A5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521168 349.00
Check your price
  • Apolipoprotein B Monoclonal Antibody for IF, ICC, Flow, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514741
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
apo B-100, apoB-100, apoB-48, apolipoprotein B-100, apolipoprotein B49
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514741 349.00
Check your price
  • Apolipoprotein B Monoclonal Antibody for IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514724
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
apo B-100, apoB-100, apoB-48, apolipoprotein B-100, apolipoprotein B46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514724 349.00
Check your price
  • Apolipoprotein B Monoclonal Antibody for IF, ICC, IHC, Flow, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514740
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
apo B-100, apoB-100, apoB-48, apolipoprotein B-100, apolipoprotein B47
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514740 349.00
Check your price
  • Apolipoprotein B Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514671
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
apo B-100, apoB-100, apoB-48, apolipoprotein B-100, apolipoprotein B48
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514671 349.00
Check your price
  • Apolipoprotein B Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518986
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
APOB; apolipoprotein B (including Ag(x) antigen); FLDB; apoB-100; apoB-48; apolipoprotein B; apolipoprotein B48
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518986 349.00
Check your price
  • Apolipoprotein C1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518706
  • Size 100ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Apolipoprotein C1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518706 349.00
Check your price
  • Apolipoprotein F Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518977
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2.5-3.8 µg/mL), Western Blot (0.3-1 ug/ml)
Alternate names:
Lipid transfer inhibitor protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518977 349.00
Check your price