Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  260  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1605 
7231 - 7260 of 48136 products displayed
  • AMPK alpha-2 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519367
  • Size 100 ug
Applications
ELISA (1.5-3.0 ug/ml), Western Blot (0.3-1.0 ug/ml)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519367 349.00
Check your price
  • Angiogenin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534422
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Ribonuclease 5; Angiogenin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534422 349.00
Check your price
  • Anillin Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518199
  • Size 100 ug
Applications
Immunohistochemistry (3-10µg/ml)
Alternate names:
ANLN; Scraps; ANILLIN; anillin, actin binding protein (scraps homolog, Drosophila); anillin (Drosophila Scraps homolog), actin binding protein; anillin, actin binding protein; DKFZp779A055; scra
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518199 349.00
Check your price
  • Anillin Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518540
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
ANILLIN; Scraps; ANLN; anillin, actin binding protein (scraps homolog, Drosophila) ; DKFZp779A055; anillin (Drosophila Scraps homolog), actin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518540 349.00
Check your price
  • ANKK1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518997
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
X-kinase; Sugen kinase 288; Protein kinase PKK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518997 349.00
Check your price
  • Annexin A1 Antibody
  • Host Goat
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA519084
  • Size 100 ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519084 349.00
Check your price
  • Annexin A10 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518004
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Annexin-14; Annexin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518004 349.00
Check your price
  • Annexin A11 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518003
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Calcyclin-associated annexin 50; Annexin-11; Annexin XI; 56 kDa autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518003 349.00
Check your price
  • Annexin A2 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19006
  • Size 100 ug
Applications
Western Blot (0.5-3 µg/ml)
Alternate names:
p36; Protein I; Placental anticoagulant protein IV; Lipocortin II; Chromobindin-8; Calpactin-1 heavy chain; Calpactin I heavy chain; Annexin-2; Annexin II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19006 349.00
Check your price
  • Anosmin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521149
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Kallmann syndrome protein; Adhesion molecule-like X-linked
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521149 349.00
Check your price
  • AOC3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518638
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Vascular adhesion protein 1; Semicarbazide-sensitive amine oxidase; HPAO; Copper amine oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518638 349.00
Check your price
  • AP2 gamma Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518346
  • Size 100 ug
Applications
Immunohistochemistry (2-3µg/ml)
Alternate names:
Transcription factor ERF-1; Activating enhancer-binding protein 2 gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518346 349.00
Check your price
  • AP2M1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520745
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Plasma membrane adaptor AP-2 50 kDa protein; HA2 50 kDa subunit; Clathrin coat-associated protein AP50; Clathrin coat assembly protein AP50; Clathrin assembly protein complex 2 mu medium chain; Adaptor-related protein complex 2 subunit mu; Adaptor protein complex AP-2 subunit mu; Adaptin-mu2; AP-2 mu chain; Mu2-adaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520745 349.00
Check your price
  • AP3B2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521163
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Neuron-specific vesicle coat protein beta-NAP; Clathrin assembly protein complex 3 beta-2 large chain; Beta-3B-adaptin; Adaptor-related protein complex 3 subunit beta-2; Adaptor protein complex AP-3 subunit beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521163 349.00
Check your price
  • AP3M1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521162
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Mu3A-adaptin; Mu-adaptin 3A; Adaptor-related protein complex 3 subunit mu-1; AP-3 adaptor complex mu3A subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521162 349.00
Check your price
  • AP3S1 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA521161
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Sigma-adaptin 3a; Sigma-3A-adaptin; Adaptor-related protein complex 3 subunit sigma-1; AP-3 complex subunit sigma-3A; Clathrin-associated/assembly/adaptor protein, small 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521161 349.00
Check your price
  • APAF1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519894
  • Size 100 ug
Applications
Immunohistochemistry (20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
APAF-1, CED4; apoptotic protease-activating factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519894 349.00
Check your price
  • APAF1 Antibody (Monoclonal, 2E10)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515743
  • Size 100 ug
Applications
Immunohistochemistry (0.5 µg/ml), Western Blot (0.25-0.5 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515743 349.00
Check your price
  • APAF1 Antibody (Monoclonal, 5E11)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515744
  • Size 100 ug
Applications
Immunohistochemistry (0.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515744 349.00
Check your price
  • APAF1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519893
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (1 ug/ml), Western Blot (1 ug/ml)
Alternate names:
APAF-1, CED4; apoptotic protease-activating factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519893 349.00
Check your price
  • APBA1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518702
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; Adapter protein X11alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518702 349.00
Check your price
  • APBA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520525
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; Adapter protein X11alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520525 349.00
Check your price
  • APBA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534418
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (0.5 ug/mL)
Alternate names:
Neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; Adapter protein X11alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534418 349.00
Check your price
  • APBA2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521053
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Neuronal Munc18-1-interacting protein 2; Neuron-specific X11L protein; Adapter protein X11beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521053 349.00
Check your price
  • APC1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520941
  • Size 100 ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Testis-specific gene 24 protein; Mitotic checkpoint regulator; Cyclosome subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520941 349.00
Check your price
  • APC11 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520955
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Cyclosome subunit 11; Hepatocellular carcinoma-associated RING finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520955 349.00
Check your price
  • APC13 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520956
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Cyclosome subunit 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520956 349.00
Check your price
  • APC2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520944
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Adenomatous polyposis coli protein-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520944 349.00
Check your price
  • APC4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520945
  • Size 100 ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Cyclosome subunit 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520945 349.00
Check your price
  • APC5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520946
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Cyclosome subunit 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520946 349.00
Check your price