Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1630 
7441 - 7470 of 48881 products displayed
  • CARD10 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534364
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
CARD-containing MAGUK protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534364 349.00
Check your price
  • CARD11 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518145
  • Size 100ug
Applications
Immunohistochemistry (2-5µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
CARD-containing MAGUK protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518145 349.00
Check your price
  • CARD11 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520032
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
CARD-containing MAGUK protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520032 349.00
Check your price
  • CARD14 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534363
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
CARD-containing MAGUK protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534363 349.00
Check your price
  • CARD6 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518154
  • Size 100ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
CARD6; caspase recruitment domain family, member 6; caspase recruitment domain protein 6; CINCIN1; CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518154 349.00
Check your price
  • CARD8 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520031
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (2-4 µg/ml)
Alternate names:
Tumor up-regulated CARD-containing antagonist of CASP9; DACAR; CARD-inhibitor of NF-kappa-B-activating ligand; Apoptotic protein NDPP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520031 349.00
Check your price
  • CARD9 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519993
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (2.5 µg/ml)
Alternate names:
RP11-413M3.9, CANDF2, hCARD9; caspase recruitment domain-containing protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519993 349.00
Check your price
  • CASKIN1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534424
  • Size 100ug
Applications
Western Blot (1 ug/mL)
Alternate names:
CASK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534424 349.00
Check your price
  • Caspase 1 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA516215
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (0.5-2 µg/ml)
Alternate names:
p45; Interleukin-1 beta-converting enzyme; Interleukin-1 beta convertase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516215 349.00
Check your price
  • Caspase 1 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520112
  • Size 100ug
Applications
Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
p45; Interleukin-1 beta-converting enzyme; Interleukin-1 beta convertase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520112 349.00
Check your price
  • Caspase 1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520113
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
p45; Interleukin-1 beta-converting enzyme; Interleukin-1 beta convertase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520113 349.00
Check your price
  • Caspase 10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519870
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
ICE-like apoptotic protease 4; FAS-associated death domain protein interleukin-1B-converting enzyme 2; Apoptotic protease Mch-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519870 349.00
Check your price
  • Caspase 12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519962
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519962 349.00
Check your price
  • Caspase 12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523369
  • Size 100ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
UNQ9415, CASP-12, CASP12P1, caspase 12 pseudogene 1, inactive caspase-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523369 349.00
Check your price
  • Caspase 12 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520033
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520033 349.00
Check your price
  • Caspase 12 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519963
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519963 349.00
Check your price
  • Caspase 12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520034
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520034 349.00
Check your price
  • Caspase 13 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519949
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
BOS_14235, CASP13; CASP-13; CASP-4; ERICE; caspase 13; caspase 4, apoptosis-related cysteine protease; caspase-13; caspase-4; evolutionary related interleukin-1-beta-converting enzyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519949 349.00
Check your price
  • Caspase 14 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520297
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Mini-ICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520297 349.00
Check your price
  • Caspase 2 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520107
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protease ICH-1; Neural precursor cell expressed developmentally down-regulated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520107 349.00
Check your price
  • Caspase 3 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534413
  • Size 100ug
Applications
Immunohistochemistry (5 ug/mL)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534413 349.00
Check your price
  • Caspase 4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518300
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Protease TX; Protease ICH-2; ICE(rel)-II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518300 349.00
Check your price
  • Caspase 4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520109
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Protease ICH-3; Caspase-11; Protease TX; Protease ICH-2; ICE(rel)-II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520109 349.00
Check your price
  • Caspase 4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520108
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Protease ICH-3; Caspase-11; Protease TX; Protease ICH-2; ICE(rel)-II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520108 349.00
Check your price
  • Caspase 5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520110
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Protease TY; Protease ICH-3; ICE(rel)-III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520110 349.00
Check your price
  • Caspase 5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520111
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protease TY; Protease ICH-3; ICE(rel)-III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520111 349.00
Check your price
  • Caspase 6 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520117
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Apoptotic protease Mch-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520117 349.00
Check your price
  • Caspase 6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520116
  • Size 100ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Apoptotic protease Mch-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520116 349.00
Check your price
  • Caspase 7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520114
  • Size 100ug
Applications
Immunohistochemistry (2 ug/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520114 349.00
Check your price
  • Caspase 7 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520115
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520115 349.00
Check your price