Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  270  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7531 - 7560 of 48882 products displayed
  • CCR3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519861
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Eosinophil eotaxin receptor; Macrophage inflammatory protein 1-alpha receptor-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519861 349.00
Check your price
  • CCR3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519859
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Eosinophil eotaxin receptor; Macrophage inflammatory protein 1-alpha receptor-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519859 349.00
Check your price
  • CCR5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519862
  • Size 100ug
Applications
Immunohistochemistry (20 ug/ml), Western Blot (1:1000)
Alternate names:
HIV-1 fusion coreceptor; CHEMR13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519862 349.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519911
  • Size 100ug
Applications
Western Blot (0.5 µg/ml)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519911 349.00
Check your price
  • CD10 Monoclonal Antibody for IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515231
  • Size 100ug
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515231 349.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522803
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522803 349.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522804
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522804 349.00
Check your price
  • CD11b Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518727
  • Size 100ug
Applications
Immunohistochemistry (2-4 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518727 349.00
Check your price
  • CD123 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523460
  • Size 100ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra, CD123 antigen, IL-3 receptor alpha SP2 isoform, IL-3 receptor subunit alpha, IL-3R subunit alpha, IL-3R-alpha, IL-3RA, interleukin-3 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523460 349.00
Check your price
  • CD150 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521123
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 ug/ml)
Alternate names:
SLAM family member 1; IPO-3; CDw150
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521123 349.00
Check your price
  • CD180 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520250
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 78; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520250 349.00
Check your price
  • CD180 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520251
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520251 349.00
Check your price
  • CD180 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520252
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 78; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520252 349.00
Check your price
  • CD19 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515741
  • Size 100ug
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
T-cell surface antigen Leu-12; Differentiation antigen CD19; B-lymphocyte surface antigen B4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515741 349.00
Check your price
  • CD225 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520989
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Mouse ifitm-like protein 2; Fragilis protein 2; Dispanin subfamily A member 2a; Leu-13 antigen; Interferon-inducible protein 9-27; Interferon-induced protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520989 349.00
Check your price
  • CD225 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520988
  • Size 100ug
Applications
Western Blot (2.5-5 µg/ml)
Alternate names:
Leu-13 antigen; Interferon-inducible protein 9-27; Interferon-induced protein 17; Dispanin subfamily A member 2a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520988 349.00
Check your price
  • CD229 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521135
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Signaling lymphocytic activation molecule 3; SLAM family member 3; Lymphocyte antigen 9; Cell surface molecule Ly-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521135 349.00
Check your price
  • CD244 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521136
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Signaling lymphocytic activation molecule 4; SLAM family member 4; NK cell type I receptor protein 2B4; NK cell activation-inducing ligand; Non-MHC restricted killing associated
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521136 349.00
Check your price
  • CD248 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520436
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Tumor endothelial marker 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520436 349.00
Check your price
  • CD248 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520437
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Tumor endothelial marker 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520437 349.00
Check your price
  • CD249 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518393
  • Size 100ug
Applications
Immunohistochemistry (3-5µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Differentiation antigen gp160; Aminopeptidase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518393 349.00
Check your price
  • CD26 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518732
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
T-cell activation antigen CD26; GP110 glycoprotein; Dipeptidyl peptidase IV; Bile canaliculus domain-specific membrane glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518732 349.00
Check your price
  • CD263 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110161
  • Size 100ug
Applications
Flow Cytometry (3 ug/ml)
Alternate names:
TRAIL receptor without an intracellular domain; TNF-related apoptosis-inducing ligand receptor 3; Lymphocyte inhibitor of TRAIL; Decoy receptor 1; Decoy TRAIL receptor without death domain; Antagonist decoy receptor for TRAIL/Apo-2L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110161 349.00
Check your price
  • CD263 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110162
  • Size 100ug
Applications
Flow Cytometry (3 ug/ml)
Alternate names:
TRAIL receptor without an intracellular domain; TNF-related apoptosis-inducing ligand receptor 3; Lymphocyte inhibitor of TRAIL; Decoy receptor 1; Decoy TRAIL receptor without death domain; Antagonist decoy receptor for TRAIL/Apo-2L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110162 349.00
Check your price
  • CD263 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519932
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
TRAIL receptor without an intracellular domain; TNF-related apoptosis-inducing ligand receptor 3; Lymphocyte inhibitor of TRAIL; Decoy receptor 1; Decoy TRAIL receptor without death domain; Antagonist decoy receptor for TRAIL/Apo-2L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519932 349.00
Check your price
  • CD263 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519959
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
TRAIL receptor without an intracellular domain; TNF-related apoptosis-inducing ligand receptor 3; Lymphocyte inhibitor of TRAIL; Decoy receptor 1; Decoy TRAIL receptor without death domain; Antagonist decoy receptor for TRAIL/Apo-2L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519959 349.00
Check your price
  • CD264 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110163
  • Size 100ug
Applications
Flow Cytometry (3 ug/ml)
Alternate names:
TRAIL receptor with a truncated death domain; TNF-related apoptosis-inducing ligand receptor 4; Decoy receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110163 349.00
Check your price
  • CD264 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519896
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1:1000)
Alternate names:
TRAIL receptor with a truncated death domain; TNF-related apoptosis-inducing ligand receptor 4; Decoy receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519896 349.00
Check your price
  • CD266 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520275
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Tweak-receptor; Fibroblast growth factor-inducible immediate-early response protein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520275 349.00
Check your price
  • CD2BP2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518286
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 ug/ml), Western Blot (0.3-1.0 ug/ml)
Alternate names:
U5 snRNP 52K protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518286 349.00
Check your price