Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1606 
7771 - 7800 of 48165 products displayed
  • CYTIP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518019
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Pleckstrin homology Sec7 and coiled-coil domains-binding protein; Cytohesin-binding protein HE; Cytohesin-associated scaffolding protein; Cytohesin binder and regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518019 349.00
Check your price
  • Cytohesin 2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA518833
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
SEC7 homolog B; PH, SEC7 and coiled-coil domain-containing protein 2; ARF nucleotide-binding site opener; ARF exchange factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518833 349.00
Check your price
  • Cytokeratin 13 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519049
  • Size 100ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519049 349.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody, APC conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Mammals
  • Conjugate APC
  • Catalog # MA110325
  • Size 100ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • D4-GDI Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519352
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Rho-GDI beta; Ly-GDI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519352 349.00
Check your price
  • DAAM1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523222
  • Size 100ug
Applications
Western Blot (4.0-6 µg/ml)
Alternate names:
disheveled-associated activator of morphogenesis 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523222 349.00
Check your price
  • DAAM2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523201
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
RP1-278E11.1, dJ90A20A.1, disheveled-associated activator of morphogenesis 2, dishevelled-associated activator of morphogenesis 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523201 349.00
Check your price
  • DACH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518909
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
OTTHUMP00000040839; dachshund homolog 1 (Drosophila) ; DACH1; RP11-512J14.1; DACH; FLJ10138; dachshund homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518909 349.00
Check your price
  • DACH1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Human, Mouse, Primate
  • Conjugate Unconjugated
  • Catalog # PA523483
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (5-7 µg/ml)
Alternate names:
RP11-512J14.1, DACH, dachshund homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523483 349.00
Check your price
  • DACH2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518897
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
DACH2; dachshund homolog 2 (Drosophila); RP11-345E19.1; FLJ31391; MGC138545; dachshund 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518897 349.00
Check your price
  • DACT1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Primate, Rat, Clawed frog, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523216
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 ug/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Thymus-expressed novel gene 3 protein; MDpr1; Frodo homolog; Dapper antagonist of catenin 1; Hepatocellular carcinoma novel gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523216 349.00
Check your price
  • DACT2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520629
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Dapper antagonist of catenin 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520629 349.00
Check your price
  • DACT3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520630
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Dapper antagonist of catenin 3; Arginine-rich region 1 protein; Antagonist of beta-catenin Dapper homolog 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520630 349.00
Check your price
  • DAD1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520062
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Defender against cell death 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520062 349.00
Check your price
  • DAGLA Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518985
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Neural stem cell-derived dendrite regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518985 349.00
Check your price
  • DAP Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534564
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Rap7a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534564 349.00
Check your price
  • DAP10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518600
  • Size 100ug
Applications
Western Blot (0.2-0.6 ug/ml)
Alternate names:
Transmembrane adapter protein KAP10; Membrane protein DAP10; DNAX-activation protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518600 349.00
Check your price
  • DAP3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518042
  • Size 100ug
Applications
Immunocytochemistry (3-10 ug/ml), Immunofluorescence (3-10 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Ionizing radiation resistance conferring protein; Death-associated protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518042 349.00
Check your price
  • DAPK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519961
  • Size 100ug
Applications
Immunohistochemistry (2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
DAP-kinase-related protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519961 349.00
Check your price
  • DARC Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518424
  • Size 100ug
Applications
Flow Cytometry (1/1000), Immunohistochemistry (3µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Plasmodium vivax receptor; Glycoprotein D; Fy glycoprotein; Duffy antigen/chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518424 349.00
Check your price
  • DARC Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520357
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Plasmodium vivax receptor; Glycoprotein D; Fy glycoprotein; Duffy antigen/chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520357 349.00
Check your price
  • DARC Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520360
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Plasmodium vivax receptor; Glycoprotein D; Fy glycoprotein; Duffy antigen/chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520360 349.00
Check your price
  • DAZL Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518404
  • Size 100ug
Applications
Immunohistochemistry (1-3µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
SPGY-like-autosomal; Deleted in azoospermia-like 1; DAZ-like autosomal; DAZ homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518404 349.00
Check your price
  • DBX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA534391
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (5 ug/mL), Immunohistochemistry (5 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
Developing brain homeobox protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534391 349.00
Check your price
  • DC-SIGN Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515745
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Dendritic cell-specific ICAM-3-grabbing non-integrin 1; C-type lectin domain family 4 member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515745 349.00
Check your price
  • DC-SIGN Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519967
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Dendritic cell-specific ICAM-3-grabbing non-integrin 1; C-type lectin domain family 4 member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519967 349.00
Check your price
  • DC-SIGN Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519966
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Dendritic cell-specific ICAM-3-grabbing non-integrin 1; C-type lectin domain family 4 member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519966 349.00
Check your price
  • DCAMKL1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520908
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Doublecortin-like kinase 1; Doublecortin-like and CAM kinase-like 1; Doublecortin domain-containing protein 3A; Calcium/calmodulin-dependent protein kinase type I-like CPG16
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520908 349.00
Check your price
  • DCAMKL2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520910
  • Size 100ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Doublecortin-like kinase 2; Doublecortin-like and CAM kinase-like 2; CaMK-like CREB regulatory kinase 2; Doublecortin domain-containing protein 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520910 349.00
Check your price
  • DCBLD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522809
  • Size 100ug
Applications
Western Blot (1:1000)
Alternate names:
Endothelial and smooth muscle cell-derived neuropilin-like protein; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522809 349.00
Check your price