Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  280  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1630 
7831 - 7860 of 48881 products displayed
  • DLL1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523457
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml)
Alternate names:
Drosophila Delta homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523457 349.00
Check your price
  • DLL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519106
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Drosophila Delta homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519106 349.00
Check your price
  • DLL3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523448
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (3-5 µg/ml)
Alternate names:
Drosophila Delta homolog 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523448 349.00
Check your price
  • DLL4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523453
  • Size 100ug
Applications
Western Blot (3-6 µg/ml)
Alternate names:
Drosophila Delta homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523453 349.00
Check your price
  • DLX3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523398
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
AI4, TDO, homeobox protein DLX-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523398 349.00
Check your price
  • DLX5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519105
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
DLX5; distal-less homeobox 5; distal-less homeo box 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519105 349.00
Check your price
  • DMAP1 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523214
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1.0-4 µg/ml)
Alternate names:
MAT1-mediated transcriptional repressor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523214 349.00
Check your price
  • DMC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521283
  • Size 100ug
Applications
Western Blot (1:500-1:3000)
Alternate names:
RP1-199H16.4, DMC1H, HsLim15, LIM15, dJ199H16.1, DMC1 homologue, disrupted meiotic cDNA1, yeast, homolog of, meiotic recombination protein DMC1/LIM15 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521283 349.00
Check your price
  • DMP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519009
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
DMP1; dentin matrix acidic phosphoprotein; dentin matrix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519009 349.00
Check your price
  • DNA Ligase I Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519162
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Polydeoxyribonucleotide synthase [ATP] 1; DNA ligase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519162 349.00
Check your price
  • DNAL1 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520573
  • Size 100ug
Applications
Immunocytochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
C14orf168, CILD16; dynein light chain 1, axonemal
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520573 349.00
Check your price
  • DNase II Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519900
  • Size 100ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
R31240_2; Lysosomal DNase II; Deoxyribonuclease II alpha; Acid DNase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519900 349.00
Check your price
  • DNMT1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (Free), IHC (P), IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA516169
  • Size 100ug
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1:10), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (0.1-0.5 µg/ml)
Alternate names:
MCMT; DNA methyltransferase HsaI; CXXC-type zinc finger protein 9; DNA methyltransferase MmuI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516169 349.00
Check your price
  • DNMT1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518878
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
MCMT; DNA methyltransferase HsaI; CXXC-type zinc finger protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518878 349.00
Check your price
  • Dnmt2 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516197
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
Met-2; DNA methyltransferase homolog MmuIIP; DNA (cytosine-5)-methyltransferase-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516197 349.00
Check your price
  • DNMT3A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516171
  • Size 100ug
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
DNA methyltransferase MmuIIIA; DNA methyltransferase HsaIIIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516171 349.00
Check your price
  • DNMT3B Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516165
  • Size 100ug
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1-2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
DNA methyltransferase MmuIIIB; DNA methyltransferase HsaIIIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516165 349.00
Check your price
  • DOCK1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517969
  • Size 100ug
Applications
Western Blot (3-4 ug/ml)
Alternate names:
180 kDa protein downstream of CRK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517969 349.00
Check your price
  • DOG-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520825
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Tumor-amplified and overexpressed sequence 2; Transmembrane protein 16A; Oral cancer overexpressed protein 2; Discovered on gastrointestinal stromal tumors protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520825 349.00
Check your price
  • DOK1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519860
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
pp62; p62(dok); Downstream of tyrosine kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519860 349.00
Check your price
  • DOK3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517975
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Downstream of tyrosine kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517975 349.00
Check your price
  • DOK5 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518309
  • Size 100ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Insulin receptor substrate 6; Downstream of tyrosine kinase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518309 349.00
Check your price
  • DOPA Decarboxylase Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518373
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
DOPA decarboxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518373 349.00
Check your price
  • DPF2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521079
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein requiem; D4, zinc and double PHD fingers family 2; BRG1-associated factor 45D; Apoptosis response zinc finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521079 349.00
Check your price
  • DPM1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA519028
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
DPM1; dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit; RP23-391M18.1; AI118379; AI194292; OTTMUSP00000017164; dolichol monophosphate mannose synthase; dolichyl-phosphate mannosyltransferase polypeptide 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519028 349.00
Check your price
  • DPM1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519027
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Mannose-P-dolichol synthase subunit 1; Dolichyl-phosphate beta-D-mannosyltransferase subunit 1; Dolichol-phosphate mannose synthase subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519027 349.00
Check your price
  • DPP10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518377
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Dipeptidyl peptidase-like protein 2; Dipeptidyl peptidase X; Dipeptidyl peptidase IV-related protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518377 349.00
Check your price
  • DPPA3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523449
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Stella-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523449 349.00
Check your price
  • DPPA3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520578
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Primordial germ cell protein 7; Compaction-associated protein 1; Stella-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520578 349.00
Check your price
  • DPPA4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523444
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Embryonic stem cell-associated transcript 15-1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523444 349.00
Check your price