Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1622 
7951 - 7980 of 48655 products displayed
  • CTRP4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520150
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
CTRP4, ZACRP4; complement C1q tumor necrosis factor-related protein 4; complement-c1q tumor necrosis factor-related protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520150 349.00
Check your price
  • CTRP4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520151
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
CTRP4, ZACRP4; complement C1q tumor necrosis factor-related protein 4; complement-c1q tumor necrosis factor-related protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520151 349.00
Check your price
  • CTRP5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA520152
  • Size 100 ug
Applications
Immunohistochemistry (10 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
UNQ303/PRO344, CTRP5, LORD; complement C1q tumor necrosis factor-related protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520152 349.00
Check your price
  • CTRP5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520153
  • Size 100 ug
Applications
Western Blot (1-4 µg/ml)
Alternate names:
UNQ303/PRO344, CTRP5, LORD; complement C1q tumor necrosis factor-related protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520153 349.00
Check your price
  • CTRP6 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520155
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
UNQ581/PRO1151, CTRP6, ZACRP6; complement C1q tumor necrosis factor-related protein 6; complement-c1q tumor necrosis factor-related protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520155 349.00
Check your price
  • CTRP6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520154
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
UNQ581/PRO1151, CTRP6, ZACRP6; complement C1q tumor necrosis factor-related protein 6; complement-c1q tumor necrosis factor-related protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520154 349.00
Check your price
  • CTRP7 Polyclonal Antibody for Western Blot, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520157
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunohistochemistry (10 ug/ml), Western Blot (1-4 µg/ml)
Alternate names:
CTRP7, ZACRP7; complement C1q tumor necrosis factor-related protein 7; complement-c1q tumor necrosis factor-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520157 349.00
Check your price
  • CTRP7 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520156
  • Size 100 ug
Applications
Immunocytochemistry (0.5 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
CTRP7, ZACRP7; complement C1q tumor necrosis factor-related protein 7; complement-c1q tumor necrosis factor-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520156 349.00
Check your price
  • CTRP7 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520198
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
CTRP7, ZACRP7; complement C1q tumor necrosis factor-related protein 7; complement-c1q tumor necrosis factor-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520198 349.00
Check your price
  • CUEDC1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520619
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
CUE domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520619 349.00
Check your price
  • CUEDC2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # PA523265
  • Size 100 ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
HOYS6, C10orf66, bA18I14.5, CUE domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523265 349.00
Check your price
  • CUEDC2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523277
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
HOYS6, C10orf66, bA18I14.5, CUE domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523277 349.00
Check your price
  • CUEDC2 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520620
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
HOYS6, C10orf66, bA18I14.5; CUE domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520620 349.00
Check your price
  • Cullin 2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519314
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
CUL2; cullin 2; MGC131970; OTTHUMP00000019437
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519314 349.00
Check your price
  • Cullin 9 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520277
  • Size 100 ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
p53-associated parkin-like cytoplasmic protein; UbcH7-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520277 349.00
Check your price
  • CX3CL1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520296
  • Size 100 ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520296 349.00
Check your price
  • CX3CR1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519910
  • Size 100 ug
Applications
Flow Cytometry (0.1 µg/ml), Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/mL), Immunohistochemistry (2 ug/mL), Western Blot (1:500-1:2000)
Alternate names:
Fractalkine receptor; RBS11; V28; G-protein coupled receptor 13; CMK-BRL-1; Beta chemokine receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519910 349.00
Check your price
  • CX3CR1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519935
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
V28; G-protein coupled receptor 13; Fractalkine receptor; CMK-BRL-1; Beta chemokine receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519935 349.00
Check your price
  • CXCR3 Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523104
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1-3 µg/ml)
Alternate names:
Interferon-inducible protein 10 receptor; G protein-coupled receptor 9; CKR-L2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523104 349.00
Check your price
  • CXCR4 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519857
  • Size 100 ug
Applications
Immunofluorescence (4 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22; Pre-B-cell-derived chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519857 349.00
Check your price
  • CXCR4 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519856
  • Size 100 ug
Applications
Flow Cytometry (0.5 µg/ml), Immunocytochemistry (20 µg/ml), Immunofluorescence (20 µg/ml), Immunoprecipitation (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22; Pre-B-cell-derived chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519856 349.00
Check your price
  • CXCR4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520468
  • Size 100 ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (4 ug/ml), Western Blot (10 µg/ml)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520468 349.00
Check your price
  • CXCR6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519888
  • Size 100 ug
Applications
Western Blot (1:1000)
Alternate names:
G-protein coupled receptor bonzo; G-protein coupled receptor STRL33; CDw186
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519888 349.00
Check your price
  • CXCR6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519301
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
G-protein coupled receptor bonzo; G-protein coupled receptor STRL33; CDw186
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519301 349.00
Check your price
  • CXCR6 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519936
  • Size 100 ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (20 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
G-protein coupled receptor bonzo; G-protein coupled receptor STRL33; CDw186
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519936 349.00
Check your price
  • CXCR7 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523192
  • Size 100 ug
Applications
Flow Cytometry (1-10 µg/ml), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (4-6 µg/ml)
Alternate names:
G-protein coupled receptor RDC1 homolog; Chemokine orphan receptor 1; C-X-C chemokine receptor type 7; G-protein coupled receptor 159
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523192 349.00
Check your price
  • CXXC4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521054
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
IDAX; CXXC finger 4; CXXC-type zinc finger protein 4; Dvl-binding protein IDAX (inhibition of the Dvl and Axin complex); inhibition of the Dvl and axin complex protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521054 349.00
Check your price
  • CYB561D2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518098
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Putative tumor suppressor protein 101F6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518098 349.00
Check your price
  • CYB5R3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519196
  • Size 100 ug
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
Diaphorase-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519196 349.00
Check your price
  • Cyclin O Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520263
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
CCNO
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520263 349.00
Check your price