Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1611 
8071 - 8100 of 48315 products displayed
  • DFFB Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519928
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
DNA fragmentation factor 40 kDa subunit; Caspase-activated nuclease; Caspase-activated deoxyribonuclease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519928 349.00
Check your price
  • DFFB Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519891
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (2 µg/ml)
Alternate names:
DNA fragmentation factor 40 kDa subunit; Caspase-activated deoxyribonuclease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519891 349.00
Check your price
  • DGAT1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA518634
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Diglyceride acyltransferase; Acyl-CoA retinol O-fatty-acyltransferase; ACAT-related gene product 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518634 349.00
Check your price
  • DGAT2 Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518667
  • Size 100 ug
Applications
ELISA (0.5-1.0ug/ml), Immunohistochemistry (3µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Diglyceride acyltransferase 2; Acyl-CoA retinol O-fatty-acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518667 349.00
Check your price
  • DHX36 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534494
  • Size 100 ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
RNA helicase associated with AU-rich element ARE; MLE-like protein 1; DEAH box protein 36; G4-resolvase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534494 349.00
Check your price
  • DHX58 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518783
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
RIG-I-like receptor LGP2; RIG-I-like receptor 3; Protein D11Lgp2 homolog; Probable ATP-dependent helicase LGP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518783 349.00
Check your price
  • DHX58 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520433
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
RIG-I-like receptor LGP2; RIG-I-like receptor 3; Protein D11Lgp2 homolog; Probable ATP-dependent helicase LGP2; Protein D11Lgp2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520433 349.00
Check your price
  • DHX58 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520434
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
RIG-I-like receptor LGP2; RIG-I-like receptor 3; Protein D11Lgp2 homolog; Probable ATP-dependent helicase LGP2; Protein D11Lgp2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520434 349.00
Check your price
  • DHX58 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520435
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
RIG-I-like receptor LGP2; RIG-I-like receptor 3; Protein D11Lgp2 homolog; Probable ATP-dependent helicase LGP2; Protein D11Lgp2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520435 349.00
Check your price
  • DHX9 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519112
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Nuclear DNA helicase II; Leukophysin; DEAH box protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519112 349.00
Check your price
  • DIABLO Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519974
  • Size 100 ug
Applications
Immunohistochemistry (5 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
Second mitochondria-derived activator of caspase; Direct IAP-binding protein with low pI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519974 349.00
Check your price
  • DIABLO Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519975
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
Second mitochondria-derived activator of caspase; Direct IAP-binding protein with low pI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519975 349.00
Check your price
  • DIAPH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519033
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
p140mDIA; Diaphanous-related formin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519033 349.00
Check your price
  • Digoxin Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514745
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514745 349.00
Check your price
  • Digoxin Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514747
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514747 349.00
Check your price
  • Digoxin Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514746
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514746 349.00
Check your price
  • DIO2 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518659
  • Size 100 ug
Applications
Immunohistochemistry (2-3µg/ml)
Alternate names:
Type-II 5'-deiodinase; Type 2 DI; DIOII; 5DII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518659 349.00
Check your price
  • DIS3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534549
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (25 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Ribosomal RNA-processing protein 44; Protein DIS3 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534549 349.00
Check your price
  • DISC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518555
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
DISC1; disrupted in schizophrenia 1 ; RP4-730B13.1; KIAA0457
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518555 349.00
Check your price
  • DISC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520422
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9; disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520422 349.00
Check your price
  • DISC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520423
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9; disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520423 349.00
Check your price
  • DISP1 Polyclonal Antibody for Western Blot, IF, ICC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518257
  • Size 100 ug
Applications
Immunocytochemistry (1-5ug/ml), Immunofluorescence (Assay Dependent)
Alternate names:
DISP1; DISPA; MGC13130; MGC16796; DKFZP434I0428; dispatched A; DISP1; dispatched homolog 1; dispatched homolog 1 (Drosophila); FLJ43740; MGC104180
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518257 349.00
Check your price
  • DISP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520631
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
DISPB, HsT16908; dispatched B; protein dispatched homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520631 349.00
Check your price
  • DIXDC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519232
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
DIX domain-containing protein 1; Coiled-coil protein DIX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519232 349.00
Check your price
  • DKC1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518681
  • Size 100 ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
snoRNP protein DKC1; Nucleolar protein family A member 4; Nucleolar protein NAP57; Nopp140-associated protein of 57 kDa; Dyskerin; CBF5 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518681 349.00
Check your price
  • DKK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA523187
  • Size 100 ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
SK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523187 349.00
Check your price
  • DKK2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518015
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
DKK2; dickkopf homolog 2 (Xenopus laevis); DKK-2; Dickkopf gene 2; dickkopf (Xenopus laevis) homolog 2; dickkopf 2; dickkopf homolog 2; dickkopf related protein-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518015 349.00
Check your price
  • DKK3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518016
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (1-10ug/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
DKK3; REIC; dickkopf homolog 3 (Xenopus laevis); DKK-3; RIG-like 5-6; RIG-like 7-1; dickkopf 3; Dickkopf gene 3; dickkopf (Xenopus laevis) homolog 3; dickkopf homolog 3; RIG; regulated in glioma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518016 349.00
Check your price
  • DKK4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518017
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
DKK4; dickkopf homolog 4 (Xenopus laevis); DKK-4; hdkk-4; Dickkopf gene 4; dickkopf (Xenopus laevis) homolog 4; MGC129562; MGC129563; dickkopf homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518017 349.00
Check your price
  • DLK1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534433
  • Size 100 ug
Applications
Immunocytochemistry (25 ug/ml), Immunofluorescence (25 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
pG2; Preadipocyte factor 1; Adipocyte differentiation inhibitor protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534433 349.00
Check your price