Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1630 
8191 - 8220 of 48882 products displayed
  • GBP5 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534541
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (25 ug/ml), Western Blot (1 ug/mL)
Alternate names:
Guanine nucleotide-binding protein 5; GTP-binding protein 5; GBP-TA antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534541 349.00
Check your price
  • GBX2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518561
  • Size 100ug
Applications
Immunohistochemistry (4-8µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Gastrulation and brain-specific homeobox protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518561 349.00
Check your price
  • GC Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518794
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Vitamin D-binding protein-macrophage activating factor; Group-specific component; Gc-globulin; Gc protein-derived macrophage activating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518794 349.00
Check your price
  • GC Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518795
  • Size 100ug
Applications
Western Blot (0.5-3 ug/ml)
Alternate names:
Vitamin D-binding protein-macrophage activating factor; Group-specific component; Gc-globulin; Gc protein-derived macrophage activating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518795 349.00
Check your price
  • GCDFP-15 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518507
  • Size 100ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
gp17; Secretory actin-binding protein; Prolactin-induced protein; Gross cystic disease fluid protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518507 349.00
Check your price
  • GCH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518885
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
GTP cyclohydrolase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518885 349.00
Check your price
  • GCKR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519103
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
GCKR; glucokinase (hexokinase 4) regulator; GKRP; glucokinase (hexokinase 4) regulatory protein; glucokinase regulator; glucokinase regulatory protein; hexokinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519103 349.00
Check your price
  • GCNT3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518857
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; C2GnT-mucin type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518857 349.00
Check your price
  • GCNT3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518858
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; C2GnT-mucin type
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518858 349.00
Check your price
  • GDF1 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534473
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
GDF1, Growth differentiation factor 1, GDF-1, embryonic growth/differentiation factor 1, DORV, DTGA3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534473 349.00
Check your price
  • GDF6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520568
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Growth/differentiation factor 16; Bone morphogenetic protein 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520568 349.00
Check your price
  • GEF5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518471
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
p60 TIM; Transforming immortalized mammary oncogene; Oncogene TIM; Guanine nucleotide regulatory protein TIM; Ephexin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518471 349.00
Check your price
  • Gelsolin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518605
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Brevin; Actin-depolymerizing factor; AGEL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518605 349.00
Check your price
  • Gemin 2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520978
  • Size 100ug
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Survival of motor neuron protein-interacting protein 1; Component of gems 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520978 349.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, IHC (P), ELISA and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518598
  • Size 100ug
Applications
ELISA (5-10ug/ml), Immunohistochemistry (Paraffin) (2-4 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
GFAP; glial fibrillary acidic protein ; FLJ45472 ; intermediate filament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518598 349.00
Check your price
  • GFI1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523495
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (6-8 µg/ml)
Alternate names:
Zinc finger protein 163; Growth factor independent protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523495 349.00
Check your price
  • GFP Tag Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA515349
  • Size 100ug
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1:200-1:1000), Western Blot (1/500-1/2000)
Alternate names:
Green Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515349 349.00
Check your price
  • GFR alpha-1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519873
  • Size 100ug
Applications
Immunohistochemistry (1 µg/ml), Western Blot (Assay Dependent)
Alternate names:
TGF-beta-related neurotrophic factor receptor 1; RET ligand 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519873 349.00
Check your price
  • GFR alpha-2 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519874
  • Size 100ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGF-beta-related neurotrophic factor receptor 2; RET ligand 2; Neurturin receptor alpha; GDNF receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519874 349.00
Check your price
  • GFR alpha-3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519875
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
GFRalpha3, Y15110; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNFR-alpha-3; GFR alpha-3; GFR-alpha-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519875 349.00
Check your price
  • GGCX Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519155
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Vitamin K gamma glutamyl carboxylase; Peptidyl-glutamate 4-carboxylase; Gamma-glutamyl carboxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519155 349.00
Check your price
  • GHDC Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534464
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
GHDC, GH3 domain-containing protein, GHDC1, DL11LGP1, LGP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534464 349.00
Check your price
  • Ghrelin Polyclonal Antibody for Western Blot, ICC, IHC, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA518409
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunohistochemistry (2-5µg/ml)
Alternate names:
Protein M46; Motilin-related peptide; Growth hormone-releasing peptide; Growth hormone secretagogue
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518409 349.00
Check your price
  • GID8 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520799
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Two hybrid-associated protein 1 with RanBPM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520799 349.00
Check your price
  • GIPC3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518227
  • Size 100ug
Applications
Western Blot (0.2-1.5 ug/ml)
Alternate names:
GIPC3; PDZ domain protein GIPC3; GIPC PDZ domain containing family, member 3; C19orf64; DKFZp686J1198; FLJ40925;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518227 349.00
Check your price
  • Girdin Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520869
  • Size 100ug
Applications
Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Hook-related protein 1; Girders of actin filament; G alpha-interacting vesicle-associated protein; Coiled-coil domain-containing protein 88A; Akt phosphorylation enhancer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520869 349.00
Check your price
  • GITRL Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520161
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Glucocorticoid-induced TNF-related ligand; Activation-inducible TNF-related ligand; GITR ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520161 349.00
Check your price
  • GKN1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Horse, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # PA523309
  • Size 100ug
Applications
Western Blot (0.1-0.2 µg/ml)
Alternate names:
18 kDa antrum mucosa protein; Protein CA11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523309 349.00
Check your price
  • GLE1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520660
  • Size 100ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
GLE1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520660 349.00
Check your price
  • GLE1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520661
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
GLE1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520661 349.00
Check your price