Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  289  290  291  292  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1605 
8311 - 8340 of 48136 products displayed
  • GEMC1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521158
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
GEMC1; geminin coiled-coil domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521158 349.00
Check your price
  • Gemin 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520978
  • Size 100 ug
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Survival of motor neuron protein-interacting protein 1; Component of gems 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520978 349.00
Check your price
  • GFAP Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518598
  • Size 100 ug
Applications
ELISA (5-10ug/ml), Immunohistochemistry (Paraffin) (2-4 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
GFAP; glial fibrillary acidic protein ; FLJ45472 ; intermediate filament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518598 349.00
Check your price
  • GFI1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523495
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (6-8 µg/ml)
Alternate names:
Zinc finger protein 163; Growth factor independent protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523495 349.00
Check your price
  • GFP Tag Antibody (Monoclonal, 4B10B2)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA515349
  • Size 100 ug
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1:200-1:1000), Western Blot (1/500-1/2000)
Alternate names:
Green Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515349 349.00
Check your price
  • GFR alpha-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519873
  • Size 100 ug
Applications
Immunohistochemistry (1 µg/ml), Western Blot (Assay Dependent)
Alternate names:
TGF-beta-related neurotrophic factor receptor 1; RET ligand 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519873 349.00
Check your price
  • GFR alpha-2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519874
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGF-beta-related neurotrophic factor receptor 2; RET ligand 2; Neurturin receptor alpha; GDNF receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519874 349.00
Check your price
  • GFR alpha-3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519875
  • Size 100 ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
GFRalpha3, Y15110; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNFR-alpha-3; GFR alpha-3; GFR-alpha-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519875 349.00
Check your price
  • GGCX Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519155
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Vitamin K gamma glutamyl carboxylase; Peptidyl-glutamate 4-carboxylase; Gamma-glutamyl carboxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519155 349.00
Check your price
  • GHDC Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534464
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
GHDC, GH3 domain-containing protein, GHDC1, DL11LGP1, LGP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534464 349.00
Check your price
  • Ghrelin Antibody
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA518409
  • Size 100 ug
Applications
Immunocytochemistry (Assay-dependent), Immunohistochemistry (2-5µg/ml)
Alternate names:
Protein M46; Motilin-related peptide; Growth hormone-releasing peptide; Growth hormone secretagogue
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518409 349.00
Check your price
  • GIPC3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518227
  • Size 100 ug
Applications
Western Blot (0.2-1.5 ug/ml)
Alternate names:
GIPC3; PDZ domain protein GIPC3; GIPC PDZ domain containing family, member 3; C19orf64; DKFZp686J1198; FLJ40925;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518227 349.00
Check your price
  • Girdin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520869
  • Size 100 ug
Applications
Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Hook-related protein 1; Girders of actin filament; G alpha-interacting vesicle-associated protein; Coiled-coil domain-containing protein 88A; Akt phosphorylation enhancer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520869 349.00
Check your price
  • GITRL Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520161
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Glucocorticoid-induced TNF-related ligand; Activation-inducible TNF-related ligand; GITR ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520161 349.00
Check your price
  • GKN1 Antibody
  • Host Rabbit
  • Target Species Horse, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # PA523309
  • Size 100 ug
Applications
Western Blot (0.1-0.2 µg/ml)
Alternate names:
18 kDa antrum mucosa protein; Protein CA11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523309 349.00
Check your price
  • GLE1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520660
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
GLE1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520660 349.00
Check your price
  • GLE1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520661
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
GLE1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520661 349.00
Check your price
  • GLI1 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523411
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Oncogene GLI; Glioma-associated oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523411 349.00
Check your price
  • GLS2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521113
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
L-glutamine amidohydrolase; L-glutaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521113 349.00
Check your price
  • GluR4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518931
  • Size 100 ug
Applications
Immunohistochemistry (5-10µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Glutamate receptor ionotropic, AMPA 4; GluR-D; AMPA-selective glutamate receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518931 349.00
Check your price
  • GluR5 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518789
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Glutamate receptor 5; Excitatory amino acid receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518789 349.00
Check your price
  • GluR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520448
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Glutamate receptor 5; Excitatory amino acid receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520448 349.00
Check your price
  • GluR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520447
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Glutamate receptor 5; Excitatory amino acid receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520447 349.00
Check your price
  • GluR6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520449
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Glutamate receptor 6; Excitatory amino acid receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520449 349.00
Check your price
  • GluR7 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518964
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Glutamate receptor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518964 349.00
Check your price
  • GluR7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520450
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Glutamate receptor 7; Excitatory amino acid receptor 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520450 349.00
Check your price
  • GLUT4 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519333
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Glucose transporter type 4, insulin-responsive; GT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519333 349.00
Check your price
  • Glutamate Dehydrogenase Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519267
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (3-5 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
GLUD1; glutamate dehydrogenase 1; GDH; GDH1; GLUD; MGC132003; glutamate dehydrogenase (NAD(P)+); GLUD2; glutamate dehydrogenase 2; GDH2; GLUDP1; glutamate dehydrogenase pseudogene 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519267 349.00
Check your price
  • GNB3/GNB4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519401
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (3-5 ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Transducin beta chain 3; Transducin beta chain 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519401 349.00
Check your price
  • GNPDA1 Antibody
  • Host Rabbit
  • Target Species Horse, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523241
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.25-0.50 µg/ml)
Alternate names:
Oscillin; Glucosamine-6-phosphate deaminase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523241 349.00
Check your price