Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  60  70  80  90  100  110  120  130  200  300  400  500  600  700  800  900  1000  1622 
1081 - 1110 of 48655 products displayed
  • CGRP Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0260502
  • Size 200 µL
Applications
ELISA (1:2000), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0260502 550.00
Check your price
  • Corynebacterium diphtheriae Toxin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB1230902
  • Size 200 µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C. diphtheriae, Diphtheria
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1230902 550.00
Check your price
  • Corynebacterium diphtheriae Toxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB1230502
  • Size 200 µl
Applications
ELISA (1:35,000)
Alternate names:
C. diphtheriae, Diphtheria
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1230502 550.00
Check your price
  • Gelsolin Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0172002
  • Size 200 µL
Applications
ELISA (1:4000), Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (Paraffin) (1:6000), Western Blot (Assay dependent)
Alternate names:
Brevin; Actin-depolymerizing factor; AGEL
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0172002 550.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF, ICC, IHC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160302
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunohistochemistry (Assay dependent), Western Blot (1:40)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160302 550.00
Check your price
  • Lamin C Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA15827
  • Size 200 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay Dependent)
Alternate names:
C type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15827 550.00
Check your price
  • NPY Monoclonal Antibody for IF, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0280802
  • Size 200 µL
Applications
ELISA (1:16,000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
NPY
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0280802 550.00
Check your price
  • Ovalbumin Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # HYB0990202
  • Size 200 µl
Applications
ELISA (1:2,500), Immunoprecipitation (Assay dependent)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0990202 550.00
Check your price
  • Ovalbumin Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # HYB0990102
  • Size 200 µl
Applications
ELISA (Assay-dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0990102 550.00
Check your price
  • Ovalbumin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # HYB0990902
  • Size 200 µl
Applications
ELISA (1:8,000)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0990902 550.00
Check your price
  • Ovalbumin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # HYB0991102
  • Size 200 µL
Applications
ELISA (1/4000)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0991102 550.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # CSI0017102
  • Size 200 µl
Applications
ELISA (1:8,000), Immunohistochemistry (1:100), Western Blot (1:100)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
CSI0017102 550.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # CSI0017402
  • Size 200 µl
Applications
ELISA (1:2,000), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
CSI0017402 550.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # CSI0017602
  • Size 200 µl
Applications
ELISA (1:8,000), Immunohistochemistry (Frozen) (1:100), Western Blot (Assay dependent)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
CSI0017602 550.00
Check your price
  • Poliovirus Type 1 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB2951502
  • Size 200 µL
Applications
ELISA (Assay Dependent)
Alternate names:
PV-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2951502 550.00
Check your price
  • Poliovirus Type 1 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB2951702
  • Size 200 µl
Applications
ELISA (1:16,000)
Alternate names:
PV-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2951702 550.00
Check your price
  • Poliovirus Type 2 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB2940602
  • Size 200 µL
Applications
ELISA (1:1000)
Alternate names:
PV-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2940602 550.00
Check your price
  • Poliovirus Type 2 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB2940202
  • Size 200 µl
Applications
ELISA (Assay Dependent)
Alternate names:
PV-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2940202 550.00
Check your price
  • Poliovirus Type 3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB3000502
  • Size 200 µL
Applications
ELISA (Assay dependent)
Alternate names:
PV-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB3000502 550.00
Check your price
  • Poliovirus Type 3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB3000602
  • Size 200 µL
Applications
ELISA (1/8000)
Alternate names:
PV-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB3000602 550.00
Check your price
  • VIP Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0230202
  • Size 200 µL
Applications
ELISA (1:1000), Western Blot (Assay dependent)
Alternate names:
VIP-R-1, Pituitary adenylate cyclase-activating polypeptide type II receptor_x000D_ PACAP type II receptor, PACAP-R-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0230202 550.00
Check your price
  • Luteinizing Hormone Releasing Hormone Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP10934
  • Size 25 mg
Applications
Control (Assay Dependent)
Alternate names:
LHRH, GnRH, Lutropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP10934 550.00
Check your price
  • F4/80 Monoclonal Antibody for Western Blot, IF, IHC, IHC (F), IHC (P), Flow, IP, RIA and IM
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516632
  • Size 250 ug
Applications
Flow Cytometry (1:50-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516632 550.00
Check your price
  • Factor XII Heavy chain Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110828
  • Size 250 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Hageman factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110828 550.00
Check your price
  • Mouse anti-Monkey IgA Secondary Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516729
  • Size 250 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516729 550.00
Check your price
  • Calicivirus Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182322
  • Size 250 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Feline calicivirus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182322 550.00
Check your price
  • CD1a Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA133803
  • Size 250 µg
Applications
Flow Cytometry (1:50-1:100 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
T-cell surface antigen T6/Leu-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133803 550.00
Check your price
  • FIV p24 gag Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183937
  • Size 250 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Feline Immunodeficiency Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183937 550.00
Check your price
  • FIV p24 gag Monoclonal Antibody for Western Blot, IF, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182184
  • Size 250 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Feline Immunodeficiency Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182184 550.00
Check your price
  • LEF1 Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112420
  • Size 250 µg
Applications
Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1-5 µg/ml)
Alternate names:
T cell-specific transcription factor 1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112420 550.00
Check your price