Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  60  70  80  90  100  110  120  130  200  300  400  500  600  700  800  900  1000  1611 
1171 - 1200 of 48310 products displayed
  • PHD3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA120196
  • Size 100 µL
Applications
Western Blot (1:1000-1:3000)
Alternate names:
Prolyl hydroxylase domain-containing protein 3; Hypoxia-inducible factor prolyl hydroxylase 3; HPH-1; SM-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120196 499.00
Check your price
  • Sigma 54 Antibody (Monoclonal, 33.7)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA15876
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15876 499.00
Check your price
  • PDE2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131128
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (1:250), Western Blot (1:500)
Alternate names:
Cyclic GMP-stimulated phosphodiesterase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131128 499.00
Check your price
  • P2X3 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA130936
  • Size 150 µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (1:1000), Western Blot (1:1000)
Alternate names:
Purinergic receptor; ATP receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130936 499.00
Check your price
  • Rat KGF Recombinant Protein
  • Host E. coli
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # RP10932
  • Size 20 µg
Applications
Control (Assay Dependent)
Alternate names:
Rat rKeratinocyte Growth Factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10932 499.00
Check your price
  • C5AR1 Antibody (Monoclonal, S5/1)
  • Host Mouse
  • Target Species Bovine, Ferret, Human, Mink, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA516937
  • Size 200 ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C5a anaphylatoxin chemotactic receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516937 499.00
Check your price
  • Kininogen Heavy Chain Antibody (Monoclonal, 2B5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110832
  • Size 200 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Williams-Fitzgerald-Flaujeac factor; High molecular weight kininogen; Fitzgerald factor; Alpha-2-thiol proteinase inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110832 499.00
Check your price
  • MMP2 Antibody (Monoclonal, VB3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MS804PABX
  • Size 200 ug
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MS804PABX 499.00
Check your price
  • MMP9 Antibody (Monoclonal, IIA5)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MS817PABX
  • Size 200 ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MS817PABX 499.00
Check your price
  • Prekallikrein Heavy Chain Antibody (Monoclonal, 13G11)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110830
  • Size 200 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Plasma prekallikrein; Kininogenin; Fletcher factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110830 499.00
Check your price
  • BrdU Antibody (Monoclonal, BU1/75 (ICR1))
  • Host Rat
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA182088
  • Size 200 µg
Applications
Flow Cytometry (1/25 -1/200), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1/25 -1/200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182088 499.00
Check your price
  • CD79a Antibody (Monoclonal, ZL7-4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA181093
  • Size 200 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (10 ug/ml), Immunoprecipitation (20 ug/ml)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181093 499.00
Check your price
  • LAIR1 Antibody (Monoclonal, NKTA255)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA133625
  • Size 200 µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
LAIR1, leukocyte-associated immunoglobulin-like receptor 1, CD305, LAIR-1, leukocyte-associated Ig-like receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133625 499.00
Check your price
  • LRP1 Antibody (Monoclonal, A2Mr alpha-2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180192
  • Size 200 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50-1/100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay dependent (Non-redu)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180192 499.00
Check your price
  • Osteopontin Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA132149
  • Size 200 µL
Applications
ELISA (1:20,000-1:100,000), Immunohistochemistry (Paraffin) (1:100-1:300), Western Blot (1:500-1:2000)
Alternate names:
Secreted phosphoprotein 1; Minopontin; Early T-lymphocyte activation 1 protein; Calcium oxalate crystal growth inhibitor protein; Bone sialoprotein 1; 2AR; Uropontin; Urinary stone protein; Nephropontin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132149 499.00
Check your price
  • Frequenin Antibody
  • Host Chicken
  • Target Species Mammal
  • Conjugate
  • Catalog # PA132140
  • Size 200µg
Applications
ELISA (1:10,000-1:50,000), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
RP23-32C16.4, 9430075O15Rik, A730032G13Rik, AI836659, Freq, Mfreq, NCS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132140 499.00
Check your price
  • beta-3 Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120714
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Beta-3 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120714 499.00
Check your price
  • beta-3 Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120715
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Beta-3 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120715 499.00
Check your price
  • CXCR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120673
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
IL-8 receptor type 2; High affinity interleukin-8 receptor B; GRO/MGSA receptor; CDw128b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120673 499.00
Check your price
  • Endothelin A Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115394
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Endothelin A receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115394 499.00
Check your price
  • GPR43 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120419
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
G-protein coupled receptor 43
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120419 499.00
Check your price
  • GPR81 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120524
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor 81; G-protein coupled receptor 104
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120524 499.00
Check your price
  • GPR81 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120523
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor 81; G-protein coupled receptor 104
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120523 499.00
Check your price
  • GRPR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115619
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
GRP-preferring bombesin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115619 499.00
Check your price
  • P2Y12 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120758
  • Size 25 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
SP1999; P2Y12 platelet ADP receptor; P2Y(cyc); P2Y(AC); P2T(AC); ADP-glucose receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120758 499.00
Check your price
  • P2Y6 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120617
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
MGC15335, P2Y6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120617 499.00
Check your price
  • SSTR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120630
  • Size 25 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
SRIF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120630 499.00
Check your price
  • CD68 Antibody (Monoclonal, ED1)
  • Host Mouse
  • Target Species Bovine, Rat
  • Conjugate Unconjugated
  • Catalog # MA516654
  • Size 250 ug
Applications
Flow Cytometry (1:50-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
GP110, LAMP4, SCARD1, CD68 antigen, macrophage antigen CD68, macrosialin, scavenger receptor class D member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516654 499.00
Check your price
  • CD68 Antibody (Monoclonal, FA-11)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516674
  • Size 250 ug
Applications
Flow Cytometry (1:50-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
GP110, LAMP4, SCARD1, CD68 antigen, macrophage antigen CD68, macrosialin, scavenger receptor class D member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516674 499.00
Check your price
  • 5-Methylcytosine Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA130675
  • Size 250 µg
Applications
Flow Cytometry (Assay-dependent), Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Frozen) (10 µg/ml), Immunoprecipitation (25-100 µg/ml), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130675 499.00
Check your price