Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  70  80  90  100  110  120  130  140  200  300  400  500  600  700  800  900  1000  1605 
1381 - 1410 of 48123 products displayed
  • Lamin A/C Antibody (Monoclonal, Jol3)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA15820
  • Size 1 mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15820 475.00
Check your price
  • Listeria monocytogenes Antibody, HRP
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate HRP
  • Catalog # PA185652
  • Size 1 mL
Applications
ELISA (1/200-1/1000), Immunohistochemistry (Frozen) (1/20-1/200), Western Blot (1/20-1/200)
Alternate names:
L. monocytogenes
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185652 475.00
Check your price
  • Macrophages / Histiocytes Antibody (Monoclonal, 3A5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135593
  • Size 1 mL
Applications
Immunofluorescence (1:20-1:50), Immunohistochemistry (Paraffin, Frozen) (1:20-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135593 475.00
Check your price
  • mGluR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138130
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
glutamate receptor, metabotropic 1, GRM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138130 475.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130408
  • Size 1 mL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-Dependent)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130408 475.00
Check your price
  • Phospho-AKT1 (Ser473) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137046
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137046 475.00
Check your price
  • PSMA Antibody (Monoclonal, SP29)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514579
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Pteroylpoly-gamma-glutamate carboxypeptidase; Prostate-specific membrane antigen; N-acetylated-alpha-linked acidic dipeptidase I; Membrane glutamate carboxypeptidase; Glutamate carboxypeptidase II; Folylpoly-gamma-glutamate carboxypeptidase; Folate hydrolase 1; Cell growth-inhibiting gene 27 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514579 475.00
Check your price
  • SMAD3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138613
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
SMAD family member 3; JV15-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138613 475.00
Check your price
  • Sodium Channel Pan Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138631
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Voltage-gated sodium channel subunit alpha Nav1.1; Sodium channel protein type I subunit alpha; Sodium channel protein brain I subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138631 475.00
Check your price
  • VEGF Receptor 1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137710
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Vascular permeability factor receptor; Tyrosine-protein kinase receptor FLT; Tyrosine-protein kinase FRT; Fms-like tyrosine kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137710 475.00
Check your price
  • PDGF-C Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112720
  • Size 10 Miniblots
Applications
ELISA (Assay Dependent), Western Blot (1:50-1:200)
Alternate names:
VEGF-E; Spinal cord-derived growth factor; Fallotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112720 475.00
Check your price
  • SGPP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112723
  • Size 10 Miniblots
Applications
Western Blot (1:400)
Alternate names:
Sphingosine-1-phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112723 475.00
Check your price
  • Human MMP3 Recombinant Protein
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77528
  • Size 10 µg
Applications
Control (Assay Dependent)
Alternate names:
Stromelysin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77528 475.00
Check your price
  • Phospho-ELK1 (Ser383) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131808
  • Size 10 µg
Applications
Western Blot (1:1000)
Alternate names:
ELK, EPHT2, FLJ37986, Hek6, NET
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131808 475.00
Check your price
  • CD25 Antibody, PE-Cyanine5 (Monoclonal, BC96)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE-Cyanine5
  • Catalog # MA516588
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516588 475.00
Check your price
  • F4/80 Antibody, APC (Monoclonal, Cl:A3-1)
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA516625
  • Size 100 TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516625 475.00
Check your price
  • KIR2DL3 Antibody, PE (Monoclonal, 190IIC311)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA181620
  • Size 100 Tests
Applications
Flow Cytometry (Neat)
Alternate names:
p58.2 MHC class-I-specific NK receptor; p58 natural killer cell receptor clone CL-6; Natural killer-associated transcript 2; NKAT2b; NKAT2a; MHC class I NK cell receptor; Killer inhibitory receptor cl 2-3; KIR-023GB; CD158 antigen-like family member B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181620 475.00
Check your price
  • MHC II Monomorphic Antibody, FITC (Monoclonal, YKIX334.2)
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181228
  • Size 100 Tests
Applications
Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181228 475.00
Check your price
  • SIRP alpha Antibody, PE (Monoclonal, ED9)
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA516664
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
Signal-regulatory protein alpha-1; Macrophage membrane protein MFP150; Macrophage fusion receptor; Inhibitory receptor SHPS-1; CD172 antigen-like family member A; Brain Ig-like molecule with tyrosine-based activation motifs
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516664 475.00
Check your price
  • B7-H4 Antibody, FITC (Monoclonal, MIH43)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516845
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
T-cell costimulatory molecule B7x; Protein B7S1; Immune costimulatory protein B7-H4; B7h.5; B7 homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516845 475.00
Check your price
  • CD11a/CD18 Antibody, FITC (Monoclonal, YKIX490.6.4)
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA516994
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
CD11 antigen-like family member A, LFA-1 alpha, LFA-1A, antigen CD11A p180, lymphocyte function-associated antigen 1 alpha polypeptide, integrin alpha-L, Cd18, LAD, LCAMB, Lfa1, MF17, Integrin beta-2 subunit, ITGB2, cluster designation 18, ITGAL, ITGB2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516994 475.00
Check your price
  • CD200 Antibody, FITC (Monoclonal, OX-104)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516897
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516897 475.00
Check your price
  • CD204 Antibody, Biotin (Monoclonal, 2F8)
  • Host Rat
  • Target Species Fish, Mouse, Pig
  • Conjugate Biotin
  • Catalog # MA516493
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516493 475.00
Check your price
  • CD31 Antibody, Biotin (Monoclonal, LCI-4)
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Biotin
  • Catalog # MA516770
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:200)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516770 475.00
Check your price
  • CD3e Antibody, FITC (Monoclonal, CD3-12)
  • Host Rat
  • Target Species Many
  • Conjugate FITC
  • Catalog # MA516623
  • Size 100 ug
Applications
Flow Cytometry (1:5-1:10)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516623 475.00
Check your price
  • CD5 Antibody, FITC (Monoclonal, FE1.1B11)
  • Host Mouse
  • Target Species Cat
  • Conjugate FITC
  • Catalog # MA516681
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516681 475.00
Check your price
  • CD5 Antibody, FITC (Monoclonal, YKIX322.3)
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181217
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181217 475.00
Check your price
  • CD68 Antibody, Biotin (Monoclonal, FA-11)
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA516675
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
GP110, LAMP4, SCARD1, CD68 antigen, macrophage antigen CD68, macrosialin, scavenger receptor class D member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516675 475.00
Check your price
  • CD8 alpha Antibody, FITC (Monoclonal, 11-39)
  • Host Mouse
  • Target Species Avian, Chicken
  • Conjugate FITC
  • Catalog # MA516441
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
CD8, Leu-2, MAL, p32, CD8a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516441 475.00
Check your price
  • F4/80 Antibody, Biotin (Monoclonal, Cl:A3-1)
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA516626
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516626 475.00
Check your price