Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  70  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1605 
1561 - 1590 of 48123 products displayed
  • Aquaporin 4 Antibody (Monoclonal, 3D2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134259
  • Size 50 µg
Applications
ELISA (1:1000-1:20,000), Immunohistochemistry (Frozen) (1:500-1:1000), Immunohistochemistry (Paraffin) (1:500-1:1000), Immunoprecipitation (1:1000-1:10,000), Western Blot (1:1000-1:10,000)
Alternate names:
WCH4; Mercurial-insensitive water channel
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134259 475.00
Check your price
  • ATP6IP2 Antibody
  • Host Goat
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132079
  • Size 50 µg
Applications
ELISA (Assay-dependent), Western Blot (0.25-1 µg/ml)
Alternate names:
Vacuolar ATP synthase membrane sector-associated protein M8-9; Renin/prorenin receptor; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H(+)-transporting lysosomal accessory protein 2; N14F; Embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132079 475.00
Check your price
  • BCMA Antibody (Monoclonal, Vicky-1)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125953
  • Size 50 µg
Applications
Flow Cytometry (1 µg/ml), Immunocytochemistry (5 µg/ml), Immunofluorescence (5 ug/ml)
Alternate names:
B-cell maturation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125953 475.00
Check your price
  • BMI-1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132487
  • Size 50 µg
Applications
ELISA (1:5000-1:30,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132487 475.00
Check your price
  • Caspase 3 Antibody
  • Host Rabbit
  • Target Species Ferret, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126426
  • Size 50 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (10-20 µg/ml), Immunohistochemistry (Paraffin) (10-20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; LICE; IRP; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126426 475.00
Check your price
  • CCL4 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128319
  • Size 50 µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
ACT2, AT744.1, G-26, LAG1, MGC104418, MGC126025, MGC126026, MIP-1-beta, MIP1B, MIP1B1, SCYA2, SCYA4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128319 475.00
Check your price
  • CCR4 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA129771
  • Size 50 µg
Applications
ELISA (1:50,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:500), Western Blot (1:1000)
Alternate names:
CC-CKR-4, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5, MGC88293
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129771 475.00
Check your price
  • CD30 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130180
  • Size 50 µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Lymphocyte activation antigen CD30; Ki-1 antigen; CD30L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130180 475.00
Check your price
  • Ceruloplasmin Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127513
  • Size 50 µg
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
Ferroxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127513 475.00
Check your price
  • Claudin 23 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124429
  • Size 50 µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (0.2-0.5 µg/ml)
Alternate names:
hCG_1646163, CLDNL, hCG1646163
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124429 475.00
Check your price
  • CX3CL1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130943
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130943 475.00
Check your price
  • Cyclophilin E Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127773
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Rotamase E; Cyclophilin-33; Cyclophilin E
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127773 475.00
Check your price
  • Desmoplakin Antibody (Monoclonal, DP-2.15)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA183118
  • Size 50 µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
250/210 kDa paraneoplastic pemphigus antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183118 475.00
Check your price
  • DLG7 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127490
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Hepatoma up-regulated protein; Disks large-associated protein DLG7; Discs large homolog 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127490 475.00
Check your price
  • Drebrin Antibody (Monoclonal, M2F6)
  • Host Mouse
  • Target Species Chicken, Cat, Guinea pig, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA120377
  • Size 50 µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Developmentally-regulated brain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120377 475.00
Check your price
  • Dynamin 1 Antibody (Monoclonal, 4E67)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191537
  • Size 50 µg
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5 mg/ml)
Alternate names:
Dynamin, brain; D100; B-dynamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191537 475.00
Check your price
  • Ephrin A4 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA130380
  • Size 50 µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
EPH-related receptor tyrosine kinase ligand 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130380 475.00
Check your price
  • Ephrin B2 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA130384
  • Size 50 µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
HTK ligand; EPH-related receptor tyrosine kinase ligand 5; ELF-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130384 475.00
Check your price
  • ETV6 Antibody (Monoclonal, BM3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191170
  • Size 50 µg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (1:100)
Alternate names:
ETS-related protein Tel1; ETS translocation variant 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191170 475.00
Check your price
  • EWSR1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127592
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Ewing sarcoma breakpoint region 1 protein; EWS oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127592 475.00
Check your price
  • EXOC3 Antibody (Monoclonal, 9H5)
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA125480
  • Size 50 µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (12.5 µg/ml), Western Blot (4 µg/ml)
Alternate names:
Exocyst complex component Sec6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125480 475.00
Check your price
  • Fibronectin Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127507
  • Size 50 µg
Applications
ELISA (0.1 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Cold-insoluble globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127507 475.00
Check your price
  • GIPC1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127811
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Synectin; SemaF cytoplasmic domain-associated protein 1; RGS19-interacting protein 1; RGS-GAIP-interacting protein; GAIP C-terminus-interacting protein; Tax interaction protein 2; GLUT1 C-terminal-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127811 475.00
Check your price
  • IGF2 Antibody, Biotin
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA186837
  • Size 50 µg
Applications
ELISA (0.25-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186837 475.00
Check your price
  • IL-21 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121355
  • Size 50 µg
Applications
ELISA (1:100,000), Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Interleukin-21, IL-21, Za11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121355 475.00
Check your price
  • IL-22 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121358
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121358 475.00
Check your price
  • IRF8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124478
  • Size 50 µg
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Interferon consensus sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124478 475.00
Check your price
  • ITGA3 Antibody
  • Host Chicken
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA127601
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127601 475.00
Check your price
  • LTBR Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130176
  • Size 50 µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (Assay dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
Tumor necrosis factor receptor type III; Tumor necrosis factor receptor 2-related protein; Tumor necrosis factor C receptor; Lymphotoxin-beta receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130176 475.00
Check your price
  • M6PR Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129512
  • Size 50 µg
Applications
Immunocytochemistry (1:150-1:250), Immunofluorescence (1:150-1:250), Western Blot (Assay dependent)
Alternate names:
46 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129512 475.00
Check your price