Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1630 
1681 - 1710 of 48882 products displayed
  • Peptide Histidine-Methionine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0200302
  • Size 200µL
Applications
ELISA (1:2,000)
Alternate names:
MGC13587, PHM27
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0200302 475.00
Check your price
  • SFTPD Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2450202
  • Size 200µl
Applications
ELISA (1:1,000), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:3,000)
Alternate names:
Lung surfactant protein D; Collectin-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2450202 475.00
Check your price
  • CD204 Monoclonal Antibody for Western Blot, IHC (F), Flow, IP and ELISA
  • Host Rat
  • Target Species Fish, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA516492
  • Size 250ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516492 475.00
Check your price
  • CD80 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # MA516723
  • Size 250ug
Applications
Flow Cytometry (1:40)
Alternate names:
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516723 475.00
Check your price
  • IFN alpha Monoclonal Antibody for IP, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 221001
  • Size 250ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
IFN-alpha 1b; IFN-alpha-1/13; interferon alpha 1b; interferon alpha-1/13; interferon alpha-D; interferon-alpha1; leIF D
Catalog # Size Price (AUD) Your Price (AUD) Qty
221001 475.00
Check your price
  • MARCO Monoclonal Antibody for Western Blot, IF, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516659
  • Size 250ug
Applications
Flow Cytometry (1:25-1:50), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:200-1:400), Western Blot (Assay Dependent)
Alternate names:
Macrophage receptor with collagenous structure
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516659 475.00
Check your price
  • Aspergillus Monoclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Fungi
  • Conjugate Unconjugated
  • Catalog # MA174434
  • Size 250µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1/300), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174434 475.00
Check your price
  • CHX10 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131286
  • Size 250µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Homeobox protein CHX10; Ceh-10 homeodomain-containing homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131286 475.00
Check your price
  • CHX10 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131287
  • Size 250µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Homeobox protein CHX10; Ceh-10 homeodomain-containing homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131287 475.00
Check your price
  • DAB1 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA131307
  • Size 250µg
Applications
Immunoprecipitation (5-10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
RP23-356E21.1, AI956902, C630028C02Rik, scm, scr, scrambler, yot
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131307 475.00
Check your price
  • Feline Leukemia Virus P27 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183882
  • Size 250µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1,500)
Alternate names:
FLV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183882 475.00
Check your price
  • RECA-1 Monoclonal Antibody for IF and IHC (F)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA180181
  • Size 250µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180181 475.00
Check your price
  • Rhizopus Arhizus Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Fungi
  • Conjugate Unconjugated
  • Catalog # MA174435
  • Size 250µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay dependent)
Alternate names:
R. Arhizus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174435 475.00
Check your price
  • SRPR Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA131289
  • Size 250µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1-5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131289 475.00
Check your price
  • CD180 Monoclonal Antibody, Biotin conjugate for IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA134128
  • Size 250µL
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134128 475.00
Check your price
  • Ephrin A4 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130382
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
EPH-related receptor tyrosine kinase ligand 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130382 475.00
Check your price
  • GPR105 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115110
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
UDP-glucose receptor; G-protein coupled receptor 105
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115110 475.00
Check your price
  • GPR119 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120479
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor 119
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120479 475.00
Check your price
  • HNF1A Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA125417
  • Size 25µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Transcription factor 1; Liver-specific transcription factor LF-B1; XLFB1a; Transcription factor 1-A; LFB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125417 475.00
Check your price
  • MC4R Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115011
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
MC4R
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115011 475.00
Check your price
  • CD11c Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # MA516602
  • Size 2mL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516602 475.00
Check your price
  • F4/80 Monoclonal Antibody for IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, RIA and IM
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516624
  • Size 2mL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516624 475.00
Check your price
  • Armenian Hamster IgG Isotype Control for IF, IHC (F), Flow and Ctrl
  • Host Hamster
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PA533220
  • Size 500ug
Applications
Control (Assay-Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533220 475.00
Check your price
  • CD13 Monoclonal Antibody for IHC (F), IHC (P), Flow and FN
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516931
  • Size 500ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Microsomal aminopeptidase; Membrane protein p161; Aminopeptidase M; Alanyl aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516931 475.00
Check your price
  • CD169 Monoclonal Antibody for IF, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516507
  • Size 500ug
Applications
Flow Cytometry (1:100 -1:1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:100)
Alternate names:
Sialic acid-binding Ig-like lectin 1; Sheep erythrocyte receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516507 475.00
Check your price
  • CD244 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516486
  • Size 500ug
Applications
Flow Cytometry (1:50-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Signaling lymphocytic activation molecule 4; SLAM family member 4; NK cell type I receptor protein 2B4; NK cell activation-inducing ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516486 475.00
Check your price
  • CD36 Monoclonal Antibody for Western Blot, IF, Flow, IP and FN
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516831
  • Size 500ug
Applications
Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Platelet glycoprotein IV; PAS-4; PAS IV; Glycoprotein IIIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516831 475.00
Check your price
  • CD45 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517378
  • Size 500ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517378 475.00
Check your price
  • CD45RC Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517461
  • Size 500ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517461 475.00
Check your price
  • Erythrocytes Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517581
  • Size 500ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517581 475.00
Check your price