Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  200  300  400  530  540  550  560  570  580  590  600  610  622  623  624  625  626  627  628  629  630  631  632  633  634  635  636  637  638  639  640  641  642  643  644  645  646  647  648  649  650  651  660  670  680  690  700  710  720  730  800  900  1000  1100  1200  1300  1400  1500  1600  1603 
19081 - 19110 of 48084 products displayed
  • TERT Antibody (Monoclonal, 2C4)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516034
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Telomerase catalytic subunit; Telomerase-associated protein 2; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516034 349.00
Check your price
  • TERT Antibody (Monoclonal, 2D8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516033
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516033 349.00
Check your price
  • TERT Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532195
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532195 349.00
Check your price
  • Tesmin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531052
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Testis-specific metallothionein-like protein; Metallothionein-like 5, testis-specific
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531052 349.00
Check your price
  • TET1 Antibody (Monoclonal, GT1462)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516312
  • Size 100 µL
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:3000)
Alternate names:
Ten-eleven translocation 1 gene protein; Leukemia-associated protein with a CXXC domain; CXXC-type zinc finger protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516312 349.00
Check your price
  • TET3 Antibody
  • Host Rabbit
  • Target Species Human, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA531860
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:500-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
methylcytosine dioxygenase TET3, probable methylcytosine dioxygenase TET3, tet oncogene family member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531860 349.00
Check your price
  • TEX13A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530769
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
testis expressed sequence 13A, testis-expressed sequence 13A protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530769 349.00
Check your price
  • TEX14 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529410
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Testis-expressed sequence 14 protein; Testis-expressed sequence 14; Sugen kinase 307; Protein kinase-like protein SgK307
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529410 349.00
Check your price
  • TFAM Antibody (Monoclonal, 18G102B2E11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA516148
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
Transcription factor 6-like 2; Transcription factor 6; Mitochondrial transcription factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516148 349.00
Check your price
  • TFAM Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527865
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
Transcription factor 6-like 2; Transcription factor 6; Mitochondrial transcription factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527865 349.00
Check your price
  • TFAM Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529571
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Transcription factor 6-like 2; Transcription factor 6; Mitochondrial transcription factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529571 349.00
Check your price
  • TFB2M Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528638
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2; Mitochondrial transcription factor B2; Mitochondrial 12S rRNA dimethylase 2; Hepatitis C virus NS5A-transactivated protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528638 349.00
Check your price
  • TFCP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530257
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transcription factor LSF; SAA3 enhancer factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530257 349.00
Check your price
  • TFEB Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521616
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Class E basic helix-loop-helix protein 35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521616 349.00
Check your price
  • TFEC Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521614
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Transcription factor EC-like; Class E basic helix-loop-helix protein 34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521614 349.00
Check your price
  • TFF1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531863
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein pS2; Polypeptide P1.A; PNR-2; Breast cancer estrogen-inducible protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531863 349.00
Check your price
  • TFR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528983
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HFE3, TFRC2, transferrin receptor protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528983 349.00
Check your price
  • TFR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529137
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HFE3, TFRC2, transferrin receptor protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529137 349.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, 4F9C10,1B11B7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515289
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
CED; DPD1; TGFB; TGFbeta; TGFB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515289 349.00
Check your price
  • TGFBI Antibody
  • Host Rabbit
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # PA527261
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RGD-containing collagen-associated protein; Kerato-epithelin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527261 349.00
Check your price
  • TGFBR3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521994
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transforming growth factor beta receptor III; Betaglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521994 349.00
Check your price
  • TGIF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521900
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
TGF-beta-induced transcription factor 2; 5'-TG-3'-interacting factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521900 349.00
Check your price
  • TGM2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFPA0117
  • Size 100 µL
Applications
Immunoprecipitation (1 µl), Western Blot (1:2000)
Alternate names:
Transglutaminase-2; Transglutaminase H; Transglutaminase C; Tissue transglutaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0117 349.00
Check your price
  • TGM2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529357
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transglutaminase-2; Transglutaminase H; Transglutaminase C; Tissue transglutaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529357 349.00
Check your price
  • TGM2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529356
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transglutaminase-2; Transglutaminase H; Transglutaminase C; Tissue transglutaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529356 349.00
Check your price
  • TGN46 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523068
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:100-1:2000)
Alternate names:
Trans-Golgi network protein TGN51; TGN48; TGN46; TGN38 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523068 349.00
Check your price
  • TGR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523182
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:1000-1:5000)
Alternate names:
Membrane-type receptor for bile acids; hBG37; G-protein coupled receptor GPCR19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523182 349.00
Check your price
  • TGR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523168
  • Size 100 µL
Applications
Western Blot (1:1000-1:5000)
Alternate names:
Membrane-type receptor for bile acids; hBG37; G-protein coupled receptor GPCR19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523168 349.00
Check your price
  • TGR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523183
  • Size 100 µL
Applications
Western Blot (1:1000-1:5000)
Alternate names:
Membrane-type receptor for bile acids; hBG37; G-protein coupled receptor GPCR19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523183 349.00
Check your price
  • TGR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527076
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hBG37; Membrane-type receptor for bile acids; G-protein coupled receptor GPCR19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527076 349.00
Check your price