Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  76  77  78  80  90  100  110  120  130  140  150  160  200  300  400  500  600  700  800  900  1000  1630 
1891 - 1920 of 48882 products displayed
  • GADD34 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA133115
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Myeloid differentiation primary response protein MyD116 homolog; Growth arrest and DNA damage-inducible protein GADD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133115 449.00
Check your price
  • GFR alpha-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132476
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
TGF-beta-related neurotrophic factor receptor 1; RET ligand 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132476 449.00
Check your price
  • GPR15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130767
  • Size 100µg
Applications
Western Blot (1:500-1:2000)
Alternate names:
Brother of Bonzo
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130767 449.00
Check your price
  • GYPC Monoclonal Antibody for Western Blot, IHC (F), Flow and HA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191163
  • Size 100µg
Applications
Flow Cytometry (Assay dependent), Hemagglutination Assay (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Sialoglycoprotein D; PAS-2'; Glycoprotein beta; Glycophorin-D; Glycoconnectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191163 449.00
Check your price
  • HDAC6 Polyclonal Antibody for Western Blot, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131473
  • Size 100µg
Applications
ChIP assay (10 µl for 10^6 cells), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Histone deacetylase mHDA2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131473 449.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot, IF, IHC (F) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183231
  • Size 100µg
Applications
ELISA (1/20-1/200), Immunofluorescence (1/10-1/50), Immunohistochemistry (Frozen) (1/10-1/50), Western Blot (1/10-1/50)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183231 449.00
Check your price
  • HTT Monoclonal Antibody for Western Blot, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA182100
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Huntington disease protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182100 449.00
Check your price
  • ICAD Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA133021
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (1:1000-1:2000), Western Blot (1:1000-1:2000)
Alternate names:
Inhibitor of CAD; DNA fragmentation factor 45 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133021 449.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130896
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130896 449.00
Check your price
  • IL-3 Polyclonal Antibody for Western Blot, Neu and Ctrl
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132180
  • Size 100µg
Applications
Control (Assay dependent), Neutralization (1-10 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
P-cell-stimulating factor; Multipotential colony-stimulating factor; Mast cell growth factor; Hematopoietic growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132180 449.00
Check your price
  • IL1RA Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182844
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IL1 inhibitor; ICIL-1RA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182844 449.00
Check your price
  • IL21R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130722
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Novel interleukin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130722 449.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130723
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130723 449.00
Check your price
  • IRAK1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130672
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (2-4 µg), Western Blot (1:1000-1:2000)
Alternate names:
IL-1 Receceptor Associated Kinase, Interleukin-1 receptor-associated kinase 1, mPLK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130672 449.00
Check your price
  • IRAK3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130775
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
IL-1 receptor-associated kinase M
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130775 449.00
Check your price
  • KCND2 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Ferret, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA130660
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Voltage-gated potassium channel subunit Kv4.2; Shal1; RK5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130660 449.00
Check your price
  • KGF Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132177
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (0.5-1 ug/ml)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132177 449.00
Check your price
  • KIF4A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131786
  • Size 100µg
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:100)
Alternate names:
Chromokinesin; Chromokinesin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131786 449.00
Check your price
  • Lass1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112918
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Protein UOG-1; Longevity assurance gene 1 protein homolog 1; LAG1 longevity assurance homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112918 449.00
Check your price
  • Lass1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112919
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Protein UOG-1; Longevity assurance gene 1 protein homolog 1; LAG1 longevity assurance homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112919 449.00
Check your price
  • Lass4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112928
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
LAG1 longevity assurance homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112928 449.00
Check your price
  • LKB1 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112767
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Renal carcinoma antigen NY-REN-19; Liver kinase B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112767 449.00
Check your price
  • LPPR4 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112728
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Plasticity-related gene 1 protein; Brain-specific phosphatidic acid phosphatase-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112728 449.00
Check your price
  • MADD Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130689
  • Size 100µg
Applications
Western Blot (1:250-1:500)
Alternate names:
Rab3 GDP/GTP exchange factor; Insulinoma glucagonoma clone 20; Differentially expressed in normal and neoplastic cells
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130689 449.00
Check your price
  • MERTK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131204
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131204 449.00
Check your price
  • Neutrophils Monoclonal Antibody for IHC (F), IHC (P), Flow, ELISA and FN
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA140038
  • Size 100µg
Applications
ELISA (Assay Dependent.), Flow Cytometry (Assay Dependent.), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140038 449.00
Check your price
  • p14ARF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131212
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131212 449.00
Check your price
  • p53DINP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130836
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
p53-dependent damage-inducible nuclear protein 1; Thymus-expressed acidic protein; Stress-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130836 449.00
Check your price
  • PAI1 Monoclonal Antibody for Western Blot, IHC (F), ELISA and FN
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA140224
  • Size 100µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent.)
Alternate names:
Serpin E1; Endothelial plasminogen activator inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140224 449.00
Check your price
  • PDE11A Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131154
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1:500)
Alternate names:
cAMP and cGMP phosphodiesterase 11A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131154 449.00
Check your price