Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  100  110  120  130  140  150  160  170  180  200  300  400  500  600  700  800  900  1000  1630 
2461 - 2490 of 48881 products displayed
  • MUC2 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA138215
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Intestinal mucin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA138215 425.00
Check your price
  • N-cadherin Polyclonal Antibody for IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137199
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Neural cadherin; CDw325
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137199 425.00
Check your price
  • NFkB p50 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130409
  • Size 1mL
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:400)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; NF-kappa-B1 p84/NF-kappa-B1 p98; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130409 425.00
Check your price
  • NFkB p52 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130410
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130410 425.00
Check your price
  • p107 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130263
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Ca(2+)/calmodulin-activated adenylyl cyclase; Adenylyl cyclase 1; Adenylate cyclase type I; ATP pyrophosphate-lyase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130263 425.00
Check your price
  • p21 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130399
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
p21; Melanoma differentiation-associated protein; CDK-interacting protein 1; Melanoma differentiation-associated protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130399 425.00
Check your price
  • p38 MAPK alpha Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130391
  • Size 1mL
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130391 425.00
Check your price
  • PAR1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138749
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
Thrombin receptor; Coagulation factor II receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138749 425.00
Check your price
  • Phospho-S6 (Ser240, Ser244) Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA139503
  • Size 1mL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA139503 425.00
Check your price
  • PLCB1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138470
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
Phospholipase C-beta-1; Phospholipase C-I; Phosphoinositide phospholipase C-beta-1; PLC-154
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138470 425.00
Check your price
  • PLCG2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138468
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Phospholipase C-gamma-2; Phospholipase C-IV; Phosphoinositide phospholipase C-gamma-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138468 425.00
Check your price
  • Procalcitonin Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175361
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175361 425.00
Check your price
  • S100A4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138589
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
S100 calcium-binding protein A4; Protein Mts1; Placental calcium-binding protein; Metastasin; Calvasculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138589 425.00
Check your price
  • S100B Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175395
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
S100 calcium-binding protein B; S-100 protein subunit beta; S-100 protein beta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175395 425.00
Check your price
  • SERPINA7 Polyclonal Antibody for RIA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127075
  • Size 1mL
Applications
Radioimmune Assays (1:25)
Alternate names:
T4-binding globulin; Serpin A7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127075 425.00
Check your price
  • Shigella Polyclonal Antibody for Western Blot, IF, IHC (F) and ELISA
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA185871
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185871 425.00
Check your price
  • SHP-2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA138602
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:30)
Alternate names:
SH-PTP2; Protein-tyrosine phosphatase SYP; SH-PTP3; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase 1D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138602 425.00
Check your price
  • SPA17 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130328
  • Size 1mL
Applications
Western Blot (1:200)
Alternate names:
Sperm autoantigenic protein 17; Sp17-1; Cancer/testis antigen 22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130328 425.00
Check your price
  • Streptococcus Group A Polyclonal Antibody for IF, ICC and ELISA
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA130490
  • Size 1mL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130490 425.00
Check your price
  • Streptococcus pneumoniae Polyclonal Antibody, FITC conjugate for IF
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA533127
  • Size 1mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
S. pneumoniae, Pneumococcus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533127 425.00
Check your price
  • Thyroglobulin Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA138758
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
AITD3, TGN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA138758 425.00
Check your price
  • TREM1 Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175482
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Triggering receptor expressed on monocytes 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175482 425.00
Check your price
  • TSH beta Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA139448
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Thyrotropin beta chain; Thyroid-stimulating hormone subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA139448 425.00
Check your price
  • VEGF Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138847
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138847 425.00
Check your price
  • CD21 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134415
  • Size 200tests
Applications
Flow Cytometry (Neat (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (1:10-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Epstein-Barr virus receptor; Complement C3d receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134415 425.00
Check your price
  • CD62L Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA517576
  • Size 200ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517576 425.00
Check your price
  • GYPC Monoclonal Antibody for Western Blot, IHC (F), Flow and HA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516592
  • Size 200ug
Applications
Flow Cytometry (Assay Dependent), Hemagglutination Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Sialoglycoprotein D; PAS-2'; Glycoprotein beta; Glycophorin-D; Glycoconnectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516592 425.00
Check your price
  • Mouse anti-Human IgG2 Fab Secondary Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516715
  • Size 200ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516715 425.00
Check your price
  • Mouse anti-Human IgG3 (Hinge region) Secondary Antibody for IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516717
  • Size 200ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516717 425.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA14806
  • Size 200µg
Applications
ELISA (0.1 to 0.4 µg/ml), Flow Cytometry (5 to 10 µl/10^6 cells), Immunofluorescence (1 to 4 µg/ml), Immunohistochemistry (Frozen) (1 to 4 µg/ml), Immunoprecipitation (4 ug/ml), Western Blot (1 to 4 µg/ml)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA14806 425.00
Check your price