Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  200  300  400  500  600  700  800  900  1000  1611 
241 - 270 of 48310 products displayed
  • Mouse anti-Human IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # SA135470
  • Size 1 mL
Applications
ELISA (1:1000), Immunohistochemistry (Frozen) (1:40), Western Blot (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135470 838.00
Check your price
  • Rat anti-Mouse IgG3 Secondary Antibody, Biotin conjugate for IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA135652
  • Size 1 mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135652 836.00
Check your price
  • CD8 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD08044
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD08044 830.00
Check your price
  • Complement C4d Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183082
  • Size 100 µg
Applications
ELISA (1:5000-1:20,000), Immunofluorescence (1:250-1:600), Immunohistochemistry (Frozen) (1:100-1:750), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; Acidic complement C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183082 824.00
Check your price
  • Rat anti-Mouse IgG Secondary Antibody for IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # SA135633
  • Size 1 mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135633 806.00
Check your price
  • CD10 Monoclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516945
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516945 800.00
Check your price
  • CD35 Monoclonal Antibody for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516650
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
C3b/C4b receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516650 800.00
Check your price
  • CD38 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516578
  • Size 1 mg
Applications
Flow Cytometry (1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T10; Cyclic ADP-ribose hydrolase 1; ADP-ribosyl cyclase 1; 2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516578 800.00
Check your price
  • CD41 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human, Mustelid, Mink, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516920
  • Size 1 mg
Applications
Flow Cytometry (1:100 -1:200)
Alternate names:
Platelet membrane glycoprotein IIb; GPalpha IIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516920 800.00
Check your price
  • CD8 Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517006
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
T-cell surface glycoprotein Lyt-3; T-cell membrane glycoprotein Ly-3; Lymphocyte antigen 3; T-cell surface glycoprotein Lyt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517006 800.00
Check your price
  • KLRB1 Monoclonal Antibody for Western Blot, IHC (F), Flow, IP, RIA and FN
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA516440
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Natural killer cell surface protein NKR-P1B allele RNK/SD/BN/F344; CD161 antigen-like family member B; Natural killer cell surface protein P1-3.2.3; CD161 antigen-like family member A; Antigen 3.2.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516440 800.00
Check your price
  • P-Selectin Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516570
  • Size 1 mg
Applications
Flow Cytometry (1:10-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:500-1:200), Western Blot (Assay Dependent)
Alternate names:
Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516570 800.00
Check your price
  • Rat anti-Mouse IgE Secondary Antibody for IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # SA135629
  • Size 1 mg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA135629 800.00
Check your price
  • Calponin 1 Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514464
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Calponin H1, smooth muscle; Basic calponin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514464 800.00
Check your price
  • CDX2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514494
  • Size 1 mL
Applications
Flow Cytometry (1:10), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:100-1:1000)
Alternate names:
Caudal-type homeobox protein 2; CDX-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514494 800.00
Check your price
  • Cytokeratin 5 Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514473
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514473 800.00
Check your price
  • Estradiol Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175014
  • Size 1 mL
Applications
ELISA (Assay Dependent)
Alternate names:
17-Beta-Estradiol
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175014 800.00
Check your price
  • PAX5 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514585
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:10-1:500)
Alternate names:
B-cell-specific transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514585 800.00
Check your price
  • PMEL Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516712
  • Size 1 mL
Applications
Immunohistochemistry (Frozen) (1:40), Immunohistochemistry (Paraffin) (1:40)
Alternate names:
Silver locus protein homolog; Premelanosome protein; P100; P1; Melanoma-associated ME20 antigen; Melanocytes lineage-specific antigen GP100; Melanocyte protein Pmel 17; ME20-M
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516712 800.00
Check your price
  • Podoplanin Monoclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183884
  • Size 1 mL
Applications
Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1/10-1/40), Western Blot (Assay dependent)
Alternate names:
T1-alpha; PA2.26 antigen; Glycoprotein 36; Aggrus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183884 800.00
Check your price
  • CD229 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA135946
  • Size 100 tests
Applications
Flow Cytometry (1:50)
Alternate names:
Signaling lymphocytic activation molecule 3; SLAM family member 3; Lymphocyte antigen 9; Cell surface molecule Ly-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135946 800.00
Check your price
  • LRP1 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA135979
  • Size 100 tests
Applications
Flow Cytometry (Neat)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135979 800.00
Check your price
  • Defensin Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182491
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (0.1-0.5 ug/ml), Immunohistochemistry (Paraffin) (0.2-1 ug/ml), Western Blot (Assay Dependent)
Alternate names:
HNP-4; Defensin, alpha 4; HNP-1; Defensin, alpha 1; HNP-3; Defensin, alpha 3; HD5(20-94); Defensin, alpha 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182491 800.00
Check your price
  • Enterovirus 71 Monoclonal Antibody for IF and FN
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183142
  • Size 100 µL
Applications
Functional Assay (neat-1/10), Immunofluorescence (1/500-1/2000)
Alternate names:
EV71
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183142 800.00
Check your price
  • CD11b Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516725
  • Size 1mg
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:500), Immunoprecipitation (Assay Dependent)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516725 800.00
Check your price
  • CD206 Monoclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135936
  • Size 200 µg
Applications
Flow Cytometry (10ul x 10^6 cells or 100ul whole blood), Immunohistochemistry (Frozen) (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Macrophage mannose receptor 1-like protein 1; Human mannose receptor; C-type lectin domain family 13 member D-like; C-type lectin domain family 13 member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135936 800.00
Check your price
  • Collagen I Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA186054
  • Size 200 µg
Applications
ELISA (1:200-1:2000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186054 800.00
Check your price
  • Nectin 2 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135890
  • Size 200 µg
Applications
Flow Cytometry (1:50), Immunohistochemistry (Frozen) (Assay dependent.), Immunoprecipitation (Assay dependent.)
Alternate names:
Poliovirus receptor-related protein 2; Herpes virus entry mediator B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135890 800.00
Check your price
  • CCKAR Polyclonal Antibody for IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA136144
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1:1500-1:2500)
Alternate names:
Cholecystokinin-1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136144 800.00
Check your price
  • CGRP Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Guinea Pig
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA136018
  • Size 50 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/800-1/1200)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136018 800.00
Check your price