Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1630 
3151 - 3180 of 48881 products displayed
  • CD205 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523654
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
DEC-205
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523654 339.00
Check your price
  • KCNN3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541014
  • Size 100µl
Applications
Immunocytochemistry (1:300-1:600), Immunofluorescence (1:300-1:600), Immunohistochemistry (Paraffin) (1:300-1:600), Western Blot (0.2-2 µg/ml)
Alternate names:
KCa2.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541014 309.00
Check your price
  • KCNN2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519205
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
KCa2.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519205 349.00
Check your price
  • KCNN2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533874
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
KCa2.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533874 349.00
Check your price
  • KCNN2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541071
  • Size 100µl
Applications
Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (1:300), Western Blot (0.2-1 µg/ml)
Alternate names:
KCa2.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541071 309.00
Check your price
  • KCNN2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541012
  • Size 100µl
Applications
Immunocytochemistry (1:300-1:500), Immunofluorescence (1:300-1:500), Immunohistochemistry (1:300-1:500), Western Blot (1-3 µg/ml )
Alternate names:
KCa2.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541012 309.00
Check your price
  • KCNN1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA541013
  • Size 100µl
Applications
Western Blot (1.25 µg/ml)
Alternate names:
KCa2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541013 309.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130549
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130549 375.00
Check your price
  • ING5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521443
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p28ING5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521443 349.00
Check your price
  • ING5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526494
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
p28ING5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526494 349.00
Check your price
  • ING3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527717
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p47ING3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527717 349.00
Check your price
  • ING3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540702
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
p47ING3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540702 309.00
Check your price
  • ING3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540703
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
p47ING3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540703 309.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525159
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525159 349.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527900
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527900 349.00
Check your price
  • IL5RA (soluble) Polyclonal Antibody for Western Blot, Flow, ELISA, Neu and IA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124792
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (50-250 ng for 10^6 cells), Inhibition Assays (1-3 µg/ml), Neutralization (1-3 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124792 375.00
Check your price
  • IL5RA Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547340
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547340 349.00
Check your price
  • IL5RA Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523581
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523581 279.00
Check your price
  • IL5RA Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523621
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
CDw125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523621 309.00
Check your price
  • CD25 Oligoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710390
  • Size 100µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Western Blot (1-2 ug/ml)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
710390 309.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody for IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110393
  • Size 100ug
Applications
ELISA (Assay-Dependent), Flow Cytometry (2-8 ug/ml), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110393 225.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110396
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110396 275.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA110394
  • Size 100Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110394 275.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110395
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110395 249.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110386
  • Size 100ug
Applications
Flow Cytometry (2-8 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110386 159.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody for IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110385
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (2-8 ug/ml), Immunohistochemistry (Paraffin, Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110385 225.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110389
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110389 275.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110388
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110388 249.00
Check your price
  • IFN gamma Oligoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710287
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
710287 309.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 134400
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
134400 304.00
Check your price