Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1630 
3181 - 3210 of 48881 products displayed
  • HPRT1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522281
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HPRT B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522281 349.00
Check your price
  • HPRT1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513692
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
HPRT B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513692 349.00
Check your price
  • Acetyl-Histone H2A (Lys5) Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 720070
  • Size 100µg
Applications
ChIP assay (5 ug/ml), Flow Cytometry (2-4 ug/million cells), Immunocytochemistry (0.5 ug/ml), Immunofluorescence (0.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Histone H2A/p
Catalog # Size Price (AUD) Your Price (AUD) Qty
720070 309.00
Check your price
  • Histone H1 Monoclonal Antibody for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513750
  • Size 500µL
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:100-1:500)
Alternate names:
TISP64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513750 309.00
Check your price
  • HEBP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530609
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p22HBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530609 349.00
Check your price
  • GUSB Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526357
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Beta-G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526357 349.00
Check your price
  • GUSB Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542545
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (2.5 µg/ml)
Alternate names:
Beta-G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542545 309.00
Check your price
  • GUSB Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135024
  • Size 1mL
Applications
ELISA (1:10), Immunohistochemistry (Frozen) (1:10), Western Blot (1:10)
Alternate names:
Beta-G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135024 375.00
Check your price
  • Trehalose Receptor Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Fruit fly
  • Conjugate Unconjugated
  • Catalog # OSR00084W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Trehalose receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00084W 349.00
Check your price
  • GPR42 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539738
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
GPR42P
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539738 309.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120033
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120033 309.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120034
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120034 349.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191847
  • Size 1mg
Applications
ELISA (Assay dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191847 399.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191849
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191849 325.00
Check your price
  • FOXP3 Polyclonal Antibody for Western Blot, ICC, IHC (P), ChIP and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19044
  • Size 100ug
Applications
ChIP assay (Assay dependent), Immunocytochemistry (2-4 ug/ml), Immunohistochemistry (Paraffin) (0.3-1 µg/ml), Western Blot (Assay dependent)
Alternate names:
Scurfin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19044 349.00
Check your price
  • FGF21 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529386
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
FGF-21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529386 349.00
Check your price
  • FGF21 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547438
  • Size 100µg
Applications
Western Blot (0.25 µg/ml)
Alternate names:
FGF-21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547438 349.00
Check your price
  • FGF20 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523480
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
FGF-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523480 349.00
Check your price
  • FGF20 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523972
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
FGF-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523972 275.00
Check your price
  • CSTL1 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539121
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
RCET11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539121 309.00
Check your price
  • CSNK1A1L Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA528272
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528272 349.00
Check your price
  • CLUAP1 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513583
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:1000)
Alternate names:
Qilin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513583 349.00
Check your price
  • CLUAP1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528519
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Qilin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528519 349.00
Check your price
  • CLIC2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541006
  • Size 100µl
Applications
Western Blot (1.25 µg/ml)
Alternate names:
XAP121
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541006 309.00
Check your price
  • CLIC2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541005
  • Size 100µl
Applications
Western Blot (1.25 µg/ml)
Alternate names:
XAP121
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541005 309.00
Check your price
  • Claudin 14 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19540
  • Size 500ug
Applications
Western Blot (1:1000)
Alternate names:
DFNB29
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19540 159.00
Check your price
  • CHTF18 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544803
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
CHL12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544803 309.00
Check your price
  • Chordin Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511458
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Chordin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511458 349.00
Check your price
  • Chordin Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547934
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Chordin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547934 349.00
Check your price
  • CENPP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531186
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CENP-P
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531186 349.00
Check your price