Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  90  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  130  140  150  160  170  180  190  200  210  300  400  500  600  700  800  900  1000  1100  1622 
3301 - 3330 of 48655 products displayed
  • Goat anti-Mouse IgG / IgM Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate AP
  • Catalog # T2192
  • Size 100 µL
Applications
ELISA (1:1,000), Immunohistochemistry (1:1,000), Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
T2192 116.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Cyanine3 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Cyanine3
  • Catalog # A10521
  • Size 500 µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10521 139.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, RPE conjugate for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M30204
  • Size 500µL
Applications
Flow Cytometry (1-2.5 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M30204 199.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, Cy3® conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M30010
  • Size 1mL
Applications
Immunofluorescence (2.5-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M30010 138.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16078SAMPLE
  • Size 50 µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16078SAMPLE 50.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16078
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16078 149.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16072
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16072 109.00
Check your price
  • Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18811
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18811 109.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16066
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16066 109.00
Check your price
  • Goat anti-Human IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18805
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18805 109.00
Check your price
  • Goat anti-Llama IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Camelid
  • Conjugate HRP
  • Catalog # A16060
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16060 199.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 32430
  • Size 2 mL
Applications
ELISA (1:60-1:500), Immunohistochemistry (1:6-1:60), Western Blot (1:60-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
32430 109.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31431
  • Size 0.2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31431 109.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31430
  • Size 2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31430 179.00
Check your price
  • Goat anti-Human IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, ICC and IHC
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # 31410
  • Size 2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31410 149.00
Check your price
  • Goat anti-Rat IgG F(ab')2 Secondary Antibody, HRP conjugate for Western Blot, ICC and IHC
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # 31474
  • Size 2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31474 149.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Cyanine5 conjugate for IF
  • Host Goat
  • Target Species Mouse
  • Conjugate Cyanine5
  • Catalog # A10524
  • Size 500 µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10524 129.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A16169
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16169 157.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A16020
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16020 200.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16079
  • Size 1 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunocytochemistry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16079 104.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16073
  • Size 2 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16073 104.00
Check your price
  • Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18812
  • Size 2 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18812 115.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate AP
  • Catalog # A16051
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16051 200.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A16163
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16163 157.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A16014
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16014 137.00
Check your price
  • Rabbit anti-Human IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate AP
  • Catalog # A18906
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18906 139.00
Check your price
  • Rabbit anti-Llama IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Camelid
  • Conjugate AP
  • Catalog # A16157
  • Size 500 µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16157 220.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16067
  • Size 2 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16067 94.00
Check your price
  • Goat anti-Human IgG (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18806
  • Size 2 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18806 85.00
Check your price
  • Goat anti-Llama IgG (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Camelid
  • Conjugate FITC
  • Catalog # A16061
  • Size 1 mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16061 179.00
Check your price