Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  90  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  130  140  150  160  170  180  190  200  210  300  400  500  600  700  800  900  1000  1100  1630 
3361 - 3390 of 48882 products displayed
  • Rabbit anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # 31329
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31329 173.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate AP
  • Catalog # 31360
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31360 181.00
Check your price
  • Goat anti-Human IgG (H+L) Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # 31529
  • Size 2mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31529 113.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16166
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16166 109.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16017
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16017 179.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A15984
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15984 Not Orderable Online
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16083
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16083 106.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16077
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16077 83.00
Check your price
  • Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18816
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18816 85.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # A16047
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16047 179.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16160
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16160 109.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16011
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16011 109.00
Check your price
  • Rabbit anti-Human IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18903
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18903 109.00
Check your price
  • Rabbit anti-Llama IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Camelid
  • Conjugate HRP
  • Catalog # A16154
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16154 199.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate AP
  • Catalog # A15978
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15978 226.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16071
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16071 92.00
Check your price
  • Goat anti-Human IgG (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18810
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18810 74.25
Check your price
  • Goat anti-Llama IgG (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Camelid
  • Conjugate TRITC
  • Catalog # A16065
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16065 179.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Donkey
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # A16041
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16041 109.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31450
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31450 179.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 31480
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31480 149.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A15981
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15981 149.00
Check your price
  • Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16082
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16082 136.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16076
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16076 138.00
Check your price
  • Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18815
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18815 138.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Rabbit
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16167
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16167 152.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Donkey
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16018
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16018 142.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Donkey
  • Target Species Sheep
  • Conjugate FITC
  • Catalog # A16049
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16049 139.00
Check your price
  • Chicken anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Chicken
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A15975
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15975 179.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16070
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16070 106.00
Check your price