Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
3931 - 3960 of 48882 products displayed
  • HA Tag Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight 800 4X PEG
  • Catalog # 26183D800
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
26183D800 159.00
Check your price
  • Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Sheep
  • Target Species Rabbit
  • Conjugate
  • Catalog # A16176
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16176 Not Orderable Online
Check your price
  • Chicken anti-Rabbit IgG (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Chicken
  • Target Species Rabbit
  • Conjugate
  • Catalog # A15988
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15988 Not Orderable Online
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Donkey
  • Target Species Rabbit
  • Conjugate TRITC
  • Catalog # A16028
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16028 139.00
Check your price
  • Rabbit anti-Feline IgG (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Rabbit
  • Target Species Cat
  • Conjugate
  • Catalog # A18882
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18882 Not Orderable Online
Check your price
  • Rabbit anti-Bovine IgG (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Rabbit
  • Target Species Bovine
  • Conjugate
  • Catalog # A24445
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24445 Not Orderable Online
Check your price
  • Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16039
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16039 200.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16033
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16033 199.00
Check your price
  • Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Chicken
  • Target Species Rabbit
  • Conjugate TRITC
  • Catalog # A15998
  • Size 500µg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15998 101.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A16027
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16027 157.00
Check your price
  • Rabbit anti-Feline IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Rabbit
  • Target Species Cat
  • Conjugate
  • Catalog # A18881
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18881 Not Orderable Online
Check your price
  • Chicken anti-Rabbit IgG (H+L) Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Chicken
  • Target Species Rabbit
  • Conjugate TRITC
  • Catalog # A15992
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15992 161.00
Check your price
  • Rabbit anti-Bovine IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Biotin
  • Catalog # A24444
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24444 139.00
Check your price
  • Rabbit anti-Goat IgG F(ab')2 Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Goat
  • Conjugate Biotin
  • Catalog # 31753
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31753 170.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, PE-Cy5.5® conjugate for Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate
  • Catalog # L42018
  • Size 500µL
Applications
Flow Cytometry (1-2.5 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
L42018 Not Orderable Online
Check your price
  • Chicken anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Chicken
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # A15997
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15997 136.00
Check your price
  • Chicken anti-Rabbit IgG (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Chicken
  • Target Species Rabbit
  • Conjugate
  • Catalog # A15991
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15991 Not Orderable Online
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, Rhodamine conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate Rhodamine
  • Catalog # 31670
  • Size 2mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31670 142.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate AP
  • Catalog # A24540
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24540 146.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate AP
  • Catalog # A24534
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24534 125.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A10547
  • Size 500µg
Applications
ELISA (1:500 to 1:2,000), Immunohistochemistry (1:500 to 1:2,000), Western Blot (1:500 to 1:2,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10547 169.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, RPE conjugate for Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate
  • Catalog # A10542
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10542 249.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A24537
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24537 149.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # A24531
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24531 109.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 525 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Qdot® 525
  • Catalog # Q22074
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22074 199.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 565 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Qdot® 565
  • Catalog # Q22077
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22077 200.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 585 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate
  • Catalog # Q22080
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22080 199.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 605 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate
  • Catalog # Q22083
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22083 199.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 625 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Qdot® 625
  • Catalog # Q22086
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22086 198.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 655 conjugate for Western Blot, ICC, IHC and Flow
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Qdot® 655
  • Catalog # Q22089
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q22089 198.00
Check your price