Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
4141 - 4170 of 48877 products displayed
  • Proinsulin Monoclonal Antibody for IHC, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122708
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay-Dependent), Radioimmune Assays (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122708 309.00
Check your price
  • Proinsulin Monoclonal Antibody for IF, ICC, IHC (F), IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122710
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Radioimmune Assays (Assay-dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122710 309.00
Check your price
  • Proinsulin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121668
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121668 349.00
Check your price
  • Proinsulin Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121667
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121667 309.00
Check your price
  • C-Peptide Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121662
  • Size 100µg
Applications
ELISA (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121662 309.00
Check your price
  • Insulin Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA135525
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135525 375.00
Check your price
  • Insulin/Proinsulin Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Guinea Pig
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA136023
  • Size 50µL
Applications
Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:1000-1:5000)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136023 1,000.00
Check your price
  • Insulin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA183252
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183252 309.00
Check your price
  • Insulin Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512032
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
proinsulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512032 249.00
Check your price
  • Insulin Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512037
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml)
Alternate names:
proinsulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512037 249.00
Check your price
  • C-Peptide Monoclonal Antibody for ICC, IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119159
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (2-5 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119159 309.00
Check your price
  • Insulin Monoclonal Antibody, Biotin conjugate for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512029
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
proinsulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512029 309.00
Check your price
  • IMPDH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527792
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
IMPDH-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527792 349.00
Check your price
  • IMPDH1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544146
  • Size 100µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
IMPDH-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544146 309.00
Check your price
  • IL-27 p28 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710175
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
710175 309.00
Check your price
  • IL-2 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710146
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710146 309.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523691
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523691 375.00
Check your price
  • IL-1 beta Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523597
  • Size 100Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523597 289.00
Check your price
  • IL-1 beta Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523546
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523546 239.00
Check your price
  • IL-1 beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523657
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523657 349.00
Check your price
  • Mouse anti-Human Lambda Light Chain Secondary Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512410
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512410 225.00
Check your price
  • Mouse anti-Human IgA Secondary Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # SA172510
  • Size 100µg
Applications
ELISA (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172510 188.00
Check your price
  • IGF1 (rat) for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP8604
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8604 301.00
Check your price
  • IGF1 (mouse) for Ctrl
  • Host E. coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP8603
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Somatomedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8603 269.00
Check your price
  • Limitin Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547798
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (1.5-7.5 ng/ml), Western Blot (0.1 µg/ml)
Alternate names:
Limitin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547798 349.00
Check your price
  • IFN alpha Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710299
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
710299 309.00
Check your price
  • Perlecan Monoclonal Antibody for IF, ICC and IHC (F)
  • Host Rat
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA134860
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (1:200)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134860 375.00
Check your price
  • Perlecan Monoclonal Antibody for IF, IHC (F) and IHC (P)
  • Host Rat
  • Target Species Bovine, Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA514641
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1-2 µg/ml)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514641 349.00
Check your price
  • Perlecan Monoclonal Antibody, Biotin conjugate for IF, IHC (F) and IHC (P)
  • Host Rat
  • Target Species Bovine, Human, Mouse, Pig
  • Conjugate Biotin
  • Catalog # MA514638
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1-2 µg/ml)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514638 399.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Bovine, Fish, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106821
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:25-1:200), Immunohistochemistry (Frozen) (1:25-1:200), Immunohistochemistry (Paraffin) (1:25-1:200), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106821 375.00
Check your price