Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  170  180  190  200  210  220  230  240  300  400  500  600  700  800  900  1000  1100  1630 
4411 - 4440 of 48877 products displayed
  • UNC13B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522159
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Munc13-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522159 349.00
Check your price
  • UNC13B Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522153
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Munc13-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522153 349.00
Check your price
  • TYK2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515303
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
JTK1; TYK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515303 349.00
Check your price
  • Tyrosine Hydroxylase Oligoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710982
  • Size 100µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (1- 2 µg/ml)
Alternate names:
Tyrosine 3-hydroxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710982 309.00
Check your price
  • TGF beta-3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Sheep
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # PA112642
  • Size 250µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
TGF-beta3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112642 325.00
Check your price
  • TCL6 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524335
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
TNG1, TNG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524335 349.00
Check your price
  • STX6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530366
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
syntaxin-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530366 349.00
Check your price
  • STX6 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514858
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:2000)
Alternate names:
syntaxin-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514858 275.00
Check your price
  • STX2 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Fruit fly, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA125502
  • Size 50µg
Applications
Immunohistochemistry (Assay-dependent), Immunoprecipitation (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Epimorphin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125502 399.00
Check your price
  • STAT6 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534814
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534814 349.00
Check your price
  • STAT6 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138675
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:75)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138675 375.00
Check your price
  • STAT6 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517673
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517673 275.00
Check your price
  • STAT6 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532603
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:75)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532603 275.00
Check your price
  • STAT6 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515659
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515659 349.00
Check your price
  • Phospho-STAT6 (Thr645) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537745
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537745 309.00
Check your price
  • Phospho-STAT6 (Thr645) Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536690
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536690 349.00
Check your price
  • Phospho-STAT6 (Tyr641) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA517902
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517902 309.00
Check your price
  • Phospho-STAT6 (Tyr641) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514920
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514920 309.00
Check your price
  • Phospho-STAT6 (Tyr641) Polyclonal Antibody, APC conjugate for Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate APC
  • Catalog # PA546901
  • Size 100Tests
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546901 Not Orderable Online
Check your price
  • Phospho-STAT6 (Tyr641) Polyclonal Antibody, PE conjugate for Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate PE
  • Catalog # PA546892
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
IL-4 Stat
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546892 279.00
Check your price
  • Syntaphilin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520529
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
bA314N13.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520529 349.00
Check your price
  • Syntaphilin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520528
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (2-4 ug/ml)
Alternate names:
bA314N13.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520528 349.00
Check your price
  • Syntaphilin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520825
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
bA314N13.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520825 309.00
Check your price
  • RPL14 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521363
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CAG-ISL 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521363 349.00
Check your price
  • RPH3A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA1774
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Exophilin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1774 349.00
Check your price
  • RAP1GAP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537034
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
ARPP-90
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537034 309.00
Check your price
  • Paxillin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA534910
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:1000-1:10,000)
Alternate names:
hCG_1778014
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534910 349.00
Check your price
  • Phospho-Paxillin (Tyr118) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Chicken, Human, Mouse, Non-human primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523327
  • Size 20ug
Applications
Western Blot (0.025-0.1 µg/ml)
Alternate names:
hCG_1778014
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523327 349.00
Check your price
  • PKC zeta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517589
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
nPKC-zeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517589 275.00
Check your price
  • PKC zeta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513756
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
nPKC-zeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513756 349.00
Check your price