Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  50  60  70  80  90  100  110  120  130  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  170  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1630 
4561 - 4590 of 48877 products displayed
  • Caldesmon HMW Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511626
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:200-1:400), Western Blot (1:100-1:1000)
Alternate names:
caldesmon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511626 309.00
Check your price
  • Caldesmon LMW Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA511774
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
caldesmon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511774 375.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128301
  • Size 50µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128301 375.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515198
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515198 349.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118375
  • Size 100µL
Applications
Western Blot (1:500-1:2,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118375 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118364
  • Size 100µL
Applications
ELISA (1:200-1:2000), Immunohistochemistry (1:200-1:2000), Inhibition Assays (1:10-1:50), Western Blot (1:200-1:2000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118364 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118357
  • Size 100µL
Applications
ELISA (1:1,000), Immunohistochemistry (1:1,000), Inhibition Assays (1:10-1:50), Western Blot (1:1,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118357 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118363
  • Size 500µg
Applications
ELISA (1-10 µg/ml), Immunohistochemistry (1-10 µg/ml), Inhibition Assays (1-10 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118363 349.00
Check your price
  • BDNF Polyclonal Antibody for IHC and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118370
  • Size 20µg
Applications
ELISA (1-10 µg/ml), Immunohistochemistry (1-10 µg/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118370 309.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118369
  • Size 500µg
Applications
ELISA (0.1 µ/ml), Immunohistochemistry (1-10 µ/ml), Inhibition Assays (1-10 µ/ml), Western Blot (1-10 µ/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118369 375.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118372
  • Size 500µg
Applications
ELISA (1-10 µg/ml), Immunohistochemistry (1-10 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118372 375.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118371
  • Size 100µL
Applications
ELISA (1:1,000), Immunohistochemistry (1:1000-1:2000), Western Blot (1:1,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118371 349.00
Check your price
  • proBDNF Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118374
  • Size 20µg
Applications
Immunohistochemistry (Frozen) (1-5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118374 309.00
Check your price
  • proBDNF Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118360
  • Size 100µL
Applications
Immunohistochemistry (1:1,000-1:5,000), Western Blot (1:1,000-1:5,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118360 349.00
Check your price
  • BDNF Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523665
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523665 349.00
Check your price
  • ATAD3B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521160
  • Size 100ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
AAA-TOB3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521160 349.00
Check your price
  • AIB1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515898
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
AIB, AIB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515898 349.00
Check your price
  • Rat Albumin Polyclonal Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA129266
  • Size 1mg
Applications
ELISA (1:200-1:30,000), Western Blot (1:1000-1:5000)
Alternate names:
Alb1, Albza
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129266 309.00
Check your price
  • Mouse Albumin Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA130899
  • Size 1mg
Applications
ELISA (1:200-1:500), Western Blot (1:5000-1:10000)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130899 219.00
Check your price
  • Mouse Albumin Polyclonal Antibody for Western Blot, IF, ICC, IDF and IE
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA128339
  • Size 2mL
Applications
Immunocytochemistry (Assay-dependent), Immunodiffusion (Assay dependent), Immunoelectrophoresis (Assay dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128339 Not Orderable Online
Check your price
  • Rat Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Sheep
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA129267
  • Size 1mg
Applications
ELISA (1:20000-1:75000), Western Blot (1:1000-1:5000)
Alternate names:
Alb1, Albza
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129267 309.00
Check your price
  • Mouse Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA130126
  • Size 1mg
Applications
ELISA (1:15,000-1:50,000), Western Blot (1:1,000-1:5,000)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130126 249.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116889
  • Size 100µg
Applications
Western Blot (1:5000-1:15,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116889 375.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP, ELISA and Neu
  • Host Rabbit
  • Target Species Avian, Fish, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA146296
  • Size 50µL
Applications
Blocking Assay (Assay-Dependent), ELISA (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Neutralization (Assay-Dependent), Western Blot (1:100-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146296 375.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121167
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121167 399.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Leech, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516914
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (2-4 µg/ml)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516914 349.00
Check your price
  • beta Actin Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA180163
  • Size 50µg
Applications
Flow Cytometry (1:10), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180163 249.00
Check your price
  • beta Actin Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA180729
  • Size 500µg
Applications
ELISA (1:10), Flow Cytometry (1:10), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:100-1:500)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180729 399.00
Check your price
  • beta Actin Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516410
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516410 225.00
Check your price
  • beta Actin Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515452
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515452 349.00
Check your price