Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  50  60  70  80  90  100  110  120  130  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1605 
4621 - 4650 of 48136 products displayed
  • Goat anti-Rabbit IgG F(ab')2, FITC
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # 31573
  • Size 2 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31573 141.00
Check your price
  • Goat anti-Rabbit IgG F(ab')2, Biotin
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # 31823
  • Size 2 mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31823 157.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG, NANOGOLD
  • Host Goat
  • Target Species Mouse
  • Conjugate NANOGOLD
  • Catalog # N24915
  • Size 1 mL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
N24915 430.00
Check your price
  • Goat anti-Mouse IgG, Alexa Fluor® 488-5 nm colloidal gold
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A31560
  • Size 500µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31560 206.00
Check your price
  • Goat anti-Mouse IgG, Alexa Fluor® 488-10 nm colloidal gold
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488-10 nm colloidal gold
  • Catalog # A31561
  • Size 500 µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31561 209.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG, Alexa Fluor® 594-FluoroNanogold
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594-FluoroNanogold
  • Catalog # A24921
  • Size 1 mL
Applications
Immunomicroscopy (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24921 526.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG, Alexa Fluor® 488-FluoroNanogold
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488-FluoroNanogold
  • Catalog # A24920
  • Size 1 mL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24920 530.00
Check your price
  • Vinculin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA514325
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514325 349.00
Check your price
  • Vinculin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA519842
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519842 349.00
Check your price
  • Vinculin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529688
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529688 349.00
Check your price
  • Vinculin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532639
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532639 349.00
Check your price
  • Vinculin Antibody (Monoclonal, SP117)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516424
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516424 499.00
Check your price
  • Vinculin Antibody (Monoclonal, VLN01)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA511690
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511690 349.00
Check your price
  • Phospho-Vinculin (Tyr821) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538546
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538546 309.00
Check your price
  • VAMP8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535300
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Endobrevin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535300 349.00
Check your price
  • TPD52 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530261
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Protein N8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530261 349.00
Check your price
  • TPD52 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518796
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Protein N8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518796 349.00
Check your price
  • Spinesin Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131504
  • Size 50 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131504 399.00
Check your price
  • Spinesin Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547223
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547223 349.00
Check your price
  • Spinesin Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546992
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546992 375.00
Check your price
  • Spinesin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531485
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531485 349.00
Check your price
  • Spinesin Antibody (Monoclonal, 376620)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524118
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524118 275.00
Check your price
  • Spinesin Antibody (Monoclonal, 282324)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523983
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523983 275.00
Check your price
  • Spinesin Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518013
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518013 349.00
Check your price
  • TMEM222 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525054
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
C1orf160
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525054 349.00
Check your price
  • Thrombomodulin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521924
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Fetomodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521924 349.00
Check your price
  • Thrombomodulin Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547550
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Fetomodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547550 375.00
Check your price
  • Thrombomodulin Antibody (Monoclonal, 501733)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524214
  • Size 100 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
Fetomodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524214 275.00
Check your price
  • Thrombomodulin Antibody (Monoclonal, 461714)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524181
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Frozen) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Fetomodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524181 275.00
Check your price
  • Thrombomodulin Antibody (Monoclonal, QBEND-40)
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA190642
  • Size 50 µL
Applications
Flow Cytometry (Neat (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (1:10-1:100), Immunohistochemistry (Paraffin) (1:10-1:50)
Alternate names:
Fetomodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190642 225.00
Check your price