Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1630 
4951 - 4980 of 48881 products displayed
  • Mouse anti-Rat IgG1 Secondary Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA133298
  • Size 100tests
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133298 304.00
Check your price
  • Mouse anti-Rat IgG1 Secondary Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # SA125262
  • Size 125µg
Applications
ELISA (0.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA125262 234.00
Check your price
  • Donkey anti-Goat IgG Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Donkey
  • Target Species Goat
  • Conjugate Biotin
  • Catalog # PA128663
  • Size 1mg
Applications
ELISA (1:140,000), Immunohistochemistry (Frozen) (1:1000-1:5000), Immunohistochemistry (Paraffin) (1:1000-1:5000), Immunomicroscopy (Assay-dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128663 208.00
Check your price
  • Mouse anti-Human IgG2 Secondary Antibody, Biotin conjugate for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 053540
  • Size 500µL
Applications
ELISA (1:500-1:1,000), Immunofluorescence (1:500-1:1,000), Immunohistochemistry (Paraffin) (1:500-1:1,000), Immunoprecipitation (1:500-1:1,000), Western Blot (1:500-1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
053540 129.00
Check your price
  • Bovine anti-Mouse IgG Secondary Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Bovine
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # SA19507
  • Size 500µg
Applications
ELISA (1:5000-1:20000), Western Blot (1:5000-1:40000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19507 154.00
Check your price
  • Bovine anti-Human IgG Secondary Antibody, Biotin conjugate for Western Blot, ICC, IHC and ELISA
  • Host Bovine
  • Target Species Human
  • Conjugate Biotin
  • Catalog # SA19506
  • Size 500µg
Applications
ELISA (1:1000-1:5000), Immunocytochemistry (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (1:5000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA19506 156.00
Check your price
  • Donkey anti-Goat IgG Secondary Antibody, Texas Red® conjugate for IF, ICC, Flow and ELISA
  • Host Donkey
  • Target Species Goat
  • Conjugate Texas Red®
  • Catalog # PA128662
  • Size 1mg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128662 196.00
Check your price
  • Vaccinia Virus H1L Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA511112
  • Size 400µL
Applications
Western Blot (1:100-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511112 349.00
Check your price
  • Goat anti-Mouse IgG3 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21151
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21151 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21121
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21121 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 546 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 546
  • Catalog # A21123
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21123 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 555
  • Catalog # A21127
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21127 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A21124
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21124 149.00
Check your price
  • Goat anti-Mouse IgG3 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A21155
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21155 149.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A21125
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21125 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 633
  • Catalog # A21126
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21126 149.00
Check your price
  • Goat anti-Mouse IgG1 Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 647
  • Catalog # A21240
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21240 169.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG Secondary Antibody, NANOGOLD conjugate for IB and IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate NANOGOLD
  • Catalog # N24916
  • Size 1mL
Applications
Immunoblot (0.2-10 µg/mL), Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
N24916 431.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, PE conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # P21129
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21129 176.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, Alexa Fluor® 488-5 nm colloidal gold conjugate for IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 488-5 nm colloidal gold
  • Catalog # A31565
  • Size 500µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31565 209.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, Alexa Fluor® 488-10 nm colloidal gold conjugate for IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 488-10 nm colloidal gold
  • Catalog # A31566
  • Size 500µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31566 209.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG Secondary Antibody, Alexa Fluor® 594-FluoroNanogold™ conjugate for IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate
  • Catalog # A24923
  • Size 1mL
Applications
Immunomicroscopy (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24923 526.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG Secondary Antibody, Alexa Fluor® 488-FluoroNanogold™ conjugate for IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate
  • Catalog # A24922
  • Size 1mL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24922 530.00
Check your price
  • 14-3-3 eta Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Dog, Human, Rabbit, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA546811
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546811 309.00
Check your price
  • 14-3-3 eta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528113
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528113 349.00
Check your price
  • 14-3-3 eta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529774
  • Size 100µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529774 349.00
Check your price
  • WEE1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515385
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Wee1A kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515385 349.00
Check your price
  • WEE1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529303
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Wee1A kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529303 349.00
Check your price
  • WEE1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126429
  • Size 50µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Wee1A kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126429 349.00
Check your price
  • Phospho-WEE1 (Ser123) Polyclonal Antibody for IHC and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512718
  • Size 400µL
Applications
Dot blot (1:500), Immunohistochemistry (1:50-1:100)
Alternate names:
Wee1A kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512718 Not Orderable Online
Check your price