Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1611 
4981 - 5010 of 48315 products displayed
  • Phospho-DNA-PK (Ser2612) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538476
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538476 309.00
Check your price
  • Phospho-DNA-PK (Thr2647) Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538475
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538475 309.00
Check your price
  • Phospho-DNA-PK (Thr2609) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512913
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512913 349.00
Check your price
  • Phospho-DNA-PK (Thr2609) Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129541
  • Size 50 µg
Applications
ELISA (1:300,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129541 349.00
Check your price
  • DNA-PK Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and GS
  • Host Mouse
  • Target Species Chicken, Hamster, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA513404
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513404 349.00
Check your price
  • DNA-PK Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA513235
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513235 399.00
Check your price
  • DNA-PK Monoclonal Antibody for IF, IHC (P) and GS
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513249
  • Size 500 µL
Applications
Gel Shift (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513249 425.00
Check your price
  • PPP4R1L Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA539447
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
C20orf192
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539447 309.00
Check your price
  • POLK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA543223
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
DINB protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543223 309.00
Check your price
  • PODXL2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542988
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542988 309.00
Check your price
  • PODXL2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521335
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521335 349.00
Check your price
  • PODXL2 Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547502
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547502 349.00
Check your price
  • PODXL2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547150
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547150 349.00
Check your price
  • PODXL2 Monoclonal Antibody for IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523932
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523932 275.00
Check your price
  • PMCH Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525442
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
MCH, pro-MCH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525442 349.00
Check your price
  • PHR1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA519042
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Evectin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519042 349.00
Check your price
  • PKP1 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 325700
  • Size 100 µg
Applications
Immunofluorescence (3-10 ug/ml), Immunoprecipitation (2 ug), Western Blot (1-3 ug/ml)
Alternate names:
Band 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
325700 309.00
Check your price
  • PGA5 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515558
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Pepsinogen-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515558 349.00
Check your price
  • PFDN6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512450
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein Ke2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512450 349.00
Check your price
  • PFDN6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Yeast, Zebrafish, Sheep, Rabbit, Bovine, Dog, Horse, Guinea pig, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA543805
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Protein Ke2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543805 309.00
Check your price
  • PDE6D Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522008
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein p17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522008 349.00
Check your price
  • PAX6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518713
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Oculorhombin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518713 349.00
Check your price
  • ORM1 Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19530
  • Size 500 ug
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Orosomucoid-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19530 225.00
Check your price
  • Opsin 3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00216W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Panopsin; Encephalopsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00216W 309.00
Check your price
  • Opsin 3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00213W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Panopsin; Encephalopsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00213W 309.00
Check your price
  • Olfr632 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00200W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Mor 5'Beta5, Olfr632
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00200W 309.00
Check your price
  • Olfr631 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00199W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
MOR 5'Beta4, Olfr631
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00199W 309.00
Check your price
  • NUMB Polyclonal Antibody for Western Blot, IF and IHC
  • Host Goat
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131840
  • Size 50 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Protein S171
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131840 399.00
Check your price
  • Numb Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA517362
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Protein S171
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517362 259.00
Check your price
  • NUMB Polyclonal Antibody for Western Blot, IF, ICC, IP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518000
  • Size 100 ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Protein S171
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518000 349.00
Check your price