Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1611 
5041 - 5070 of 48310 products displayed
  • Kallikrein 15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14211
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
ACO protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14211 309.00
Check your price
  • Shootin1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517167
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Shootin1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517167 275.00
Check your price
  • JunD Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1834
  • Size 200 µg
Applications
ChIP assay (1-3 ul), Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (10 µg/ml)
Alternate names:
Oncogene Jun-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1834 275.00
Check your price
  • JSRP1 Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Dog, Guinea pig, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3933
  • Size 100 µL
Applications
Immunofluorescence (1:250), Western Blot (1:1,500)
Alternate names:
Junctional SR
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3933 349.00
Check your price
  • JAK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44406G
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Janus kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44406G 309.00
Check your price
  • Phospho-JAK2 (Tyr1007, Tyr1008) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44426G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Janus kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44426G 304.00
Check your price
  • Phospho-JAK1 (Tyr1022, Tyr1023) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44422G
  • Size 10 blots
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (Assay Dependent)
Alternate names:
Janus kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44422G 306.00
Check your price
  • IL-9 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547584
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1.5-7.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547584 349.00
Check your price
  • Bovine IL-6 for Western Blot, IP and ELISA
  • Host Yeast
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # RBOIL6I
  • Size 20 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Bovine Interleukin 6 IL6
Catalog # Size Price (AUD) Your Price (AUD) Qty
RBOIL6I 302.00
Check your price
  • IL-25 Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # PA186925
  • Size 50 µg
Applications
ELISA (0.25-1.0 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Il25, Ly6elg
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186925 499.00
Check your price
  • hnRNP M Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA134962
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (3:25), Western Blot (1:1000)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134962 399.00
Check your price
  • hnRNP M Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA134981
  • Size 200 µL
Applications
Immunoprecipitation (3:25), Western Blot (1:500)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134981 425.00
Check your price
  • hnRNP M3-M4 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA116674
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:1000)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116674 375.00
Check your price
  • hnRNP M1-M4 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA116673
  • Size 200 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116673 309.00
Check your price
  • hnRNP M Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA191607
  • Size 50 µL
Applications
Immunoprecipitation (3:25), Western Blot (1:500)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191607 275.00
Check your price
  • HMOX1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527338
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
P32 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527338 349.00
Check your price
  • GPS2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530953
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
AMF-1, GPS-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530953 349.00
Check your price
  • GNB4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSG00022W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Transducin beta chain 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSG00022W 309.00
Check your price
  • GNB3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSG00021W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Transducin beta chain 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSG00021W 309.00
Check your price
  • GNB1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSG00020W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Transducin beta chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSG00020W 309.00
Check your price
  • GMNN Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524596
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Gem, geminin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524596 349.00
Check your price
  • GMNN Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530612
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Gem, geminin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530612 349.00
Check your price
  • CD32 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # CD3204
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RII-a; CDw32
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD3204 268.00
Check your price
  • CD32 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # CD3201
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RII-a; CDw32
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD3201 288.00
Check your price
  • CD64 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547249
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547249 349.00
Check your price
  • CD64 Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523993
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523993 275.00
Check your price
  • Factor X Polyclonal Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA143061
  • Size 1 mg
Applications
ELISA (Assay Dependent.), Western Blot (25 ug/ml)
Alternate names:
Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA143061 149.00
Check your price
  • Factor X Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA143036
  • Size 100 ug
Applications
ELISA (Assay Dependent.), Western Blot (5 ug/ml)
Alternate names:
Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143036 309.00
Check your price
  • Factor X Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA143035
  • Size 100 µg
Applications
ELISA (Assay Dependent.), Western Blot (5 ug/ml)
Alternate names:
Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143035 309.00
Check your price
  • EME1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527110
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MMS4 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527110 349.00
Check your price