Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1630 
5701 - 5730 of 48877 products displayed
  • Cyclin D3 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112298
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RP5-973N23.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112298 309.00
Check your price
  • Cyclin C Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514981
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
SRB11 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514981 349.00
Check your price
  • Cyclin C Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516227
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (10µg/mg protein lysate), Western Blot (5-10 µg/mL)
Alternate names:
SRB11 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516227 309.00
Check your price
  • Phospho-Cyclin C (Ser275) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537510
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
SRB11 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537510 309.00
Check your price
  • Phospho-Caveolin 2 (Ser36) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125626
  • Size 25µg
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
CAV, MGC12294
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125626 399.00
Check your price
  • Phospho-Caveolin 2 (Ser23) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125625
  • Size 25µg
Applications
Western Blot (0.1-0.5 µg/ml)
Alternate names:
CAV, MGC12294
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125625 399.00
Check your price
  • C6orf1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA546354
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Protein LBH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546354 309.00
Check your price
  • Phospho-beta Arrestin 1 (Ser412) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44200
  • Size 10blots
Applications
Western Blot (0.1-1.0 ug/ml)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44200 309.00
Check your price
  • ARHGEF19 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539420
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Ephexin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539420 309.00
Check your price
  • ARHGEF16 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539419
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Ephexin-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539419 309.00
Check your price
  • ARHGEF15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA531818
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Ephexin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531818 349.00
Check your price
  • Fe65 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146191
  • Size 100µL
Applications
Immunoprecipitation (2-10 µg), Western Blot (1:1000)
Alternate names:
Protein Fe65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146191 309.00
Check your price
  • FE65 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517432
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Protein Fe65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517432 275.00
Check your price
  • Fe65 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518560
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein Fe65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518560 349.00
Check your price
  • A20A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539563
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
ANKRD20A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539563 309.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120178
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
AGPT2, ANG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120178 349.00
Check your price
  • Angiopoietin 2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129640
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500)
Alternate names:
AGPT2, ANG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129640 375.00
Check your price
  • Angiogenin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120177
  • Size 100µg
Applications
ELISA (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Ribonuclease 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120177 309.00
Check your price
  • ADAMTSL1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528408
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Punctin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528408 349.00
Check your price
  • ADAMTSL1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531013
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Punctin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531013 349.00
Check your price
  • beta Actin Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # MA1140A488
  • Size 50µl
Applications
Western Blot (1:500)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140A488 225.00
Check your price
  • beta Actin Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Alexa Fluor® 555
  • Catalog # MA1140A555
  • Size 50µl
Applications
Western Blot (Assay-Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140A555 225.00
Check your price
  • beta Actin Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # MA1140A647
  • Size 50µl
Applications
Western Blot (Assay-Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140A647 225.00
Check your price
  • Acrosomal Vesicle Protein Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # MA119208
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Msa63, SP-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119208 225.00
Check your price
  • Srb4p Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # PA14933
  • Size 250µL
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
YER022W, MED17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14933 1,400.00
Check your price
  • Srb4p Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # PA14932
  • Size 50µL
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
YER022W, MED17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14932 550.00
Check your price
  • RT1.A Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA170005
  • Size 250ug
Applications
Flow Cytometry (1 ug/1 million cells)
Alternate names:
rat class I A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170005 199.00
Check your price
  • Enterovirus 71 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA532202
  • Size 100µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
EV71
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532202 349.00
Check your price
  • Glucose Oxidase Polyclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA
  • Host Goat
  • Target Species Fungi
  • Conjugate
  • Catalog # PA126994
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
GOx
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126994 Not Orderable Online
Check your price
  • Enterovirus 71 Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516317
  • Size 100ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
EV71
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516317 349.00
Check your price