Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  40  50  60  70  80  90  100  110  200  300  400  500  600  700  800  900  1000  1630 
571 - 600 of 48882 products displayed
  • MMP21 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA125234
  • Size 50µg
Applications
Western Blot (1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125234 375.00
Check your price
  • MNK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544608
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544608 309.00
Check your price
  • Ki-67 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121520
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (10-20 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121520 399.00
Check your price
  • MITD1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543864
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543864 309.00
Check your price
  • C20orf72 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA546304
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546304 309.00
Check your price
  • MFAP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544593
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544593 309.00
Check your price
  • MEF2C Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517119
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517119 309.00
Check your price
  • MCTP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543044
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543044 309.00
Check your price
  • MCTP1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542572
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542572 309.00
Check your price
  • MCTP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542573
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542573 309.00
Check your price
  • MAML3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543079
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543079 309.00
Check your price
  • Lys20/Lys21 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA190371
  • Size 250µL
Applications
Immunocytochemistry (1:1000-1:10000), Immunofluorescence (1:1000-1:10,000), Western Blot (1:10,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190371 349.00
Check your price
  • LYPD5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA544431
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544431 309.00
Check your price
  • Ly-6C Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA516666
  • Size 100ug
Applications
Flow Cytometry (Neat-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516666 275.00
Check your price
  • Phospho-LRRK2 (Ser935) Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710097
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
710097 304.00
Check your price
  • LRRK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513868
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513868 349.00
Check your price
  • LIMK2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521987
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521987 349.00
Check your price
  • LIMK2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521871
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521871 349.00
Check your price
  • LIMD1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543594
  • Size 100µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543594 309.00
Check your price
  • LENG1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546522
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546522 309.00
Check your price
  • LENG1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Horse, Human
  • Conjugate Unconjugated
  • Catalog # PA546521
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546521 309.00
Check your price
  • LCN12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA544898
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544898 309.00
Check your price
  • KTI12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545752
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545752 309.00
Check your price
  • KTI12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA545751
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545751 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Amphibian, Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512231
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:10-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512231 225.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191326
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191326 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513203
  • Size 500µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1-2 ug/ml), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513203 219.00
Check your price
  • KLHL7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544515
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544515 309.00
Check your price
  • KIF2B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540811
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540811 309.00
Check your price
  • KIF14 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126592
  • Size 50µL
Applications
Immunoprecipitation (1-4 µg/mg of lysate), Western Blot (1:2000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126592 349.00
Check your price