Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6331 - 6360 of 48882 products displayed
  • STIM1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146217
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:1000)
Alternate names:
D11S4896E, GOK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146217 375.00
Check your price
  • STIM1 Monoclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA119451
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
D11S4896E, GOK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119451 309.00
Check your price
  • STAT5 beta Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515665
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
STAT5; STAT5B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515665 349.00
Check your price
  • SRPRB Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543390
  • Size 100µl
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (0.2-1 µg/ml)
Alternate names:
Protein APMCF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543390 309.00
Check your price
  • SPOCK1 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548008
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Protein SPOCK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548008 309.00
Check your price
  • SPOCK1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524039
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein SPOCK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524039 275.00
Check your price
  • SOX14 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532595
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Protein SOX-28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532595 349.00
Check your price
  • SOX12 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538525
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Protein SOX-22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538525 309.00
Check your price
  • SOX12 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519999
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Protein SOX-22
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519999 309.00
Check your price
  • SNX7 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530623
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
sorting nexin-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530623 349.00
Check your price
  • SNX5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528526
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
sorting nexin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528526 349.00
Check your price
  • SNX5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531206
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
sorting nexin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531206 349.00
Check your price
  • SMEK1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518784
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
SMEK homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518784 349.00
Check your price
  • SERPINA10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522100
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Serpin A10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522100 349.00
Check your price
  • SERPINA10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518954
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Serpin A10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518954 349.00
Check your price
  • SEMG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA542099
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Semenogelin II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542099 309.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522804
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522804 349.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522803
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522803 349.00
Check your price
  • CD100 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA134040
  • Size 100µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134040 309.00
Check your price
  • CD100 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA516559
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516559 600.00
Check your price
  • CD100 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate APC
  • Catalog # MA516558
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516558 399.00
Check your price
  • SCUBE2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA520991
  • Size 100ug
Applications
Immunocytochemistry (20 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Protein CEGP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520991 349.00
Check your price
  • SBSN Polyclonal Antibody for Western Blot, IHC, Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA525531
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
UNQ698, HLAR698
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525531 Not Orderable Online
Check your price
  • SAA Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140181
  • Size 100µg
Applications
ELISA (1:10), Immunohistochemistry (Paraffin, Frozen) (1:10), Western Blot (1:10)
Alternate names:
Serum Amyloid-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140181 600.00
Check your price
  • SAA Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA140180
  • Size 100µg
Applications
ELISA (1:10), Immunohistochemistry (Frozen) (1:10), Immunohistochemistry (Paraffin) (1:10), Western Blot (1:10)
Alternate names:
Serum Amyloid-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140180 600.00
Check your price
  • FAM82B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542837
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Protein FAM82B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542837 309.00
Check your price
  • RIC8A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521855
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein Ric-8A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521855 349.00
Check your price
  • RIC8A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518316
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Protein Ric-8A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518316 349.00
Check your price
  • RhoC Monoclonal Antibody for AGG
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # MA17670
  • Size 100ug
Applications
Agglutination (Assay Dependent)
Alternate names:
Rho cDNA clone 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17670 309.00
Check your price
  • RhoA Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1134
  • Size 100ug
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (1:500-1:2000)
Alternate names:
Rho cDNA clone 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1134 309.00
Check your price