Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6451 - 6480 of 48882 products displayed
  • HDAC3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529026
  • Size 100µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:1000), Western Blot (1:500-1:3000)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529026 349.00
Check your price
  • HDAC3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141039
  • Size 100µg
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1:10), Immunofluorescence (1:10), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (2-5 µg/ml)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141039 309.00
Check your price
  • HDAC3 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518069
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:400-1:800)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518069 399.00
Check your price
  • HDAC3 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515443
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515443 349.00
Check your price
  • HDAC3 Monoclonal Antibody for Western Blot, IF, ICC and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515442
  • Size 100µL
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515442 349.00
Check your price
  • GSR Monoclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA182826
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Gr-1, Gr1, Ly-6G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182826 199.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533739
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (4-32 µg/ml)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533739 349.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533740
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533740 349.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533741
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (4-21 µg/ml)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533741 349.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533738
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533738 349.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120472
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120472 399.00
Check your price
  • GPR56 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA133156
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Protein TM7XN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133156 399.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # OPA106100
  • Size 100µL
Applications
Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA106100 375.00
Check your price
  • Glucagon Polyclonal Antibody for IF, IHC (F), IHC (P) and RIA
  • Host Sheep
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA185465
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
GLP1, GLP2, GRPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185465 275.00
Check your price
  • Glicentin Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA136077
  • Size 250µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50)
Alternate names:
GLP1, GLP2, GRPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136077 425.00
Check your price
  • FUBP1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541103
  • Size 100µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
DNA helicase V
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541103 309.00
Check your price
  • FUBP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528061
  • Size 100µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
DNA helicase V
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528061 349.00
Check your price
  • FUBP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530291
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
DNA helicase V
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530291 349.00
Check your price
  • FREM1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520981
  • Size 100ug
Applications
Immunocytochemistry (20 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Protein QBRICK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520981 349.00
Check your price
  • FGF15 Polyclonal Antibody for IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547870
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml)
Alternate names:
FGF-19, FGF-19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547870 349.00
Check your price
  • CD105 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A16390
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16390 108.00
Check your price
  • ECSIT Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535160
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein SITPEC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535160 349.00
Check your price
  • ECSIT Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520082
  • Size 100ug
Applications
Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protein SITPEC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520082 349.00
Check your price
  • DLL1 Monoclonal Antibody, PE conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18523
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Drosophila Delta homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18523 116.00
Check your price
  • DKK3 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515601
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Western Blot (1/500-1/2000)
Alternate names:
RIG; REIC; DKK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515601 349.00
Check your price
  • CYP3A7 Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MA3034
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (2µg per 10^6 cells), Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (1:250)
Alternate names:
CP37, P450-HFLA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3034 309.00
Check your price
  • Cullin 3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521511
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
CUL-3, cullin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521511 349.00
Check your price
  • Cullin 2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532371
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
CUL-2, cullin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532371 275.00
Check your price
  • Cullin 1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532370
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
CUL-1, cullin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532370 275.00
Check your price
  • CTHRC1 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538054
  • Size 100µg
Applications
Immunofluorescence (20 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Protein NMTC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538054 309.00
Check your price