Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1605 
6511 - 6540 of 48148 products displayed
  • WNT10B Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534315
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10-15 µg/ml)
Alternate names:
Protein Wnt-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534315 349.00
Check your price
  • WNT10B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520530
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Protein Wnt-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520530 349.00
Check your price
  • WNT10B Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515557
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Protein Wnt-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515557 349.00
Check your price
  • Vezatin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530652
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
VEZATIN, vezatin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530652 349.00
Check your price
  • Vezatin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530651
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
VEZATIN, vezatin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530651 349.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA126000
  • Size 50µL
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:100), Western Blot (1:2000)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126000 399.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517608
  • Size 200ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517608 325.00
Check your price
  • UBAP2L Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539520
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein NICE-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539520 309.00
Check your price
  • UBAP2L Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536998
  • Size 100µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Protein NICE-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536998 309.00
Check your price
  • TUBE1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544635
  • Size 100µl
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Epsilon-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544635 309.00
Check your price
  • TUBE1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA120249
  • Size 100µL
Applications
ELISA (Assay dependent), Western Blot (2-4 µg/ml)
Alternate names:
Epsilon-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120249 399.00
Check your price
  • TUBA8 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517193
  • Size 100µL
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Alpha-tubulin 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517193 309.00
Check your price
  • TUBA8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529135
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Alpha-tubulin 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529135 349.00
Check your price
  • TSPAN32 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131554
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Protein Phemx
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131554 399.00
Check your price
  • TSPAN32 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542904
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (5 µg/ml)
Alternate names:
Protein Phemx
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542904 309.00
Check your price
  • TSPAN32 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518045
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Protein Phemx
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518045 349.00
Check your price
  • 53BP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species Fish, Goat, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116565
  • Size 100µL
Applications
Flow Cytometry (1.5 µg/ml), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:400-1:2000), Western Blot (1:5000-1:25,000)
Alternate names:
TP53BP1, p202
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116565 375.00
Check your price
  • 53BP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116566
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:50-1:1000), Immunofluorescence (1:50-1:1000), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:10,000)
Alternate names:
TP53BP1, p202
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116566 325.00
Check your price
  • TMEM109 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543429
  • Size 100µl
Applications
Western Blot (1.25 µg/ml)
Alternate names:
Mitsugumin-23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543429 309.00
Check your price
  • TLR3 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18430
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18430 192.00
Check your price
  • THRSP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548103
  • Size 100µg
Applications
Western Blot (0.5 µg/ml)
Alternate names:
Spot 14 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548103 309.00
Check your price
  • Trans-Golgi Network 38 Polyclonal Antibody for Western Blot, IF, IHC (F) and IM
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184496
  • Size 100µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1/100-1/200), Immunomicroscopy (Assay Dependent), Western Blot (1/1000)
Alternate names:
TGN38B; TGN38A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184496 550.00
Check your price
  • Phospho-AS160 (Thr642) Oligoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710105
  • Size 100µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (2-5 ug/ml)
Alternate names:
Akt substrate of 160 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
710105 304.00
Check your price
  • TBC1D3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538731
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
PRC17, TBC1D3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538731 309.00
Check your price
  • TAS1R3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00181W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Sweet taste receptor T1R3
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00181W 309.00
Check your price
  • Synaptotagmin 6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0113
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:2000)
Alternate names:
Synaptotagmin VI
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0113 309.00
Check your price
  • Synaptotagmin 6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531281
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Synaptotagmin VI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531281 349.00
Check your price
  • SYNGR4 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520879
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
synaptogyrin-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520879 349.00
Check your price
  • SYNGR3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520878
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
synaptogyrin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520878 349.00
Check your price
  • STRIP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525058
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein FAM40B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525058 349.00
Check your price