Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1622 
6721 - 6750 of 48655 products displayed
  • NPS Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3212
  • Size 100 µL
Applications
Immunocytochemistry (1:5000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:2000)
Alternate names:
Neuropeptide S; NPS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3212 309.00
Check your price
  • NDUFS1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522309
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Complex I-75kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522309 349.00
Check your price
  • NDUFS1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519341
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Complex I-75kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519341 349.00
Check your price
  • NDUFS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528220
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:10,000)
Alternate names:
Complex I-75kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528220 349.00
Check your price
  • FUS2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521588
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Protein fusion-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521588 349.00
Check your price
  • FUS2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131810
  • Size 100 µg
Applications
ELISA (1:32,000), Western Blot (Assay dependent)
Alternate names:
Protein fusion-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131810 375.00
Check your price
  • FUS2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530495
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein fusion-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530495 349.00
Check your price
  • MYLK4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525869
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Sugen kinase 85
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525869 349.00
Check your price
  • MOXD1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531526
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Monooxygenase X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531526 349.00
Check your price
  • MOXD1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531527
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Monooxygenase X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531527 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511518
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511518 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520636
  • Size 100 ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520636 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520635
  • Size 100 ug
Applications
Western Blot (2 µg/ml)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520635 349.00
Check your price
  • Myoglobin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA516809
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:1000-1:2000)
Alternate names:
OTTHUMP00000197936
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516809 249.00
Check your price
  • MAP1B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530584
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MAP1.2; MAP1(X)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530584 349.00
Check your price
  • LZTS2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544945
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Protein LAPSER1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544945 309.00
Check your price
  • Ly-6G Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA183934
  • Size 100 Tests
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
Gr-1, Gr1, Ly-6G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183934 159.00
Check your price
  • LIM2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA543463
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
MP20; MP19; MP18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543463 309.00
Check your price
  • LEPROTL1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA546034
  • Size 100 µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Endospanin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546034 309.00
Check your price
  • KLHL22 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515699
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
KELCHL; KLHL22
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515699 349.00
Check your price
  • KIF4A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528142
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Chromokinesin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528142 349.00
Check your price
  • KIF4A Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530492
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Chromokinesin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530492 349.00
Check your price
  • KIF4A Polyclonal Antibody for Western Blot and IF
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518059
  • Size 100 ug
Applications
Immunofluorescence (Assay Dependent), Western Blot (Assay-dependent)
Alternate names:
Chromokinesin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518059 349.00
Check your price
  • Jagged2 Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514978
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
protein jagged-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514978 275.00
Check your price
  • Jagged2 Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514979
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
protein jagged-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514979 275.00
Check your price
  • ITGB6 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547588
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547588 349.00
Check your price
  • ITGB6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547309
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547309 349.00
Check your price
  • ITGB6 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA535903
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535903 309.00
Check your price
  • ITGB6 Monoclonal Antibody for IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524161
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (8-25 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524161 275.00
Check your price
  • ITGB6 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523968
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523968 275.00
Check your price