Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1622 
6841 - 6870 of 48655 products displayed
  • CDH4 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511433
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Retinal cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511433 349.00
Check your price
  • P-cadherin Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # 324000
  • Size 100 µg
Applications
Immunofluorescence (2-10 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Placental cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
324000 305.00
Check your price
  • P-cadherin Monoclonal Antibody for Western Blot, IF, IHC (F), IP, ELISA and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 135800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Placental cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
135800 330.00
Check your price
  • CCR3 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18365
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Eosinophil eotaxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18365 108.00
Check your price
  • CCR3 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A18388
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Eosinophil eotaxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18388 Not Orderable Online
Check your price
  • CCD92 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524939
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Limkain beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524939 349.00
Check your price
  • CCDC130 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524672
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
9 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524672 349.00
Check your price
  • CALHM3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Horse, Human
  • Conjugate Unconjugated
  • Catalog # PA544479
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Protein FAM26A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544479 309.00
Check your price
  • CALHM1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520895
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Protein FAM26C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520895 349.00
Check your price
  • Bcl-X Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # AHO0222
  • Size 500 µl
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apoptosis regulator Bcl-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0222 304.00
Check your price
  • Bcl-xL Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A15375
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Apoptosis regulator Bcl-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15375 318.00
Check your price
  • VISTA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547032
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein L259
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547032 399.00
Check your price
  • BarX1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540622
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
BARX homeobox 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540622 309.00
Check your price
  • AXIN1 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710204
  • Size 100 µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Axis inhibition protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710204 309.00
Check your price
  • SERCA3 ATPase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527691
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Calcium pump 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527691 349.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547063
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (0.1-0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ENOVIN, EVN, NBN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547063 349.00
Check your price
  • beta-Arrestin 2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523296
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.25-0.5 µg/ml)
Alternate names:
Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523296 349.00
Check your price
  • beta-Arrestin 2 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518375
  • Size 100 µg
Applications
Immunohistochemistry (4-6 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (0.3-1 ug/ml)
Alternate names:
Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518375 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535089
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535089 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519582
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519582 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519583
  • Size 100 µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunoprecipitation (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519583 349.00
Check your price
  • Phospho-beta Arrestin 1 (Ser412) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538259
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538259 309.00
Check your price
  • Phospho-beta Arrestin 1 (Ser412) Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515217
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515217 309.00
Check your price
  • ARHGDIG Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519354
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
Rho-GDI gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519354 349.00
Check your price
  • Angiopoietin 4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534780
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Angiopoietin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534780 349.00
Check your price
  • Angiopoietin 4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534646
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Angiopoietin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534646 349.00
Check your price
  • ADSL Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529964
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Adenylosuccinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529964 349.00
Check your price
  • Adrenomedullin Polyclonal Antibody for IF, ICC and IHC (F)
  • Host Guinea Pig
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA136016
  • Size 50 µL
Applications
Immunofluorescence (1:500), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Ap, H39316, RATAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136016 900.00
Check your price
  • Septin-8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531356
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
2-Sep, septin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531356 349.00
Check your price
  • Rattin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA146257
  • Size 100 µL
Applications
Western Blot (1:500)
Alternate names:
Humanin homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146257 309.00
Check your price