Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1605 
7681 - 7710 of 48136 products displayed
  • Apolipoprotein C2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127196
  • Size 100 µL
Applications
ELISA (1:40,000-1:160,000), Immunohistochemistry (Assay-Dependent), Western Blot (1:40,000-1:160,000)
Alternate names:
Apolipoprotein C2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127196 309.00
Check your price
  • Apolipoprotein C2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA122561
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein C2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122561 349.00
Check your price
  • Apolipoprotein C1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518706
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Apolipoprotein C1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518706 349.00
Check your price
  • Apolipoprotein C1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA122560
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein C1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122560 349.00
Check your price
  • Apolipoprotein C1 Antibody, HRP
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA129013
  • Size 100 µL
Applications
ELISA (Assay dependent), Western Blot (1:1000-1:3000)
Alternate names:
Apolipoprotein C1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129013 309.00
Check your price
  • Apolipoprotein A2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535301
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535301 349.00
Check your price
  • Apolipoprotein A2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130491
  • Size 1 mL
Applications
ELISA (1:80,000-1:400,000), Western Blot (Assay dependent)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130491 425.00
Check your price
  • Apolipoprotein A2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126900
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126900 375.00
Check your price
  • Apolipoprotein A2 Antibody, HRP
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA128972
  • Size 100 µL
Applications
ELISA (1:1500-1:6000), Western Blot (1:1500-1:6000)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128972 309.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129508
  • Size 50 µg
Applications
Western Blot (1:1500-1:2500)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129508 425.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514310
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514310 349.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520721
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520721 349.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # PA521166
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521166 349.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 5F4F5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515391
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515391 349.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529557
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529557 349.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA122559
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (Paraffin) (1:250-1:500), Western Blot (1:500-1:1000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122559 349.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Hamster, Human
  • Conjugate Unconjugated
  • Catalog # PA127156
  • Size 1 mL
Applications
Immunodiffusion (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127156 475.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA123059
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123059 375.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA122558
  • Size 100 µg
Applications
ELISA (1:10,000-1:20,000), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122558 375.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519784
  • Size 100 µg
Applications
Immunofluorescence (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519784 349.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, G2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183002
  • Size 500 µg
Applications
ELISA (1:5000), Immunohistochemistry (Frozen) (1:40), Western Blot (1/2000-1/8,000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183002 399.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 2G4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0127
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0127 375.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 1402)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123058
  • Size 100 µg
Applications
ELISA (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123058 375.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 532)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514732
  • Size 100 ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514732 349.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 513)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514670
  • Size 100 ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514670 349.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 311)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514668
  • Size 100 ug
Applications
ELISA (1µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514668 349.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 6001)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514667
  • Size 100 ug
Applications
ELISA (1µg/ml), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514667 349.00
Check your price
  • Apolipoprotein A1 Antibody, HRP
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA128965
  • Size 100 µL
Applications
ELISA (1:4000-1:32,000), Western Blot (1:4000-1:32,000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128965 309.00
Check your price
  • Alivin 1 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14357
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Alivin-1; AMIGO-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14357 309.00
Check your price
  • AGL Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512142
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Glycogen debrancher
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512142 349.00
Check your price