Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1630 
7951 - 7980 of 48877 products displayed
  • Phospho-Cardiac Troponin I pThr142 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535713
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535713 Not Orderable Online
Check your price
  • Phospho-Cardiac Troponin I (Ser150) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535410
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535410 349.00
Check your price
  • Phospho-Cardiac Troponin I (Ser22, Ser23) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540251
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540251 309.00
Check your price
  • Phospho-Cardiac Troponin I (Ser22, Ser23) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538341
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538341 309.00
Check your price
  • Phospho-Cardiac Troponin I (Ser22, Ser23) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA122702
  • Size 200µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122702 349.00
Check your price
  • Phospho-Cardiac Troponin I (Ser23, Ser24) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535411
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535411 349.00
Check your price
  • TMEM147 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521894
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein NIFIE 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521894 349.00
Check your price
  • Talin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527646
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ILWEQ, TLN, talin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527646 349.00
Check your price
  • Tyrosine Hydroxylase Polyclonal Antibody for Western Blot, IF, IHC and DB
  • Host Sheep
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA14679
  • Size 100µL
Applications
Dot blot (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (1:1000), Western Blot (1:1000)
Alternate names:
Tyrosine 3-hydroxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14679 375.00
Check your price
  • TAAR2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00115W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
G-protein coupled receptor 58
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00115W 309.00
Check your price
  • TAAR2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00080W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
G-protein coupled receptor 58
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00080W 309.00
Check your price
  • SYT13 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525396
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Synaptotagmin XIII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525396 349.00
Check your price
  • SYT13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532101
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Synaptotagmin XIII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532101 349.00
Check your price
  • Synaptotagmin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14699
  • Size 100µL
Applications
Western Blot (1/1000)
Alternate names:
p65; Synaptotagmin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14699 375.00
Check your price
  • Phospho-Synaptotagmin 1 (Thr202) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14642
  • Size 100µL
Applications
Immunohistochemistry (1:400), Western Blot (1:1000)
Alternate names:
p65; Synaptotagmin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14642 375.00
Check your price
  • Phospho-Synaptotagmin 1 (Ser309) Polyclonal Antibody for Western Blot, IHC and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14641
  • Size 100µL
Applications
Dot blot (1:1000), Immunohistochemistry (1:400), Western Blot (1:1000)
Alternate names:
p65; Synaptotagmin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14641 309.00
Check your price
  • SPT3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538548
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
SPT3-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538548 309.00
Check your price
  • SPT3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536134
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
SPT3-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536134 309.00
Check your price
  • SPT3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540315
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.4 µg/ml)
Alternate names:
SPT3-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540315 309.00
Check your price
  • STX12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSS00028W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
STX12, STX13, STX14, MGC51957
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSS00028W 309.00
Check your price
  • STRC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA535991
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000)
Alternate names:
DFNB16, stereocilin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535991 Not Orderable Online
Check your price
  • ST7L Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535289
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
ST7-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535289 349.00
Check your price
  • SRC Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1152
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-3 µg/ml)
Alternate names:
pp60c-src; Proto-oncogene c-Src
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1152 309.00
Check your price
  • SRC (active) Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO0051
  • Size 50µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1.0 ug/ml)
Alternate names:
pp60c-src; Proto-oncogene c-Src
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0051 303.00
Check your price
  • SRC Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701396
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Western Blot (1-3 µg/ml)
Alternate names:
pp60c-src; Proto-oncogene c-Src
Catalog # Size Price (AUD) Your Price (AUD) Qty
701396 309.00
Check your price
  • Sclerostin Polyclonal Antibody for IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537943
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (4-6 µg/ml)
Alternate names:
sclerosteosis, VBCH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537943 309.00
Check your price
  • Sclerostin Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546977
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
sclerosteosis, VBCH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546977 375.00
Check your price
  • Sclerostin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547134
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
sclerosteosis, VBCH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547134 349.00
Check your price
  • Sclerostin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523897
  • Size 100µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
sclerosteosis, VBCH
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523897 275.00
Check your price
  • SNRK Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522167
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SNF1-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522167 349.00
Check your price