Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1605 
7981 - 8010 of 48136 products displayed
  • ITGA9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527771
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Integrin alpha-RLC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527771 349.00
Check your price
  • ISL1 Antibody (Monoclonal, 1B1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515516
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Isl-1; ISLET1; ISL1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515516 349.00
Check your price
  • ISL1 Antibody (Monoclonal, 1H9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515515
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Isl-1; ISLET1; ISL1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515515 349.00
Check your price
  • Phospho-IRF3 (Ser396) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 720012
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Interferon regulatory factor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
720012 319.00
Check your price
  • IMMP2L Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA526991
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
IMP2-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526991 349.00
Check your price
  • IMMP1L Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524322
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
IMP1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524322 349.00
Check your price
  • IL-27 p28 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141055
  • Size 100 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141055 309.00
Check your price
  • IL-27 p28 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA124498
  • Size 50 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124498 375.00
Check your price
  • IL-2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175205
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175205 1
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546957
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.15-0.75 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546957 375.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA546923
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546923 375.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA546915
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.75-3.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546915 375.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA547850
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.25-0.8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547850 349.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Cat
  • Conjugate Unconjugated
  • Catalog # PA547217
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.03-0.15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547217 349.00
Check your price
  • IL-2 Antibody (Monoclonal, 8D5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517097
  • Size 100 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517097 275.00
Check your price
  • IL-2 Antibody (Monoclonal, 5355)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523696
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523696 375.00
Check your price
  • IL-2 Antibody (Monoclonal, JES6-IAI2)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523798
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.125-0.75 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523798 349.00
Check your price
  • IL-2 Antibody, PE (Monoclonal, 5334)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523618
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523618 309.00
Check your price
  • IL-2 Antibody, FITC (Monoclonal, 5334)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523580
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523580 279.00
Check your price
  • Bovine IL-1 beta Recombinant Protein
  • Host Yeast
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # RBOIL1BI
  • Size 20 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Bovine Interleukin 1 beta IL1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
RBOIL1BI 313.00
Check your price
  • IL-13 Antibody (Monoclonal, A130D12G51E4)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ARC0132
  • Size 500 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell activation protein P600
Catalog # Size Price (AUD) Your Price (AUD) Qty
ARC0132 303.00
Check your price
  • IL-13 Antibody (Monoclonal, 21H6H5)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 701123
  • Size 100 µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
T-cell activation protein P600
Catalog # Size Price (AUD) Your Price (AUD) Qty
701123 309.00
Check your price
  • Human IL-11 Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL1110
  • Size 10 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL1110 309.00
Check your price
  • RABL5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539390
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Rab-like protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539390 309.00
Check your price
  • Interferon alpha-2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521926
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521926 349.00
Check your price
  • Interferon alpha-2b Antibody Polyclonal (PA1-28126)
  • Host Chicken
  • Target Species Human
  • Conjugate
  • Catalog # PA128126
  • Size 50µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128126 439.00
Check your price
  • Interferon alpha-2 Antibody (Monoclonal, N39)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135514
  • Size 100 µg
Applications
Blocking Assay (Assay Dependent), ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135514 309.00
Check your price
  • Interferon alpha-2 Antibody (Monoclonal, KT5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA172505
  • Size 100 µg
Applications
ELISA (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA172505 199.00
Check your price
  • Interferon alpha-2 Antibody (Monoclonal, N27)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135513
  • Size 100 µg
Applications
ELISA (Assay dependent.), Inhibition Assays (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135513 309.00
Check your price
  • Interferon alpha-2 Antibody (Monoclonal, 17G6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517263
  • Size 100 µL
Applications
Immunoprecipitation (2 ug), Western Blot (0.1 µg/ml)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517263 349.00
Check your price