Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1630 
8131 - 8160 of 48877 products displayed
  • Hemopexin Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529521
  • Size 100µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Beta-1B-glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529521 349.00
Check your price
  • Hemopexin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534799
  • Size 100µL
Applications
Western Blot (Assay dependent)
Alternate names:
Beta-1B-glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534799 349.00
Check your price
  • HMX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540574
  • Size 100µl
Applications
Western Blot (1:1000)
Alternate names:
Homeobox protein H6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540574 309.00
Check your price
  • HMOX1 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA and IA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125475
  • Size 50µg
Applications
ELISA (1:10,000), Immunohistochemistry (Frozen) (5-10 µg/ml), Immunohistochemistry (Paraffin) (5-10 µg/ml), Inhibition Assays (Assay Dependent), Western Blot (5-10 µg/ml)
Alternate names:
P32 protein; HSP32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125475 399.00
Check your price
  • HMGB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16617
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
High mobility group protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16617 Not Orderable Online
Check your price
  • HGF Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701283
  • Size 100µg
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Scatter factor; Hepatopoietin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
701283 309.00
Check your price
  • HEPH Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542974
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
HEPH, CPL, KIAA0698
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542974 309.00
Check your price
  • HAL Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA529123
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
HIS, HSTD, histidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529123 349.00
Check your price
  • GSS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537307
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Glutathione synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537307 399.00
Check your price
  • GPSM2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518646
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Mosaic protein LGN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518646 349.00
Check your price
  • Glypican 2 Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548007
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cerebroglycan, GPC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548007 309.00
Check your price
  • Glypican 2 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548010
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cerebroglycan, GPC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548010 309.00
Check your price
  • Glypican 2 Monoclonal Antibody for Western Blot and IHC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524012
  • Size 100µg
Applications
Immunohistochemistry (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cerebroglycan, GPC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524012 275.00
Check your price
  • GP1BB Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522267
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Antigen CD42b-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522267 349.00
Check your price
  • GnRH1 Polyclonal Antibody for IHC (F), IHC (P), ELISA and RIA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA136121
  • Size 250µL
Applications
ELISA (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (1:50), Radioimmune Assays (Assay dependent.)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136121 375.00
Check your price
  • GnRH1 Monoclonal Antibody for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110816
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110816 349.00
Check your price
  • GnRH1 Monoclonal Antibody for IHC, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110814
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110814 399.00
Check your price
  • GnRH1 Monoclonal Antibody for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110815
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110815 349.00
Check your price
  • GnRH1 Monoclonal Antibody for IHC, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110817
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110817 375.00
Check your price
  • GFR alpha-4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539188
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Persephin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539188 309.00
Check your price
  • GFR alpha-4 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547182
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Persephin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547182 349.00
Check your price
  • Phospho-GATA4 (Ser105) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 44948
  • Size 10blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-1.0 ug/ml)
Alternate names:
GATA-binding factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
44948 309.00
Check your price
  • Prothrombin Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127519
  • Size 50µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127519 425.00
Check your price
  • Thrombin Polyclonal Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA143040
  • Size 1mg
Applications
ELISA (10 ug/ml), Western Blot (10 ug/ml)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA143040 199.00
Check your price
  • Prothrombin Polyclonal Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA143062
  • Size 1mg
Applications
ELISA (Assay Dependent.), Western Blot (25 ug/ml)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA143062 119.00
Check your price
  • Prothrombin Polyclonal Antibody for Western Blot and ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA143039
  • Size 1mg
Applications
ELISA (10 µg/ml), Western Blot (10 µg/ml)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA143039 159.00
Check your price
  • Thrombin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA143019
  • Size 100µg
Applications
ELISA (Assay Dependent.)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143019 225.00
Check your price
  • Prothrombin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA143020
  • Size 100ug
Applications
ELISA (Assay Dependent.), Inhibition Assays (Assay Dependent.), Western Blot (5 ug/ml)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143020 225.00
Check your price
  • Prothrombin Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA143034
  • Size 100ug
Applications
ELISA (Assay Dependent.), Western Blot (Assay Dependent)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143034 309.00
Check your price
  • Prothrombin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517629
  • Size 200ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Coagulation factor II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517629 325.00
Check your price