Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1630 
841 - 870 of 48882 products displayed
  • ZNF578 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA543573
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543573 309.00
Check your price
  • ZNF575 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539567
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539567 309.00
Check your price
  • ZNF573 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541220
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541220 309.00
Check your price
  • ZNF572 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524264
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524264 349.00
Check your price
  • ZNF571 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531499
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531499 349.00
Check your price
  • ZNF570 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541209
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541209 309.00
Check your price
  • ZNF567 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531310
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531310 349.00
Check your price
  • ZNF567 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531309
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531309 349.00
Check your price
  • ZNF566 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524049
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524049 349.00
Check your price
  • ZNF564 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA541211
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541211 309.00
Check your price
  • ZNF563 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA544866
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544866 309.00
Check your price
  • ZNF563 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA543634
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543634 309.00
Check your price
  • ZNF560 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA541224
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541224 309.00
Check your price
  • ZNF559 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541184
  • Size 100µl
Applications
Western Blot (5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541184 309.00
Check your price
  • ZNF559 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541185
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541185 309.00
Check your price
  • ZNF557 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA541175
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541175 309.00
Check your price
  • ZNF554 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530907
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530907 349.00
Check your price
  • ZNF550 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530912
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530912 349.00
Check your price
  • ZNF549 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541681
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541681 309.00
Check your price
  • ZNF548 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541233
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541233 309.00
Check your price
  • ZNF543 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530932
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530932 349.00
Check your price
  • ZNF541 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538788
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538788 309.00
Check your price
  • ZNF540 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541229
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541229 309.00
Check your price
  • ZNF536 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524338
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524338 349.00
Check your price
  • ZNF530 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532092
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532092 349.00
Check your price
  • ZNF528 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541182
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541182 309.00
Check your price
  • ZNF514 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531307
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531307 349.00
Check your price
  • ZNF491 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541219
  • Size 100µl
Applications
Western Blot (0.125 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541219 309.00
Check your price
  • ZNF480 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA543714
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543714 309.00
Check your price
  • ZNF454 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541250
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541250 309.00
Check your price