Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  70  80  90  100  110  120  130  140  200  300  400  500  600  700  800  900  1000  1630 
1351 - 1380 of 48881 products displayed
  • Rabbit anti-Mouse IgM Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 31196
  • Size 2mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31196 173.00
Check your price
  • Platelet Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517901
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517901 249.00
Check your price
  • Platelet Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517900
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517900 119.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24460
  • Size 1mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24460 139.00
Check your price
  • Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18802
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18802 214.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18790
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18790 169.00
Check your price
  • Goat anti-Human IgE Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18796
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18796 215.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18784
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18784 157.00
Check your price
  • Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18799
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18799 279.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18787
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18787 149.00
Check your price
  • Goat anti-Human IgE Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18793
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18793 179.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18781
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18781 109.00
Check your price
  • Goat anti-Mouse IgM Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31440
  • Size 2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31440 209.00
Check your price
  • Goat anti-Human IgM Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # 31415
  • Size 2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31415 199.00
Check your price
  • Goat anti-Mouse IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M31101
  • Size 500µL
Applications
Flow Cytometry (1:80-1:320), Immunofluorescence (1:80-1:320)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M31101 Not Orderable Online
Check your price
  • Goat anti-Mouse IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M31001
  • Size 1mL
Applications
Flow Cytometry (1:80-1:320), Immunofluorescence (1:80-1:320)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M31001 Not Orderable Online
Check your price
  • Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18800
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18800 241.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18788
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18788 118.00
Check your price
  • Goat anti-Human IgE Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18794
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18794 157.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18782
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18782 94.00
Check your price
  • Goat anti-Human IgM Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # 31575
  • Size 2mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31575 146.00
Check your price
  • Rabbit anti-Mouse IgM Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # 31333
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31333 228.00
Check your price
  • Goat anti-Mouse IgM Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31992
  • Size 2mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31992 191.00
Check your price
  • Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18804
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18804 241.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18792
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18792 118.00
Check your price
  • Goat anti-Human IgE Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18798
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18798 157.00
Check your price
  • Goat anti-Human IgA Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18786
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18786 96.00
Check your price
  • Rabbit anti-Mouse IgM Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31456
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31456 209.00
Check your price
  • Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18803
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18803 252.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18791
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18791 136.00
Check your price