Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  90  100  110  120  130  140  150  160  170  200  300  400  500  600  700  800  900  1000  1630 
2191 - 2220 of 48881 products displayed
  • Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18823
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18823 109.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # A16084
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16084 109.00
Check your price
  • Goat anti-Human IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18817
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18817 109.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31437
  • Size 2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31437 199.00
Check your price
  • Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16091
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16091 117.00
Check your price
  • Goat anti-Human IgG Fc Highly Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18830
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18830 73.25
Check your price
  • Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18824
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18824 116.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # A16085
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16085 109.00
Check your price
  • Goat anti-Human IgG Fc Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18818
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18818 94.00
Check your price
  • Rabbit anti-Human IgG Fc Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate AP
  • Catalog # 31318
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31318 194.00
Check your price
  • Rabbit anti-Mouse IgG Fc Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # 31332
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31332 194.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31547
  • Size 2mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31547 142.00
Check your price
  • Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16095
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16095 106.00
Check your price
  • Goat anti-Human IgG Fc Highly Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18834
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18834 96.00
Check your price
  • Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18828
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18828 85.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, TRITC conjugate for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16089
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16089 93.00
Check your price
  • Goat anti-Human IgG Fc Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A18822
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18822 73.00
Check your price
  • Rabbit anti-Human IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # 31423
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31423 199.00
Check your price
  • Rabbit anti-Mouse IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31455
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31455 199.00
Check your price
  • Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16094
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16094 136.00
Check your price
  • Goat anti-Human IgG Fc Highly Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18833
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18833 94.00
Check your price
  • Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18827
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18827 147.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, Biotin conjugate for Western Blot, IF, Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # A16088
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16088 139.00
Check your price
  • Goat anti-Human IgG Fc Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18821
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18821 104.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31805
  • Size 2mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31805 174.00
Check your price
  • Rabbit anti-Mouse IgG Fc Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31555
  • Size 1.5mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31555 118.00
Check your price
  • Rabbit anti-Human IgG Fc Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate FITC
  • Catalog # 31535
  • Size 1.5mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31535 133.00
Check your price
  • Lucifer Yellow Polyclonal Antibody, Biotin conjugate for ICC and IHC
  • Host Rabbit
  • Target Species Chemical
  • Conjugate Biotin
  • Catalog # A5751
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A5751 333.00
Check your price
  • Rabbit anti-Mouse IgG Fc Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31813
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31813 189.00
Check your price
  • Rabbit anti-Human IgG Fc Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 31789
  • Size 1.5mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31789 174.00
Check your price