Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1630 
3001 - 3030 of 48882 products displayed
  • FBXO27 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522811
  • Size 100ug
Applications
Western Blot (1:500-1:1000)
Alternate names:
F-box/G-domain protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522811 349.00
Check your price
  • FBXO32 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519056
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Muscle atrophy F-box protein; Atrogin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519056 349.00
Check your price
  • FBXO43 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518188
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (5-10µg/ml), Immunofluorescence (5-10ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Endogenous meiotic inhibitor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518188 349.00
Check your price
  • FBXO44 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518187
  • Size 100ug
Applications
Western Blot (2-6.0ug/ml)
Alternate names:
F-box/G-domain protein 3; F-box protein FBX30
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518187 349.00
Check your price
  • FBXO6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110884
  • Size 100ug
Applications
Western Blot (1-5 ug/ml)
Alternate names:
F-box/G-domain protein 2; F-box protein that recognizes sugar chains 2; F-box only protein 6b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110884 309.00
Check your price
  • FBXO6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522810
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
F-box/G-domain protein 2; F-box protein that recognizes sugar chains 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522810 349.00
Check your price
  • FBXO9 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523474
  • Size 100ug
Applications
Immunocytochemistry (5µg/ml), Immunofluorescence (5µg/ml), Western Blot (4-6 µg/ml)
Alternate names:
Renal carcinoma antigen NY-REN-57; Cross-immune reaction antigen 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523474 349.00
Check your price
  • FBXW2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518189
  • Size 100ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.3-1.5 ug/ml)
Alternate names:
Protein MD6; F-box and WD-40 domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518189 349.00
Check your price
  • FceR1 alpha Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517074
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
IgE Fc receptor subunit alpha; Fc-epsilon RI-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517074 309.00
Check your price
  • FCGR2B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518362
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (3-6 µg/ml), Western Blot (0.3-1 µg/ml)
Alternate names:
Fc-gamma RII-c; CDw32; Fc-gamma RII-b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518362 349.00
Check your price
  • FCHO2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534390
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (2.5 ug/mL), Immunohistochemistry (2.5 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
FCHO2, FCH domain only 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534390 349.00
Check your price
  • Fe65 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518560
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein Fe65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518560 349.00
Check your price
  • Feline Leukemia Virus Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514707
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
FLV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514707 349.00
Check your price
  • FEM1A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518060
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Prostaglandin E receptor 4-associated protein; FEM1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518060 349.00
Check your price
  • FEM1C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517940
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
FEM1-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517940 349.00
Check your price
  • Ferritin Heavy Chain Polyclonal Antibody for Western Blot, IHC, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519058
  • Size 100ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Cell proliferation-inducing gene 15 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519058 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514733
  • Size 100ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514733 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514694
  • Size 100ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514694 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514736
  • Size 100ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514736 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514693
  • Size 100ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 0
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514693 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514735
  • Size 100ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514735 349.00
Check your price
  • Ferritin Polyclonal Antibody for Western Blot, IHC, ELISA and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519059
  • Size 100ug
Applications
ELISA (1-2ug/ml), Immunohistochemistry (3-6ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
FTL; ferritin, light polypeptide; MGC71996; L apoferritin; ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519059 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518608
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518608 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520396
  • Size 100ug
Applications
Western Blot (1 ug/ml)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520396 349.00
Check your price
  • FEZ2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520456
  • Size 100ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Zygin-2; Zygin II; Zygin-related protein types I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520456 349.00
Check your price
  • FEZ2 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520459
  • Size 100ug
Applications
Immunofluorescence (5 µg/ml), Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Zygin-2; Zygin II; Zygin-related protein types I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520459 349.00
Check your price
  • FGF20 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523480
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
FGF-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523480 349.00
Check your price
  • FGF21 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518811
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
; fibroblast growth factor 21; FGF21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518811 349.00
Check your price
  • FGF23 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518636
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Tumor-derived hypophosphatemia-inducing factor; Phosphatonin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518636 349.00
Check your price
  • FGF4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520483
  • Size 100ug
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Transforming protein KS3; Heparin-binding growth factor 4; Heparin secretory-transforming protein 1; K-fibroblast growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520483 349.00
Check your price