Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1630 
3061 - 3090 of 48877 products displayed
  • FNBP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523250
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Rapostlin; Formin-binding protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523250 349.00
Check your price
  • FNBP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518302
  • Size 100ug
Applications
Western Blot (0.25-2 ug/ml)
Alternate names:
Formin-binding protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518302 349.00
Check your price
  • FNBP1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523414
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Rapostlin; Formin-binding protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523414 349.00
Check your price
  • FNIP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520691
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
O6-methylguanine-induced apoptosis 1 protein; FNIP1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520691 349.00
Check your price
  • FNIP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520690
  • Size 100ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
O6-methylguanine-induced apoptosis 1 protein; FNIP1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520690 349.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514726
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514726 349.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for Western Blot, IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514711
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514711 349.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514710
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514710 349.00
Check your price
  • Follicle Stimulating Hormone Monoclonal Antibody for Western Blot, IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514695
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514695 349.00
Check your price
  • FOXA1 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518168
  • Size 100ug
Applications
Immunohistochemistry (5-10 µg/ml), Western Blot (0.2-0.6 ug/ml)
Alternate names:
Forkhead box protein A1; Transcription factor 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518168 349.00
Check your price
  • FOXA2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518169
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518169 349.00
Check your price
  • FOXB1 Polyclonal Antibody for IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518170
  • Size 100ug
Applications
Immunocytochemistry (0.1-0.3ug/ml), Immunofluorescence (0.1-0.3ug/ml)
Alternate names:
Transcription factor FKH-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518170 349.00
Check your price
  • FOXC1 Polyclonal Antibody for Western Blot, IHC, ChIP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA518171
  • Size 100ug
Applications
ChIP assay (Assay-dependent), Immunohistochemistry (3 µg/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
Transcription factor FKH-1; Mesoderm/mesenchyme forkhead 1; Forkhead-related transcription factor 3; Forkhead-related protein FKHL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518171 349.00
Check your price
  • FOXC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518458
  • Size 100ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
Forkhead-related transcription factor 3; Forkhead-related protein FKHL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518458 349.00
Check your price
  • FOXC2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517077
  • Size 100ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
Transcription factor FKH-14; Mesenchyme fork head protein 1; Forkhead-related protein FKHL14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517077 309.00
Check your price
  • FOXC2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518172
  • Size 100ug
Applications
Immunohistochemistry (2-4 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Transcription factor FKH-14; Mesenchyme fork head protein 1; Forkhead-related protein FKHL14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518172 349.00
Check your price
  • FOXE1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518173
  • Size 100ug
Applications
Immunohistochemistry (2-3µg/ml)
Alternate names:
Thyroid transcription factor 2; HNF-3/fork head-like protein 5; HFKH4; Forkhead-related protein FKHL15; Forkhead box protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518173 349.00
Check your price
  • FOXG1 Polyclonal Antibody for Western Blot, IF, ICC and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19043
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Forkhead-related protein FKHL3; Forkhead-related protein FKHL2; Forkhead-related protein FKHL1; Forkhead box protein G1C; Forkhead box protein G1B; Forkhead box protein G1A; Brain factor 2; Brain factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19043 309.00
Check your price
  • FOXH1 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534474
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Forkhead activin signal transducer 2; Forkhead activin signal transducer 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534474 349.00
Check your price
  • FOXI1 Polyclonal Antibody for IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518174
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4 ug/ml)
Alternate names:
Hepatocyte nuclear factor 3 forkhead homolog 3; Forkhead-related transcription factor 6; Forkhead-related protein FKHL10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518174 349.00
Check your price
  • FOXL2 Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518175
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
Pituitary forkhead factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518175 349.00
Check your price
  • FOXL2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131950
  • Size 100ug
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Pituitary forkhead factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131950 375.00
Check your price
  • FOXN1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518191
  • Size 100ug
Applications
Immunohistochemistry (20-30µg/ml)
Alternate names:
Winged-helix transcription factor nude
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518191 349.00
Check your price
  • FOXO1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517078
  • Size 100ug
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517078 309.00
Check your price
  • FOXO1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518696
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518696 349.00
Check your price
  • FOXO1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520972
  • Size 100ug
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520972 349.00
Check your price
  • FOXO1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523132
  • Size 100ug
Applications
Western Blot (0.05-0.50 µg/ml)
Alternate names:
Forkhead in rhabdomyosarcoma; Forkhead box protein O1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523132 349.00
Check your price
  • FOXO3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523191
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523191 349.00
Check your price
  • FOXO3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA131322
  • Size 100ug
Applications
ELISA (Assay-dependent), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131322 375.00
Check your price
  • FOXO3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19042
  • Size 100ug
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19042 309.00
Check your price