Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  20  30  40  50  60  70  80  90  100  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  140  150  160  170  180  190  200  210  300  400  500  600  700  800  900  1000  1100  1630 
3571 - 3600 of 48881 products displayed
  • IL-17F Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516228
  • Size 100ug
Applications
Western Blot (0.2-2 µg/ml)
Alternate names:
Cytokine ML-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516228 349.00
Check your price
  • IL-18 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519131
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Interleukin-1 gamma; Interferon gamma-inducing factor; Iboctadekin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519131 349.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519921
  • Size 100ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519921 349.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519920
  • Size 100ug
Applications
Western Blot (1:1000)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519920 349.00
Check your price
  • IL-23 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520239
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520239 349.00
Check your price
  • IL-23 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520238
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-23 subunit p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520238 349.00
Check your price
  • IL-25 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516230
  • Size 100ug
Applications
Flow Cytometry (1-2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Interleukin-17E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516230 349.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520241
  • Size 100ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520241 349.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520240
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520240 349.00
Check your price
  • IL-31 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520220
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (2.5-10 µg/ml)
Alternate names:
1700013B14Rik; IL-31; interleukin-31
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520220 349.00
Check your price
  • IL-31 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520219
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (2.5-10 µg/ml)
Alternate names:
1700013B14Rik; IL-31; interleukin-31
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520219 349.00
Check your price
  • IL-32 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523199
  • Size 100ug
Applications
Western Blot (2-5 µg/ml)
Alternate names:
Tumor necrosis factor alpha-inducing factor; Natural killer cells protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523199 349.00
Check your price
  • IL-32 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520221
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (5-10 ug/ml)
Alternate names:
Tumor necrosis factor alpha-inducing factor; Natural killer cells protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520221 349.00
Check your price
  • IL-32 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520222
  • Size 100ug
Applications
Western Blot (2.5-5 µg/ml)
Alternate names:
Tumor necrosis factor alpha-inducing factor; Natural killer cells protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520222 349.00
Check your price
  • IL-33 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515772
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515772 349.00
Check your price
  • IL-33 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516242
  • Size 100ug
Applications
Flow Cytometry (0.25 µg/0.5X10^6 cells), Western Blot (1-10 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516242 349.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523150
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523150 349.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520397
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520397 349.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520398
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520398 349.00
Check your price
  • IL-34 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520594
  • Size 100ug
Applications
Western Blot (0.25 µg/ml)
Alternate names:
C16orf77, IL-34; interleukin-34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520594 349.00
Check your price
  • IL-4 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M2IL4BI
  • Size 100ug
Applications
ELISA (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
M2IL4BI 349.00
Check your price
  • IL12RB1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519320
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
IL-12 receptor beta component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519320 349.00
Check your price
  • IL15RA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534512
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
IL-15RA, Interleukin-15 receptor subunit alpha, IL15RA, CD215
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534512 349.00
Check your price
  • IL17RA Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518746
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
interleukin 17 receptor A; AW538159; Cdw217; IL17r; VDw217;_x000D_ interleukin 17 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518746 349.00
Check your price
  • IL17RA Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534571
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
CDw217
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534571 349.00
Check your price
  • IL17RE Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534499
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
IL-17RE, Interleukin-17 receptor E, IL-17 receptor E
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534499 349.00
Check your price
  • IL21R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519983
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Novel interleukin receptor; Novel cytokine receptor 8; Lymphocyte receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519983 349.00
Check your price
  • IL21R Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519982
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Novel interleukin receptor; Novel cytokine receptor 8; Lymphocyte receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519982 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519986
  • Size 100ug
Applications
Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519986 349.00
Check your price
  • IL22RA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519987
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR11; Cytokine receptor family 2 member 9; Cytokine receptor class-II member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519987 349.00
Check your price