Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  20  30  40  50  60  70  80  90  100  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  150  160  170  180  190  200  210  220  300  400  500  600  700  800  900  1000  1100  1605 
3781 - 3810 of 48135 products displayed
  • AIM Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520242
  • Size 100 ug
Applications
Immunohistochemistry (10 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
SP-alpha; IgM-associated peptide; CT-2; Apoptosis inhibitory 6; Apoptosis inhibitor expressed by macrophages
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520242 349.00
Check your price
  • AIM2 Antibody (Monoclonal, 10M2B3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516259
  • Size 100 ug
Applications
Western Blot (0.3-2 µg/ml)
Alternate names:
Absent in melanoma 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516259 349.00
Check your price
  • AIM2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519275
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (4-6µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Absent in melanoma 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519275 349.00
Check your price
  • AIPL1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520623
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520623 349.00
Check your price
  • AIPL1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520624
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520624 349.00
Check your price
  • AIRE Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518640
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518640 349.00
Check your price
  • AIRE Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518067
  • Size 100 ug
Applications
ChIP assay (Assay Dependent), Immunohistochemistry (2.5µg/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518067 349.00
Check your price
  • AIRE Antibody
  • Host Goat
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA518525
  • Size 100 ug
Applications
Immunohistochemistry (2.5µg/ml), Western Blot (0.5-1.0 ug/ml)
Alternate names:
Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518525 349.00
Check your price
  • AIRE Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523275
  • Size 100 ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523275 349.00
Check your price
  • AKAP10 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518959
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
AKAP10; A kinase (PRKA) anchor protein 10; D-AKAP2; MGC9414; PRKA10; A-kinase anchor protein 10; dual-specificity A-kinase anchoring protein 2; mitochondrial A kinase PPKA anchor protein 10; protein kinase A anchoring protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518959 349.00
Check your price
  • AKAP3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518261
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Sperm oocyte-binding protein; Protein kinase A-anchoring protein 3; Fibrousheathin-1; Fibrousheathin I; Fibrous sheath protein of 95 kDa; Cancer/testis antigen 82; A-kinase anchor protein 110 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518261 349.00
Check your price
  • AKAP450 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518345
  • Size 100 ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Protein yotiao; Protein kinase A-anchoring protein 9; Protein hyperion; Centrosome- and Golgi-localized PKN-associated protein; AKAP 120-like protein; A-kinase anchor protein 450 kDa; A-kinase anchor protein 350 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518345 349.00
Check your price
  • AKAP8 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518265
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
A-kinase anchor protein 95 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518265 349.00
Check your price
  • Akirin1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520606
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
C1orf108, RP11-781D11.2, STRF2; akirin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520606 349.00
Check your price
  • Akirin1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520605
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
C1orf108, RP11-781D11.2, STRF2; akirin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520605 349.00
Check your price
  • Akirin2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520608
  • Size 100 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Fourteen-three-three beta interacting-protein 1; Fourteen-three-three beta interactant 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520608 349.00
Check your price
  • Akirin2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520607
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
C6orf166, FBI1, dJ486L4.2; akirin-2; fourteen-three-three beta interactant 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520607 349.00
Check your price
  • AKR1A1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518073
  • Size 100 ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Aldo-keto reductase family 1 member A1; Aldehyde reductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518073 349.00
Check your price
  • AKR1B10 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518882
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518882 349.00
Check your price
  • AKR1C3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518339
  • Size 100 ug
Applications
Western Blot (0.01-0.1 ug/ml)
Alternate names:
Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; Testosterone 17-beta-dehydrogenase 5; Prostaglandin F synthase; Indanol dehydrogenase; HA1753; Dihydrodiol dehydrogenase type I; Dihydrodiol dehydrogenase 3; Chlordecone reductase homolog HAKRb; 3-alpha-hydroxysteroid dehydrogenase type 2; 3-alpha-HSD type II, brain; 17-beta-hydroxysteroid dehydrogenase type 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518339 349.00
Check your price
  • AKR1C4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518511
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
HAKRA; Dihydrodiol dehydrogenase 4; Chlordecone reductase; 3-alpha-hydroxysteroid dehydrogenase type I; 3-alpha-HSD1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518511 349.00
Check your price
  • AKT3 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19004
  • Size 100 ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
RAC-PK-gamma; Protein kinase B gamma; Protein kinase Akt-3; STK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19004 309.00
Check your price
  • Alcohol dehydrogenase Pan Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518332
  • Size 100 ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; Alcohol dehydrogenase subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518332 349.00
Check your price
  • ALDH18A1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519392
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Aldehyde dehydrogenase family 18 member A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519392 349.00
Check your price
  • ALDH1A1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518483
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Aldehyde dehydrogenase, cytosolic; Aldehyde dehydrogenase family 1 member A1; ALHDII; ALDH-E1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518483 349.00
Check your price
  • ALDH1B1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519327
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519327 349.00
Check your price
  • ALDH2 Antibody (Monoclonal, 4G6A3)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517029
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:75), Immunofluorescence (1:75), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
ALDH1; ALDH-E2; ALDH class 2; ALDHI; AHD-M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517029 309.00
Check your price
  • ALDH3A1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519326
  • Size 100 ug
Applications
Immunohistochemistry (3-5 ug/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
Aldehyde dehydrogenase family 3 member A1; Aldehyde dehydrogenase 3; ALDHIII; Dioxin-inducible aldehyde dehydrogenase 3; Aldehyde dehydrogenase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519326 349.00
Check your price
  • ALDH3A1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520602
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Tumor-associated aldehyde dehydrogenase; HTC-ALDH; Aldehyde dehydrogenase family 3 member A1; Aldehyde dehydrogenase 3; ALDHIII; Dioxin-inducible aldehyde dehydrogenase 3; Aldehyde dehydrogenase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520602 349.00
Check your price
  • ALDH3A2 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519330
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Microsomal aldehyde dehydrogenase; Aldehyde dehydrogenase family 3 member A2; Aldehyde dehydrogenase 4; Aldehyde dehydrogenase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519330 349.00
Check your price