Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
3931 - 3960 of 48882 products displayed
  • MAVS Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520349
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Virus-induced-signaling adapter; Putative NF-kappa-B-activating protein 031N; Interferon beta promoter stimulator protein 1; CARD adapter inducing interferon beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520349 349.00
Check your price
  • MAVS Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520348
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Virus-induced-signaling adapter; Putative NF-kappa-B-activating protein 031N; Interferon beta promoter stimulator protein 1; CARD adapter inducing interferon beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520348 349.00
Check your price
  • MAX Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19067
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Myc-associated factor X; Class D basic helix-loop-helix protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19067 309.00
Check your price
  • MBD2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518619
  • Size 100ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Methyl-CpG-binding protein MBD2; Demethylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518619 349.00
Check your price
  • MBNL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519051
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Triplet-expansion RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519051 349.00
Check your price
  • MC2R Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518867
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Melanocortin receptor 2; Adrenocorticotropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518867 349.00
Check your price
  • MC3R Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518367
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
melanocortin 3 receptor; MC3R; MC3; BMIQ9; MC3-R; OB20; OQTL; Obesity quantitative trait locus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518367 349.00
Check your price
  • MCL1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515236
  • Size 100ug
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515236 349.00
Check your price
  • MCL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518280
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518280 349.00
Check your price
  • MCL1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520121
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520121 349.00
Check your price
  • MCL1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520120
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520120 349.00
Check your price
  • MCL1 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520122
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520122 349.00
Check your price
  • MCP-1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514767
  • Size 100ug
Applications
Western Blot (1/500-1/2000)
Alternate names:
Small-inducible cytokine A2; Monocyte secretory protein JE; Monocyte chemotactic protein 1; Monocyte chemotactic and activating factor; Monocyte chemoattractant protein 1; HC11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514767 349.00
Check your price
  • MCP-1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA517040
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Small-inducible cytokine A2; Platelet-derived growth factor-inducible protein JE; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1; Monocyte secretory protein JE; Monocyte chemotactic and activating factor; HC11; Immediate-early serum-responsive protein JE
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517040 309.00
Check your price
  • MCP-1 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M2MCP1BI
  • Size 100ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Small-inducible cytokine A2; Monocyte secretory protein JE; Monocyte chemotactic protein 1; Monocyte chemotactic and activating factor; Monocyte chemoattractant protein 1; HC11
Catalog # Size Price (AUD) Your Price (AUD) Qty
M2MCP1BI 349.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534505
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Small-inducible cytokine A2; Platelet-derived growth factor-inducible protein JE; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1; Monocyte secretory protein JE; Monocyte chemotactic and activating factor; HC11; Immediate-early serum-responsive protein JE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534505 349.00
Check your price
  • MCT2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517960
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Solute carrier family 16 member 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517960 349.00
Check your price
  • MD1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520253
  • Size 100ug
Applications
Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protein MD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520253 349.00
Check your price
  • MD1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520261
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Protein MD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520261 349.00
Check your price
  • MDA5 Polyclonal Antibody for Western Blot, IHC, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518164
  • Size 100ug
Applications
Immunohistochemistry (10µg/ml), Western Blot (Assay-dependent)
Alternate names:
RNA helicase-DEAD box protein 116; RIG-I-like receptor 2; Murabutide down-regulated protein; Melanoma differentiation-associated protein 5; Interferon-induced with helicase C domain protein 1; Helicase with 2 CARD domains; Clinically amyopathic dermatomyositis autoantigen 140 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518164 349.00
Check your price
  • MDA5 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520338
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
RNA helicase-DEAD box protein 116; RIG-I-like receptor 2; Murabutide down-regulated protein; Melanoma differentiation-associated protein 5; Interferon-induced with helicase C domain protein 1; Helicase with 2 CARD domains; Clinically amyopathic dermatomyositis autoantigen 140 kDa; Interferon induced with helicase C domain protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520338 349.00
Check your price
  • MDM2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518444
  • Size 100ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
p53-binding protein Mdm2; Oncoprotein Mdm2; Double minute 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518444 349.00
Check your price
  • MECR Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534490
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
Nuclear receptor-binding factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534490 349.00
Check your price
  • MED28 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520572
  • Size 100ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Tumor angiogenesis marker EG-1; Merlin and Grb2-interacting cytoskeletal protein; Mediator complex subunit 28; Endothelial-derived protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520572 349.00
Check your price
  • MED4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520571
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Mediator complex subunit 4; Vitamin D3 receptor-interacting protein complex 36 kDa component; TRAP/SMCC/PC2 subunit p36 subunit; Activator-recruited cofactor 36 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520571 349.00
Check your price
  • MEF2C Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517119
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517119 309.00
Check your price
  • MEFV Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518410
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Marenostrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518410 349.00
Check your price
  • MEIG1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521156
  • Size 100ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Protein expressed in male leptotene and zygotene spermatocytes 278
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521156 349.00
Check your price
  • MEIS1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519050
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
MEIS1; Meis homeobox 1; MGC43380; Meis1, myeloid ecotropic viral integration site 1 homolog; WUGSC:H_NH0444B04.1; leukemogenic homolog protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519050 349.00
Check your price
  • MEIS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523376
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Myeloid ecotropic viral integration site 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523376 349.00
Check your price